Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97225606-97226141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP19
Entrez Gene ID
84986
Gene Name
Rho GTPase activating protein 19
Gene Aliases
-
UniGene
Hs.80305
Gene Chromosome Location
Chr.10: 97222173-97292673 on Build GRCh38
Transcripts
NM_001204300.1 NM_001256423.1 NM_032900.5 NR_037909.1

Gene Symbol
ARHGAP19-SLIT1
Entrez Gene ID
100533184
Gene Name
ARHGAP19-SLIT1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80305
Gene Chromosome Location
Chr.10: 97153042-97292673 on Build GRCh38
Transcripts
NM_001204300.1 NM_001256423.1 NM_032900.5 NR_037909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3442393 c.1480C>T p.L494F Chr.10: 97226127-97226127 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs796250362 Chr.10: 97225790-97225791 on Build GRCh38
rs751426206 Chr.10: 97225749-97225749 on Build GRCh38
rs1052085259 Chr.10: 97225962-97225962 on Build GRCh38
rs770406805 Chr.10: 97226111-97226111 on Build GRCh38
rs951459762 Chr.10: 97225750-97225750 on Build GRCh38
rs79123398 Chr.10: 97225709-97225709 on Build GRCh38
rs959793567 Chr.10: 97225814-97225814 on Build GRCh38
rs886281818 Chr.10: 97226091-97226091 on Build GRCh38
rs936678162 Chr.10: 97225984-97225984 on Build GRCh38
rs935007140 Chr.10: 97225954-97225954 on Build GRCh38
rs74152149 Chr.10: 97225982-97225982 on Build GRCh38
rs112443290 Chr.10: 97225938-97225938 on Build GRCh38
rs760306872 Chr.10: 97226081-97226082 on Build GRCh38
rs969484524 Chr.10: 97225743-97225743 on Build GRCh38
rs539595111 Chr.10: 97226032-97226032 on Build GRCh38
rs140463273 Chr.10: 97225677-97225677 on Build GRCh38
rs982635373 Chr.10: 97225748-97225748 on Build GRCh38
rs941964763 Chr.10: 97225847-97225847 on Build GRCh38
rs933251423 Chr.10: 97226043-97226043 on Build GRCh38
rs963050200 Chr.10: 97225701-97225701 on Build GRCh38
rs776019405 Chr.10: 97226114-97226114 on Build GRCh38
rs12572241 Chr.10: 97225855-97225855 on Build GRCh38
rs758484058 Chr.10: 97226025-97226025 on Build GRCh38
rs367993388 Chr.10: 97225798-97225799 on Build GRCh38
rs553408676 Chr.10: 97225688-97225688 on Build GRCh38
rs905375110 Chr.10: 97226054-97226054 on Build GRCh38
rs372806906 Chr.10: 97226080-97226080 on Build GRCh38
rs1004592337 Chr.10: 97225658-97225661 on Build GRCh38
rs74152150 Chr.10: 97225986-97225986 on Build GRCh38
rs978203599 Chr.10: 97225944-97225944 on Build GRCh38
rs188858831 Chr.10: 97225997-97225997 on Build GRCh38
rs578069856 Chr.10: 97225798-97225798 on Build GRCh38
rs371711711 Chr.10: 97225857-97225857 on Build GRCh38
rs35858317 Chr.10: 97225798-97225798 on Build GRCh38
rs907783473 Chr.10: 97225845-97225845 on Build GRCh38
rs931722704 Chr.10: 97225873-97225873 on Build GRCh38
rs181857867 Chr.10: 97225900-97225900 on Build GRCh38
rs200076395 Chr.10: 97226090-97226090 on Build GRCh38
rs776574463 Chr.10: 97226032-97226033 on Build GRCh38
rs1011139008 Chr.10: 97225983-97225983 on Build GRCh38
rs777371736 Chr.10: 97226079-97226079 on Build GRCh38
rs755655623 Chr.10: 97226071-97226071 on Build GRCh38
rs184839583 Chr.10: 97225990-97225990 on Build GRCh38
rs771691009 Chr.10: 97225824-97225824 on Build GRCh38

View Full Product Details