Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97320675-97321173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRAT1
Entrez Gene ID
10023
Gene Name
frequently rearranged in advanced T-cell lymphomas 1
Gene Aliases
-
UniGene
Hs.126057
Gene Chromosome Location
Chr.10: 97319265-97321915 on Build GRCh38
Transcripts
NM_005479.3

SNP Detail

SNP ID Chromosome Location
rs984772599 Chr.10: 97320947-97320947 on Build GRCh38
rs972622003 Chr.10: 97320897-97320897 on Build GRCh38
rs188824732 Chr.10: 97320786-97320786 on Build GRCh38
rs913808888 Chr.10: 97320759-97320759 on Build GRCh38
rs3781373 Chr.10: 97320828-97320828 on Build GRCh38
rs896128420 Chr.10: 97320881-97320881 on Build GRCh38
rs764919317 Chr.10: 97320860-97320860 on Build GRCh38
rs979214574 Chr.10: 97321117-97321117 on Build GRCh38
rs937861963 Chr.10: 97321149-97321149 on Build GRCh38
rs953613225 Chr.10: 97320850-97320850 on Build GRCh38
rs577464259 Chr.10: 97320800-97320800 on Build GRCh38
rs542289668 Chr.10: 97320926-97320926 on Build GRCh38
rs995682289 Chr.10: 97320972-97320972 on Build GRCh38
rs954097170 Chr.10: 97321000-97321000 on Build GRCh38
rs1019605919 Chr.10: 97320864-97320864 on Build GRCh38
rs368674669 Chr.10: 97320733-97320733 on Build GRCh38
rs555122396 Chr.10: 97320697-97320697 on Build GRCh38
rs189847912 Chr.10: 97321056-97321056 on Build GRCh38
rs1001935435 Chr.10: 97320796-97320796 on Build GRCh38
rs961939669 Chr.10: 97320967-97320967 on Build GRCh38
rs752121421 Chr.10: 97320875-97320875 on Build GRCh38
rs141543730 Chr.10: 97320766-97320766 on Build GRCh38
rs184577692 Chr.10: 97321002-97321002 on Build GRCh38
rs762552622 Chr.10: 97321017-97321017 on Build GRCh38
rs1045684693 Chr.10: 97320966-97320966 on Build GRCh38
rs925149316 Chr.10: 97321125-97321125 on Build GRCh38
rs948648754 Chr.10: 97320962-97320962 on Build GRCh38
rs892905141 Chr.10: 97321142-97321142 on Build GRCh38
rs1013689490 Chr.10: 97320903-97320903 on Build GRCh38
rs1334891 Chr.10: 97321114-97321114 on Build GRCh38
rs574720922 Chr.10: 97320898-97320898 on Build GRCh38
rs750293899 Chr.10: 97320705-97320705 on Build GRCh38
rs550919413 Chr.10: 97321100-97321100 on Build GRCh38
rs1019728496 Chr.10: 97321016-97321016 on Build GRCh38
rs986364957 Chr.10: 97320851-97320851 on Build GRCh38
rs999089908 Chr.10: 97320746-97320746 on Build GRCh38
rs965366957 Chr.10: 97321048-97321048 on Build GRCh38
rs907004001 Chr.10: 97320734-97320734 on Build GRCh38
rs529930770 Chr.10: 97321129-97321129 on Build GRCh38
rs193168823 Chr.10: 97320939-97320939 on Build GRCh38
rs572712277 Chr.10: 97320957-97320957 on Build GRCh38
rs774953245 Chr.10: 97320919-97320919 on Build GRCh38
rs1027196168 Chr.10: 97320977-97320977 on Build GRCh38
rs544546875 Chr.10: 97320820-97320820 on Build GRCh38
rs566718362 Chr.10: 97320719-97320719 on Build GRCh38
rs966275343 Chr.10: 97320883-97320883 on Build GRCh38
rs903607932 Chr.10: 97320794-97320794 on Build GRCh38
rs534401971 Chr.10: 97320958-97320958 on Build GRCh38
rs1040266367 Chr.10: 97320735-97320735 on Build GRCh38

View Full Product Details