Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116679421-116679914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA12A
Entrez Gene ID
259217
Gene Name
heat shock protein family A (Hsp70) member 12A
Gene Aliases
-
UniGene
Hs.648448
Gene Chromosome Location
Chr.10: 116671192-116851241 on Build GRCh38
Transcripts
NM_025015.2 XM_005269672.3 XM_005269673.4 XM_011539579.2 XM_011539580.2 XM_017016032.1

SNP Detail

SNP ID Chromosome Location
rs782127907 Chr.10: 116679752-116679752 on Build GRCh38
rs781872827 Chr.10: 116679644-116679644 on Build GRCh38
rs782197880 Chr.10: 116679724-116679724 on Build GRCh38
rs782248180 Chr.10: 116679475-116679475 on Build GRCh38
rs781985770 Chr.10: 116679751-116679751 on Build GRCh38
rs782026978 Chr.10: 116679586-116679586 on Build GRCh38
rs782204643 Chr.10: 116679718-116679718 on Build GRCh38
rs374166494 Chr.10: 116679639-116679639 on Build GRCh38
rs782314856 Chr.10: 116679486-116679486 on Build GRCh38
rs782461502 Chr.10: 116679666-116679666 on Build GRCh38
rs535724982 Chr.10: 116679612-116679612 on Build GRCh38
rs377732716 Chr.10: 116679463-116679463 on Build GRCh38
rs1014422410 Chr.10: 116679862-116679862 on Build GRCh38
rs77942176 Chr.10: 116679578-116679578 on Build GRCh38
rs782459296 Chr.10: 116679802-116679802 on Build GRCh38
rs17095105 Chr.10: 116679471-116679471 on Build GRCh38
rs781839610 Chr.10: 116679517-116679517 on Build GRCh38
rs782207349 Chr.10: 116679560-116679560 on Build GRCh38
rs781903901 Chr.10: 116679534-116679534 on Build GRCh38
rs782636112 Chr.10: 116679712-116679712 on Build GRCh38
rs368507473 Chr.10: 116679614-116679614 on Build GRCh38
rs781854268 Chr.10: 116679500-116679500 on Build GRCh38
rs782513391 Chr.10: 116679537-116679537 on Build GRCh38
rs61753067 Chr.10: 116679695-116679695 on Build GRCh38
rs781934341 Chr.10: 116679624-116679624 on Build GRCh38
rs782405246 Chr.10: 116679733-116679733 on Build GRCh38
rs200287668 Chr.10: 116679760-116679760 on Build GRCh38
rs782754614 Chr.10: 116679794-116679794 on Build GRCh38
rs782408049 Chr.10: 116679581-116679581 on Build GRCh38
rs781918083 Chr.10: 116679723-116679723 on Build GRCh38
rs1024483926 Chr.10: 116679868-116679868 on Build GRCh38
rs375001436 Chr.10: 116679806-116679806 on Build GRCh38
rs555671481 Chr.10: 116679613-116679613 on Build GRCh38
rs782352033 Chr.10: 116679721-116679721 on Build GRCh38
rs782431209 Chr.10: 116679519-116679519 on Build GRCh38
rs782389933 Chr.10: 116679607-116679607 on Build GRCh38
rs369761934 Chr.10: 116679453-116679453 on Build GRCh38
rs550568601 Chr.10: 116679485-116679485 on Build GRCh38
rs200971754 Chr.10: 116679550-116679550 on Build GRCh38
rs782746967 Chr.10: 116679759-116679759 on Build GRCh38
rs146698512 Chr.10: 116679481-116679481 on Build GRCh38
rs782717575 Chr.10: 116679771-116679771 on Build GRCh38
rs201032256 Chr.10: 116679538-116679538 on Build GRCh38
rs781799434 Chr.10: 116679665-116679665 on Build GRCh38
rs782128836 Chr.10: 116679513-116679513 on Build GRCh38
rs782506966 Chr.10: 116679708-116679708 on Build GRCh38
rs558137014 Chr.10: 116679785-116679785 on Build GRCh38
rs367658412 Chr.10: 116679811-116679811 on Build GRCh38
rs61742406 Chr.10: 116679520-116679520 on Build GRCh38
rs782468606 Chr.10: 116679718-116679718 on Build GRCh38
rs1051406561 Chr.10: 116679737-116679737 on Build GRCh38
rs956352987 Chr.10: 116679849-116679849 on Build GRCh38
rs782219344 Chr.10: 116679539-116679539 on Build GRCh38
rs993435166 Chr.10: 116679523-116679523 on Build GRCh38
rs782748326 Chr.10: 116679664-116679664 on Build GRCh38
rs782466965 Chr.10: 116679651-116679651 on Build GRCh38
rs1031743623 Chr.10: 116679823-116679823 on Build GRCh38
rs782467654 Chr.10: 116679454-116679454 on Build GRCh38
rs376895420 Chr.10: 116679490-116679490 on Build GRCh38
rs781894611 Chr.10: 116679784-116679784 on Build GRCh38
rs370721153 Chr.10: 116679634-116679634 on Build GRCh38
rs570403259 Chr.10: 116679487-116679487 on Build GRCh38
rs782055432 Chr.10: 116679765-116679765 on Build GRCh38
rs1006769914 Chr.10: 116679776-116679776 on Build GRCh38
rs782232321 Chr.10: 116679589-116679589 on Build GRCh38
rs782794995 Chr.10: 116679492-116679492 on Build GRCh38
rs61741265 Chr.10: 116679540-116679540 on Build GRCh38
rs781969339 Chr.10: 116679801-116679801 on Build GRCh38
rs540763581 Chr.10: 116679885-116679885 on Build GRCh38
rs948988598 Chr.10: 116679544-116679544 on Build GRCh38
rs904549897 Chr.10: 116679788-116679788 on Build GRCh38
rs373345193 Chr.10: 116679482-116679482 on Build GRCh38
rs782674186 Chr.10: 116679682-116679682 on Build GRCh38

View Full Product Details