Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 223746-224253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT3
Entrez Gene ID
23410
Gene Name
sirtuin 3
Gene Aliases
SIR2L3
UniGene
Hs.716456
Gene Chromosome Location
Chr.11: 215030-236950 on Build GRCh38
Transcripts
NM_001017524.2 NM_012239.5 XM_005252835.1 XM_011519956.1 XM_011519957.1 XM_017017428.1 XM_017017429.1 XM_017017430.1 XM_017017431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2112658 c.928C>T p.P310S Chr.11: 224119-224119 on Build GRCh38
4425803 c.808-4C>G p.? Chr.11: 224243-224243 on Build GRCh38
5503692 c.916G>A p.V306M Chr.11: 224131-224131 on Build GRCh38
926004 c.856G>A p.V286I Chr.11: 224191-224191 on Build GRCh38
4425802 c.969+7C>T p.? Chr.11: 224071-224071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781721642 Chr.11: 223840-223840 on Build GRCh38
rs761925596 Chr.11: 224056-224056 on Build GRCh38
rs144507767 Chr.11: 223832-223832 on Build GRCh38
rs143392832 Chr.11: 224219-224219 on Build GRCh38
rs564207737 Chr.11: 224001-224001 on Build GRCh38
rs201905714 Chr.11: 224157-224157 on Build GRCh38
rs781079662 Chr.11: 224225-224225 on Build GRCh38
rs200605181 Chr.11: 224101-224101 on Build GRCh38
rs142592164 Chr.11: 224211-224211 on Build GRCh38
rs746994524 Chr.11: 224017-224017 on Build GRCh38
rs11246011 Chr.11: 223904-223904 on Build GRCh38
rs868391020 Chr.11: 224116-224116 on Build GRCh38
rs200188428 Chr.11: 224039-224039 on Build GRCh38
rs768289126 Chr.11: 223860-223860 on Build GRCh38
rs555602013 Chr.11: 223968-223968 on Build GRCh38
rs780840198 Chr.11: 224178-224179 on Build GRCh38
rs551570 Chr.11: 224088-224088 on Build GRCh38
rs765121861 Chr.11: 223858-223859 on Build GRCh38
rs766977903 Chr.11: 224203-224203 on Build GRCh38
rs754199589 Chr.11: 224079-224079 on Build GRCh38
rs201289946 Chr.11: 224179-224179 on Build GRCh38
rs931911891 Chr.11: 223803-223803 on Build GRCh38
rs1050069813 Chr.11: 224009-224009 on Build GRCh38
rs200105251 Chr.11: 224144-224144 on Build GRCh38
rs202035351 Chr.11: 224195-224195 on Build GRCh38
rs549940 Chr.11: 223819-223819 on Build GRCh38
rs532779708 Chr.11: 223778-223778 on Build GRCh38
rs773624991 Chr.11: 223823-223829 on Build GRCh38
rs568268299 Chr.11: 223859-223859 on Build GRCh38
rs752653410 Chr.11: 223859-223860 on Build GRCh38
rs201762879 Chr.11: 224131-224131 on Build GRCh38
rs72479380 Chr.11: 223796-223796 on Build GRCh38
rs149395025 Chr.11: 223823-223823 on Build GRCh38
rs774505562 Chr.11: 224049-224049 on Build GRCh38
rs145518170 Chr.11: 224176-224176 on Build GRCh38
rs763584627 Chr.11: 223862-223863 on Build GRCh38
rs763108948 Chr.11: 223802-223802 on Build GRCh38
rs749174328 Chr.11: 223786-223786 on Build GRCh38
rs369924462 Chr.11: 224099-224099 on Build GRCh38
rs570857 Chr.11: 224012-224012 on Build GRCh38
rs199967856 Chr.11: 224165-224165 on Build GRCh38
rs201048317 Chr.11: 223854-223854 on Build GRCh38
rs1010377667 Chr.11: 224091-224091 on Build GRCh38
rs201217517 Chr.11: 224156-224156 on Build GRCh38
rs11246013 Chr.11: 223940-223940 on Build GRCh38
rs765568528 Chr.11: 224206-224206 on Build GRCh38
rs12790038 Chr.11: 223998-223998 on Build GRCh38
rs199515394 Chr.11: 224070-224070 on Build GRCh38
rs199770042 Chr.11: 223855-223855 on Build GRCh38
rs868118182 Chr.11: 224171-224171 on Build GRCh38
rs755355075 Chr.11: 224087-224087 on Build GRCh38
rs746455495 Chr.11: 224214-224214 on Build GRCh38
rs757954291 Chr.11: 223860-223861 on Build GRCh38
rs748216064 Chr.11: 223796-223797 on Build GRCh38
rs115737484 Chr.11: 223967-223967 on Build GRCh38
rs201895298 Chr.11: 224064-224064 on Build GRCh38
rs747796290 Chr.11: 224105-224105 on Build GRCh38
rs777213044 Chr.11: 224136-224136 on Build GRCh38
rs760457646 Chr.11: 223787-223787 on Build GRCh38
rs773102485 Chr.11: 223897-223897 on Build GRCh38
rs756909662 Chr.11: 224028-224028 on Build GRCh38
rs766639509 Chr.11: 223856-223857 on Build GRCh38
rs201576457 Chr.11: 224205-224205 on Build GRCh38
rs1002218886 Chr.11: 224107-224107 on Build GRCh38
rs147722093 Chr.11: 224194-224194 on Build GRCh38
rs11246010 Chr.11: 223868-223868 on Build GRCh38
rs924016767 Chr.11: 223806-223806 on Build GRCh38
rs778621111 Chr.11: 224095-224095 on Build GRCh38
rs756649033 Chr.11: 224067-224067 on Build GRCh38
rs200209873 Chr.11: 224181-224181 on Build GRCh38
rs757902572 Chr.11: 224114-224114 on Build GRCh38
rs376256203 Chr.11: 223809-223809 on Build GRCh38
rs11246012 Chr.11: 223926-223926 on Build GRCh38
rs751587211 Chr.11: 223824-223824 on Build GRCh38
rs760971529 Chr.11: 223832-223843 on Build GRCh38
rs377119925 Chr.11: 223895-223895 on Build GRCh38
rs12222188 Chr.11: 224063-224063 on Build GRCh38
rs201772736 Chr.11: 224201-224201 on Build GRCh38
rs371073141 Chr.11: 224027-224027 on Build GRCh38
rs796833849 Chr.11: 223770-223770 on Build GRCh38
rs868582935 Chr.11: 223847-223847 on Build GRCh38
rs12223279 Chr.11: 224204-224204 on Build GRCh38
rs776299097 Chr.11: 223857-223858 on Build GRCh38
rs766201880 Chr.11: 223788-223788 on Build GRCh38
rs142088229 Chr.11: 224102-224102 on Build GRCh38
rs761235190 Chr.11: 223773-223773 on Build GRCh38
rs973181497 Chr.11: 224193-224193 on Build GRCh38
rs570772 Chr.11: 224048-224048 on Build GRCh38
rs747679324 Chr.11: 223876-223876 on Build GRCh38
rs202141195 Chr.11: 224180-224180 on Build GRCh38
rs201454850 Chr.11: 224040-224040 on Build GRCh38
rs562348241 Chr.11: 224192-224192 on Build GRCh38
rs199552510 Chr.11: 224212-224212 on Build GRCh38
rs773359296 Chr.11: 224196-224196 on Build GRCh38
rs975639946 Chr.11: 224023-224023 on Build GRCh38
rs757341603 Chr.11: 223833-223833 on Build GRCh38
rs539431373 Chr.11: 223929-223929 on Build GRCh38
rs11246014 Chr.11: 223976-223976 on Build GRCh38
rs770872784 Chr.11: 224026-224026 on Build GRCh38
rs760852779 Chr.11: 224199-224199 on Build GRCh38
rs772254049 Chr.11: 223802-223803 on Build GRCh38
rs576629528 Chr.11: 223955-223955 on Build GRCh38
rs752890938 Chr.11: 223792-223792 on Build GRCh38

View Full Product Details