Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1923469-1923971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNNT3
Entrez Gene ID
7140
Gene Name
troponin T3, fast skeletal type
Gene Aliases
TNTF
UniGene
Hs.73454
Gene Chromosome Location
Chr.11: 1919551-1938706 on Build GRCh38
Transcripts
NM_001042780.2 NM_001042781.2 NM_001042782.2 NM_001297646.1 NM_006757.3 XM_006718290.3 XM_006718293.2 XM_006718294.3 XM_006718296.3 XM_006718299.2 XM_006718300.3 XM_006718302.3 XM_011520343.2 XM_017018205.1 XM_017018206.1 XM_017018207.1 XM_017018208.1 XM_017018209.1 XM_017018210.1 XM_017018211.1 XM_017018212.1 XM_017018213.1 XM_017018214.1 XM_017018215.1 XM_017018216.1 XM_017018217.1 XM_017018218.1 XM_017018219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5586824 c.43G>A p.E15K Chr.11: 1923566-1923566 on Build GRCh38
5586825 c.43G>A p.E15K Chr.11: 1923566-1923566 on Build GRCh38
4333901 c.40_42delGAA p.E18delE Chr.11: 1923563-1923565 on Build GRCh38
2093377 c.40_42delGAA p.E18delE Chr.11: 1923563-1923565 on Build GRCh38
2093376 c.40_42delGAA p.E18delE Chr.11: 1923563-1923565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047880316 Chr.11: 1923688-1923688 on Build GRCh38
rs372467045 Chr.11: 1923590-1923590 on Build GRCh38
rs950253441 Chr.11: 1923921-1923921 on Build GRCh38
rs774300369 Chr.11: 1923840-1923840 on Build GRCh38
rs540889944 Chr.11: 1923494-1923494 on Build GRCh38
rs1051891699 Chr.11: 1923685-1923685 on Build GRCh38
rs78662878 Chr.11: 1923944-1923944 on Build GRCh38
rs867832480 Chr.11: 1923867-1923867 on Build GRCh38
rs115630871 Chr.11: 1923677-1923677 on Build GRCh38
rs201230428 Chr.11: 1923529-1923529 on Build GRCh38
rs369789445 Chr.11: 1923569-1923569 on Build GRCh38
rs199720054 Chr.11: 1923597-1923597 on Build GRCh38
rs765524276 Chr.11: 1923595-1923596 on Build GRCh38
rs373689006 Chr.11: 1923602-1923602 on Build GRCh38
rs199583374 Chr.11: 1923601-1923601 on Build GRCh38
rs1011164381 Chr.11: 1923810-1923811 on Build GRCh38
rs752762981 Chr.11: 1923544-1923544 on Build GRCh38
rs745695630 Chr.11: 1923580-1923580 on Build GRCh38
rs770689910 Chr.11: 1923831-1923831 on Build GRCh38
rs753016317 Chr.11: 1923596-1923596 on Build GRCh38
rs975018438 Chr.11: 1923635-1923635 on Build GRCh38
rs900781691 Chr.11: 1923676-1923676 on Build GRCh38
rs530639254 Chr.11: 1923927-1923974 on Build GRCh38
rs751384508 Chr.11: 1923622-1923623 on Build GRCh38
rs374167020 Chr.11: 1923495-1923495 on Build GRCh38
rs984045145 Chr.11: 1923868-1923868 on Build GRCh38
rs375214921 Chr.11: 1923628-1923628 on Build GRCh38
rs765750042 Chr.11: 1923603-1923603 on Build GRCh38
rs758500654 Chr.11: 1923602-1923603 on Build GRCh38
rs201240144 Chr.11: 1923595-1923595 on Build GRCh38
rs946414206 Chr.11: 1923653-1923653 on Build GRCh38
rs975510300 Chr.11: 1923855-1923855 on Build GRCh38
rs376786651 Chr.11: 1923562-1923562 on Build GRCh38
rs751685193 Chr.11: 1923620-1923620 on Build GRCh38
rs1024007574 Chr.11: 1923589-1923589 on Build GRCh38
rs544457579 Chr.11: 1923926-1923926 on Build GRCh38
rs367656256 Chr.11: 1923527-1923527 on Build GRCh38
rs200379236 Chr.11: 1923547-1923547 on Build GRCh38
rs530511047 Chr.11: 1923933-1923933 on Build GRCh38
rs386749737 Chr.11: 1923922-1923923 on Build GRCh38
rs908009365 Chr.11: 1923904-1923904 on Build GRCh38
rs930844655 Chr.11: 1923684-1923684 on Build GRCh38
rs760610444 Chr.11: 1923522-1923522 on Build GRCh38
rs1039366040 Chr.11: 1923655-1923655 on Build GRCh38
rs558294893 Chr.11: 1923821-1923821 on Build GRCh38
rs539981352 Chr.11: 1923744-1923744 on Build GRCh38
rs755208612 Chr.11: 1923534-1923534 on Build GRCh38
rs573228622 Chr.11: 1923824-1923824 on Build GRCh38
rs776532883 Chr.11: 1923504-1923504 on Build GRCh38
rs758690476 Chr.11: 1923608-1923608 on Build GRCh38
rs372388005 Chr.11: 1923540-1923540 on Build GRCh38
rs373325754 Chr.11: 1923930-1923930 on Build GRCh38
rs995275291 Chr.11: 1923748-1923748 on Build GRCh38
rs114106473 Chr.11: 1923844-1923844 on Build GRCh38
rs899269850 Chr.11: 1923932-1923932 on Build GRCh38
rs754215349 Chr.11: 1923536-1923537 on Build GRCh38
rs202011950 Chr.11: 1923505-1923505 on Build GRCh38
rs1023616937 Chr.11: 1923778-1923778 on Build GRCh38
rs1004893327 Chr.11: 1923550-1923550 on Build GRCh38
rs757270420 Chr.11: 1923566-1923566 on Build GRCh38
rs987147420 Chr.11: 1923493-1923493 on Build GRCh38
rs554525505 Chr.11: 1923783-1923783 on Build GRCh38
rs766937596 Chr.11: 1923599-1923599 on Build GRCh38
rs3781958 Chr.11: 1923707-1923707 on Build GRCh38
rs920917763 Chr.11: 1923857-1923857 on Build GRCh38
rs1045819189 Chr.11: 1923922-1923922 on Build GRCh38
rs201902036 Chr.11: 1923866-1923866 on Build GRCh38
rs114684124 Chr.11: 1923624-1923624 on Build GRCh38
rs764062440 Chr.11: 1923610-1923610 on Build GRCh38
rs760043392 Chr.11: 1923563-1923565 on Build GRCh38
rs146986911 Chr.11: 1923714-1923714 on Build GRCh38
rs973423737 Chr.11: 1923581-1923581 on Build GRCh38
rs1001472532 Chr.11: 1923770-1923773 on Build GRCh38
rs932242410 Chr.11: 1923862-1923862 on Build GRCh38
rs776803034 Chr.11: 1923591-1923591 on Build GRCh38
rs969311609 Chr.11: 1923779-1923779 on Build GRCh38
rs148051896 Chr.11: 1923923-1923923 on Build GRCh38
rs956487944 Chr.11: 1923823-1923823 on Build GRCh38
rs1034860288 Chr.11: 1923785-1923785 on Build GRCh38
rs368864786 Chr.11: 1923598-1923598 on Build GRCh38
rs777936161 Chr.11: 1923623-1923623 on Build GRCh38
rs769459985 Chr.11: 1923506-1923506 on Build GRCh38
rs371623979 Chr.11: 1923523-1923523 on Build GRCh38
rs540645991 Chr.11: 1923839-1923839 on Build GRCh38
rs535257707 Chr.11: 1923702-1923702 on Build GRCh38
rs768888979 Chr.11: 1923563-1923563 on Build GRCh38
rs200418997 Chr.11: 1923525-1923525 on Build GRCh38
rs1015948893 Chr.11: 1923612-1923612 on Build GRCh38
rs201048428 Chr.11: 1923605-1923605 on Build GRCh38
rs780802963 Chr.11: 1923577-1923577 on Build GRCh38
rs868582873 Chr.11: 1923841-1923841 on Build GRCh38

View Full Product Details