Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5453317-5453825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR51B5
Entrez Gene ID
282763
Gene Name
olfactory receptor family 51 subfamily B member 5
Gene Aliases
HOR5'Beta5, OR11-37
UniGene
Hs.690458
Gene Chromosome Location
Chr.11: 5340883-5505652 on Build GRCh38
Transcripts
NM_001005567.2 NR_038321.1 NM_001004754.2

Gene Symbol
OR51I2
Entrez Gene ID
390064
Gene Name
olfactory receptor family 51 subfamily I member 2
Gene Aliases
OR11-38
UniGene
Hs.553733
Gene Chromosome Location
Chr.11: 5453408-5454477 on Build GRCh38
Transcripts
NM_001005567.2 NR_038321.1 NM_001004754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5361061 c.213G>A p.V71V Chr.11: 5453701-5453701 on Build GRCh38
6551662 c.221C>T p.S74F Chr.11: 5453709-5453709 on Build GRCh38
4033568 c.226G>A p.A76T Chr.11: 5453714-5453714 on Build GRCh38
1218713 c.307C>A p.L103I Chr.11: 5453795-5453795 on Build GRCh38
5079934 c.166C>A p.H56N Chr.11: 5453654-5453654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753761760 Chr.11: 5453723-5453723 on Build GRCh38
rs147328747 Chr.11: 5453551-5453551 on Build GRCh38
rs371172345 Chr.11: 5453495-5453495 on Build GRCh38
rs376825797 Chr.11: 5453465-5453465 on Build GRCh38
rs796735267 Chr.11: 5453363-5453363 on Build GRCh38
rs769839256 Chr.11: 5453537-5453537 on Build GRCh38
rs200708823 Chr.11: 5453684-5453684 on Build GRCh38
rs543285499 Chr.11: 5453772-5453772 on Build GRCh38
rs371546106 Chr.11: 5453625-5453625 on Build GRCh38
rs761110245 Chr.11: 5453639-5453639 on Build GRCh38
rs745490584 Chr.11: 5453444-5453444 on Build GRCh38
rs374880132 Chr.11: 5453442-5453442 on Build GRCh38
rs779879912 Chr.11: 5453514-5453514 on Build GRCh38
rs1043054927 Chr.11: 5453340-5453340 on Build GRCh38
rs117662980 Chr.11: 5453355-5453355 on Build GRCh38
rs752460723 Chr.11: 5453402-5453402 on Build GRCh38
rs780173605 Chr.11: 5453669-5453670 on Build GRCh38
rs780911656 Chr.11: 5453657-5453657 on Build GRCh38
rs749199557 Chr.11: 5453696-5453696 on Build GRCh38
rs764230863 Chr.11: 5453515-5453515 on Build GRCh38
rs372429022 Chr.11: 5453776-5453776 on Build GRCh38
rs1012708668 Chr.11: 5453422-5453422 on Build GRCh38
rs772350779 Chr.11: 5453656-5453656 on Build GRCh38
rs767799957 Chr.11: 5453523-5453525 on Build GRCh38
rs564251258 Chr.11: 5453497-5453497 on Build GRCh38
rs755933130 Chr.11: 5453685-5453685 on Build GRCh38
rs776467569 Chr.11: 5453461-5453461 on Build GRCh38
rs764534110 Chr.11: 5453736-5453736 on Build GRCh38
rs749144584 Chr.11: 5453779-5453779 on Build GRCh38
rs549065495 Chr.11: 5453463-5453463 on Build GRCh38
rs866650577 Chr.11: 5453801-5453801 on Build GRCh38
rs751811140 Chr.11: 5453543-5453543 on Build GRCh38
rs773209144 Chr.11: 5453538-5453538 on Build GRCh38
rs770378310 Chr.11: 5453440-5453440 on Build GRCh38
rs777529437 Chr.11: 5453778-5453778 on Build GRCh38
rs80243568 Chr.11: 5453719-5453719 on Build GRCh38
rs763220863 Chr.11: 5453753-5453753 on Build GRCh38
rs193242103 Chr.11: 5453370-5453370 on Build GRCh38
rs543574678 Chr.11: 5453653-5453653 on Build GRCh38
rs34041261 Chr.11: 5453389-5453390 on Build GRCh38
rs909671496 Chr.11: 5453630-5453630 on Build GRCh38
rs773828247 Chr.11: 5453792-5453793 on Build GRCh38
rs750975545 Chr.11: 5453754-5453754 on Build GRCh38
rs764047191 Chr.11: 5453456-5453456 on Build GRCh38
rs144839036 Chr.11: 5453735-5453735 on Build GRCh38
rs766593008 Chr.11: 5453473-5453473 on Build GRCh38
rs113366643 Chr.11: 5453741-5453741 on Build GRCh38
rs578230333 Chr.11: 5453693-5453693 on Build GRCh38
rs375923603 Chr.11: 5453640-5453640 on Build GRCh38
rs765976440 Chr.11: 5453651-5453651 on Build GRCh38
rs774287640 Chr.11: 5453712-5453712 on Build GRCh38
rs753160596 Chr.11: 5453563-5453563 on Build GRCh38
rs768307813 Chr.11: 5453529-5453529 on Build GRCh38
rs757116270 Chr.11: 5453788-5453788 on Build GRCh38
rs139674475 Chr.11: 5453585-5453585 on Build GRCh38
rs756570371 Chr.11: 5453576-5453576 on Build GRCh38
rs527688089 Chr.11: 5453455-5453455 on Build GRCh38
rs373816161 Chr.11: 5453773-5453773 on Build GRCh38
rs766538002 Chr.11: 5453539-5453539 on Build GRCh38
rs138083584 Chr.11: 5453491-5453491 on Build GRCh38
rs74049540 Chr.11: 5453664-5453664 on Build GRCh38
rs1036650842 Chr.11: 5453425-5453425 on Build GRCh38
rs751628694 Chr.11: 5453542-5453542 on Build GRCh38
rs776541215 Chr.11: 5453531-5453531 on Build GRCh38
rs770911627 Chr.11: 5453706-5453706 on Build GRCh38
rs201440151 Chr.11: 5453584-5453584 on Build GRCh38
rs142661859 Chr.11: 5453642-5453642 on Build GRCh38
rs145612844 Chr.11: 5453517-5453517 on Build GRCh38
rs1056813104 Chr.11: 5453731-5453731 on Build GRCh38
rs549847849 Chr.11: 5453532-5453532 on Build GRCh38
rs2030094 Chr.11: 5453608-5453608 on Build GRCh38
rs750656594 Chr.11: 5453632-5453633 on Build GRCh38
rs572697636 Chr.11: 5453757-5453757 on Build GRCh38
rs757396021 Chr.11: 5453594-5453594 on Build GRCh38
rs965338906 Chr.11: 5453552-5453552 on Build GRCh38
rs778549994 Chr.11: 5453508-5453508 on Build GRCh38
rs895526400 Chr.11: 5453415-5453415 on Build GRCh38
rs746623494 Chr.11: 5453591-5453591 on Build GRCh38
rs148530767 Chr.11: 5453637-5453637 on Build GRCh38
rs775815831 Chr.11: 5453733-5453733 on Build GRCh38
rs777196043 Chr.11: 5453739-5453739 on Build GRCh38
rs756151657 Chr.11: 5453650-5453653 on Build GRCh38
rs376306443 Chr.11: 5453750-5453750 on Build GRCh38
rs771779740 Chr.11: 5453513-5453513 on Build GRCh38
rs771904526 Chr.11: 5453452-5453452 on Build GRCh38
rs867120171 Chr.11: 5453610-5453610 on Build GRCh38
rs779753433 Chr.11: 5453589-5453589 on Build GRCh38
rs758615846 Chr.11: 5453471-5453471 on Build GRCh38
rs765845929 Chr.11: 5453742-5453742 on Build GRCh38
rs374169531 Chr.11: 5453562-5453562 on Build GRCh38
rs751864277 Chr.11: 5453487-5453487 on Build GRCh38
rs867474081 Chr.11: 5453771-5453771 on Build GRCh38
rs756627327 Chr.11: 5453667-5453667 on Build GRCh38
rs560595035 Chr.11: 5453435-5453435 on Build GRCh38
rs932804784 Chr.11: 5453352-5453352 on Build GRCh38
rs368086802 Chr.11: 5453593-5453593 on Build GRCh38
rs145725236 Chr.11: 5453636-5453636 on Build GRCh38
rs750561734 Chr.11: 5453468-5453468 on Build GRCh38
rs754921159 Chr.11: 5453776-5453779 on Build GRCh38
rs867332360 Chr.11: 5453548-5453548 on Build GRCh38
rs745434189 Chr.11: 5453512-5453512 on Build GRCh38
rs902784615 Chr.11: 5453354-5453354 on Build GRCh38
rs367656709 Chr.11: 5453775-5453775 on Build GRCh38
rs533547313 Chr.11: 5453364-5453364 on Build GRCh38
rs754539560 Chr.11: 5453756-5453756 on Build GRCh38
rs147506036 Chr.11: 5453490-5453490 on Build GRCh38
rs144341122 Chr.11: 5453648-5453648 on Build GRCh38
rs747979911 Chr.11: 5453598-5453598 on Build GRCh38
rs756690972 Chr.11: 5453502-5453502 on Build GRCh38
rs745960137 Chr.11: 5453715-5453715 on Build GRCh38

View Full Product Details