Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5967425-5967954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR56A5
Entrez Gene ID
390084
Gene Name
olfactory receptor family 56 subfamily A member 5
Gene Aliases
OR56A5P
UniGene
Hs.719888
Gene Chromosome Location
Chr.11: 5967553-5968494 on Build GRCh38
Transcripts
NM_001146033.1

SNP Detail

SNP ID Chromosome Location
rs760085520 Chr.11: 5967623-5967623 on Build GRCh38
rs908536005 Chr.11: 5967702-5967702 on Build GRCh38
rs753458858 Chr.11: 5967629-5967629 on Build GRCh38
rs770333100 Chr.11: 5967652-5967652 on Build GRCh38
rs777903755 Chr.11: 5967865-5967865 on Build GRCh38
rs776809229 Chr.11: 5967568-5967568 on Build GRCh38
rs200855128 Chr.11: 5967513-5967513 on Build GRCh38
rs760036347 Chr.11: 5967698-5967698 on Build GRCh38
rs970511193 Chr.11: 5967607-5967607 on Build GRCh38
rs762731313 Chr.11: 5967720-5967720 on Build GRCh38
rs141209218 Chr.11: 5967683-5967683 on Build GRCh38
rs763234735 Chr.11: 5967612-5967612 on Build GRCh38
rs557968232 Chr.11: 5967555-5967555 on Build GRCh38
rs764673542 Chr.11: 5967717-5967717 on Build GRCh38
rs764888953 Chr.11: 5967645-5967645 on Build GRCh38
rs746335097 Chr.11: 5967462-5967462 on Build GRCh38
rs548174741 Chr.11: 5967572-5967572 on Build GRCh38
rs372544375 Chr.11: 5967815-5967815 on Build GRCh38
rs770111382 Chr.11: 5967595-5967595 on Build GRCh38
rs373154113 Chr.11: 5967871-5967871 on Build GRCh38
rs905526340 Chr.11: 5967526-5967526 on Build GRCh38
rs917751233 Chr.11: 5967584-5967584 on Build GRCh38
rs780097306 Chr.11: 5967539-5967539 on Build GRCh38
rs751458523 Chr.11: 5967728-5967728 on Build GRCh38
rs761322572 Chr.11: 5967714-5967714 on Build GRCh38
rs761133010 Chr.11: 5967793-5967793 on Build GRCh38
rs747542588 Chr.11: 5967920-5967920 on Build GRCh38
rs746177474 Chr.11: 5967768-5967768 on Build GRCh38
rs1032001657 Chr.11: 5967485-5967485 on Build GRCh38
rs772330706 Chr.11: 5967785-5967785 on Build GRCh38
rs369691077 Chr.11: 5967862-5967862 on Build GRCh38
rs758456519 Chr.11: 5967598-5967598 on Build GRCh38
rs1000961430 Chr.11: 5967467-5967467 on Build GRCh38
rs746963298 Chr.11: 5967561-5967561 on Build GRCh38
rs779043133 Chr.11: 5967907-5967907 on Build GRCh38
rs201804826 Chr.11: 5967651-5967651 on Build GRCh38
rs576642792 Chr.11: 5967910-5967910 on Build GRCh38
rs769313199 Chr.11: 5967795-5967795 on Build GRCh38
rs774792496 Chr.11: 5967622-5967622 on Build GRCh38
rs757040332 Chr.11: 5967928-5967928 on Build GRCh38
rs61738948 Chr.11: 5967676-5967676 on Build GRCh38
rs866578507 Chr.11: 5967805-5967805 on Build GRCh38
rs777735064 Chr.11: 5967675-5967675 on Build GRCh38
rs772921173 Chr.11: 5967806-5967806 on Build GRCh38
rs766762752 Chr.11: 5967617-5967617 on Build GRCh38
rs773318737 Chr.11: 5967596-5967596 on Build GRCh38
rs768131592 Chr.11: 5967700-5967700 on Build GRCh38
rs781119502 Chr.11: 5967681-5967681 on Build GRCh38
rs774737515 Chr.11: 5967696-5967696 on Build GRCh38
rs755583006 Chr.11: 5967509-5967509 on Build GRCh38
rs552689850 Chr.11: 5967761-5967761 on Build GRCh38
rs997271647 Chr.11: 5967457-5967457 on Build GRCh38
rs1027656867 Chr.11: 5967686-5967686 on Build GRCh38
rs541837280 Chr.11: 5967893-5967893 on Build GRCh38
rs935703338 Chr.11: 5967534-5967534 on Build GRCh38
rs771210055 Chr.11: 5967763-5967763 on Build GRCh38
rs751260621 Chr.11: 5967845-5967845 on Build GRCh38
rs781116138 Chr.11: 5967731-5967731 on Build GRCh38
rs748200584 Chr.11: 5967682-5967682 on Build GRCh38
rs746975434 Chr.11: 5967825-5967826 on Build GRCh38
rs1000610464 Chr.11: 5967838-5967838 on Build GRCh38
rs762678081 Chr.11: 5967812-5967812 on Build GRCh38
rs1024818924 Chr.11: 5967890-5967890 on Build GRCh38
rs776028165 Chr.11: 5967789-5967789 on Build GRCh38
rs1049902172 Chr.11: 5967762-5967762 on Build GRCh38
rs953141865 Chr.11: 5967465-5967465 on Build GRCh38
rs756979437 Chr.11: 5967515-5967515 on Build GRCh38
rs752605920 Chr.11: 5967736-5967736 on Build GRCh38
rs1029137462 Chr.11: 5967463-5967463 on Build GRCh38
rs756849900 Chr.11: 5967644-5967644 on Build GRCh38
rs540432832 Chr.11: 5967767-5967767 on Build GRCh38
rs927002158 Chr.11: 5967776-5967776 on Build GRCh38
rs752659451 Chr.11: 5967860-5967860 on Build GRCh38
rs375405523 Chr.11: 5967825-5967825 on Build GRCh38
rs764917891 Chr.11: 5967593-5967593 on Build GRCh38
rs777765292 Chr.11: 5967754-5967754 on Build GRCh38
rs759334315 Chr.11: 5967855-5967855 on Build GRCh38
rs1033516747 Chr.11: 5967911-5967911 on Build GRCh38
rs753411177 Chr.11: 5967514-5967514 on Build GRCh38
rs193118599 Chr.11: 5967915-5967915 on Build GRCh38
rs757346444 Chr.11: 5967902-5967902 on Build GRCh38
rs373559237 Chr.11: 5967790-5967790 on Build GRCh38
rs773445988 Chr.11: 5967685-5967685 on Build GRCh38
rs1021050358 Chr.11: 5967658-5967658 on Build GRCh38
rs771264926 Chr.11: 5967688-5967688 on Build GRCh38
rs749536257 Chr.11: 5967687-5967687 on Build GRCh38
rs1032428934 Chr.11: 5967849-5967849 on Build GRCh38
rs368896033 Chr.11: 5967729-5967729 on Build GRCh38
rs767240410 Chr.11: 5967856-5967856 on Build GRCh38
rs7113015 Chr.11: 5967677-5967677 on Build GRCh38
rs116341877 Chr.11: 5967719-5967719 on Build GRCh38
rs983398624 Chr.11: 5967706-5967706 on Build GRCh38
rs768831543 Chr.11: 5967562-5967562 on Build GRCh38
rs188179224 Chr.11: 5967791-5967791 on Build GRCh38
rs778571561 Chr.11: 5967522-5967522 on Build GRCh38
rs79668572 Chr.11: 5967525-5967525 on Build GRCh38
rs763564644 Chr.11: 5967628-5967628 on Build GRCh38
rs890646377 Chr.11: 5967667-5967667 on Build GRCh38
rs563159606 Chr.11: 5967908-5967908 on Build GRCh38
rs538383484 Chr.11: 5967484-5967484 on Build GRCh38
rs534407077 Chr.11: 5967721-5967721 on Build GRCh38
rs745737471 Chr.11: 5967523-5967523 on Build GRCh38

View Full Product Details