Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35441275-35441809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
UniGene
Hs.55044
Gene Chromosome Location
Chr.11: 35431827-35530300 on Build GRCh38
Transcripts
NM_001001991.2 NM_001282675.1 NM_001282676.1 NM_015430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2115869 c.954C>T p.N318N Chr.11: 35441611-35441611 on Build GRCh38
6302929 c.872T>C p.V291A Chr.11: 35441693-35441693 on Build GRCh38
2115865 c.1007A>G p.Y336C Chr.11: 35441558-35441558 on Build GRCh38
5999982 c.955G>A p.G319R Chr.11: 35441610-35441610 on Build GRCh38
3447016 c.951C>T p.I317I Chr.11: 35441614-35441614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762193264 Chr.11: 35441740-35441740 on Build GRCh38
rs771781749 Chr.11: 35441476-35441476 on Build GRCh38
rs747971433 Chr.11: 35441688-35441688 on Build GRCh38
rs146700269 Chr.11: 35441572-35441572 on Build GRCh38
rs192795455 Chr.11: 35441723-35441723 on Build GRCh38
rs771892466 Chr.11: 35441460-35441460 on Build GRCh38
rs760914031 Chr.11: 35441651-35441651 on Build GRCh38
rs772231139 Chr.11: 35441705-35441705 on Build GRCh38
rs769787876 Chr.11: 35441694-35441694 on Build GRCh38
rs565994772 Chr.11: 35441355-35441355 on Build GRCh38
rs1040565656 Chr.11: 35441385-35441385 on Build GRCh38
rs779555385 Chr.11: 35441667-35441667 on Build GRCh38
rs745585185 Chr.11: 35441540-35441540 on Build GRCh38
rs910809379 Chr.11: 35441411-35441411 on Build GRCh38
rs572768296 Chr.11: 35441602-35441602 on Build GRCh38
rs759667343 Chr.11: 35441646-35441646 on Build GRCh38
rs186146967 Chr.11: 35441546-35441546 on Build GRCh38
rs149018193 Chr.11: 35441607-35441607 on Build GRCh38
rs534940410 Chr.11: 35441360-35441360 on Build GRCh38
rs749891554 Chr.11: 35441663-35441663 on Build GRCh38
rs201085073 Chr.11: 35441456-35441456 on Build GRCh38
rs989849737 Chr.11: 35441617-35441617 on Build GRCh38
rs759302630 Chr.11: 35441459-35441459 on Build GRCh38
rs778164985 Chr.11: 35441742-35441742 on Build GRCh38
rs750406778 Chr.11: 35441477-35441477 on Build GRCh38
rs768548437 Chr.11: 35441609-35441609 on Build GRCh38
rs748516329 Chr.11: 35441593-35441593 on Build GRCh38
rs1054105129 Chr.11: 35441313-35441314 on Build GRCh38
rs562004347 Chr.11: 35441611-35441611 on Build GRCh38
rs751829263 Chr.11: 35441578-35441578 on Build GRCh38
rs375293926 Chr.11: 35441531-35441531 on Build GRCh38
rs760583701 Chr.11: 35441467-35441467 on Build GRCh38
rs910179142 Chr.11: 35441631-35441632 on Build GRCh38
rs943615467 Chr.11: 35441439-35441439 on Build GRCh38
rs1044237725 Chr.11: 35441783-35441783 on Build GRCh38
rs775490541 Chr.11: 35441463-35441463 on Build GRCh38
rs143655671 Chr.11: 35441626-35441626 on Build GRCh38
rs773314273 Chr.11: 35441702-35441702 on Build GRCh38
rs757951569 Chr.11: 35441665-35441665 on Build GRCh38
rs761012112 Chr.11: 35441706-35441706 on Build GRCh38
rs11033121 Chr.11: 35441354-35441354 on Build GRCh38
rs79736526 Chr.11: 35441303-35441303 on Build GRCh38
rs767585784 Chr.11: 35441648-35441648 on Build GRCh38
rs772382832 Chr.11: 35441526-35441526 on Build GRCh38
rs763105648 Chr.11: 35441564-35441564 on Build GRCh38
rs796870782 Chr.11: 35441299-35441299 on Build GRCh38
rs751881828 Chr.11: 35441487-35441487 on Build GRCh38
rs141335779 Chr.11: 35441586-35441586 on Build GRCh38
rs539863644 Chr.11: 35441316-35441316 on Build GRCh38
rs777146563 Chr.11: 35441583-35441583 on Build GRCh38
rs369562255 Chr.11: 35441613-35441613 on Build GRCh38
rs777207310 Chr.11: 35441447-35441447 on Build GRCh38
rs764102291 Chr.11: 35441470-35441470 on Build GRCh38
rs544392330 Chr.11: 35441682-35441682 on Build GRCh38
rs770476180 Chr.11: 35441527-35441527 on Build GRCh38
rs754326970 Chr.11: 35441658-35441658 on Build GRCh38
rs776910564 Chr.11: 35441727-35441727 on Build GRCh38
rs113713187 Chr.11: 35441302-35441303 on Build GRCh38
rs570854035 Chr.11: 35441315-35441315 on Build GRCh38
rs372808412 Chr.11: 35441660-35441660 on Build GRCh38
rs768442771 Chr.11: 35441561-35441561 on Build GRCh38
rs766662203 Chr.11: 35441486-35441486 on Build GRCh38
rs371928843 Chr.11: 35441606-35441606 on Build GRCh38
rs759896615 Chr.11: 35441581-35441581 on Build GRCh38
rs139863360 Chr.11: 35441480-35441480 on Build GRCh38
rs375277907 Chr.11: 35441452-35441452 on Build GRCh38
rs753235834 Chr.11: 35441503-35441503 on Build GRCh38
rs779010834 Chr.11: 35441318-35441318 on Build GRCh38
rs754382086 Chr.11: 35441591-35441591 on Build GRCh38
rs943660828 Chr.11: 35441680-35441680 on Build GRCh38
rs750982014 Chr.11: 35441743-35441743 on Build GRCh38
rs771883617 Chr.11: 35441553-35441553 on Build GRCh38
rs374907179 Chr.11: 35441610-35441610 on Build GRCh38
rs934330212 Chr.11: 35441730-35441730 on Build GRCh38
rs1006215751 Chr.11: 35441514-35441514 on Build GRCh38
rs150252565 Chr.11: 35441587-35441587 on Build GRCh38
rs774515894 Chr.11: 35441627-35441627 on Build GRCh38
rs775829954 Chr.11: 35441649-35441649 on Build GRCh38
rs987677701 Chr.11: 35441671-35441671 on Build GRCh38
rs367709317 Chr.11: 35441605-35441605 on Build GRCh38
rs879694939 Chr.11: 35441303-35441303 on Build GRCh38
rs748746852 Chr.11: 35441525-35441525 on Build GRCh38
rs200531458 Chr.11: 35441556-35441556 on Build GRCh38
rs764585639 Chr.11: 35441656-35441656 on Build GRCh38
rs1013953610 Chr.11: 35441779-35441779 on Build GRCh38
rs760519296 Chr.11: 35441558-35441558 on Build GRCh38
rs775085957 Chr.11: 35441557-35441557 on Build GRCh38
rs199686522 Chr.11: 35441704-35441704 on Build GRCh38
rs764484315 Chr.11: 35441588-35441588 on Build GRCh38
rs188272517 Chr.11: 35441677-35441677 on Build GRCh38
rs368261001 Chr.11: 35441453-35441453 on Build GRCh38
rs919347870 Chr.11: 35441618-35441618 on Build GRCh38
rs16927482 Chr.11: 35441601-35441601 on Build GRCh38
rs1051898526 Chr.11: 35441386-35441386 on Build GRCh38
rs756752377 Chr.11: 35441599-35441599 on Build GRCh38
rs776657487 Chr.11: 35441562-35441562 on Build GRCh38
rs746718471 Chr.11: 35441673-35441673 on Build GRCh38
rs778375596 Chr.11: 35441506-35441506 on Build GRCh38
rs113777401 Chr.11: 35441499-35441499 on Build GRCh38
rs1042761343 Chr.11: 35441449-35441449 on Build GRCh38
rs756521194 Chr.11: 35441504-35441504 on Build GRCh38

View Full Product Details