Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45805304-45805835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C1
Entrez Gene ID
55343
Gene Name
solute carrier family 35 member C1
Gene Aliases
CDG2C, FUCT1
UniGene
Hs.12211
Gene Chromosome Location
Chr.11: 45804072-45813016 on Build GRCh38
Transcripts
NM_001145265.1 NM_001145266.1 NM_018389.4 XM_011520203.2

SNP Detail

SNP ID Chromosome Location
rs751618860 Chr.11: 45805766-45805766 on Build GRCh38
rs1047069608 Chr.11: 45805543-45805543 on Build GRCh38
rs538320044 Chr.11: 45805811-45805811 on Build GRCh38
rs1022746927 Chr.11: 45805451-45805451 on Build GRCh38
rs950498553 Chr.11: 45805559-45805559 on Build GRCh38
rs886048311 Chr.11: 45805409-45805409 on Build GRCh38
rs772429552 Chr.11: 45805532-45805532 on Build GRCh38
rs530489331 Chr.11: 45805586-45805586 on Build GRCh38
rs186564976 Chr.11: 45805788-45805788 on Build GRCh38
rs187368323 Chr.11: 45805698-45805698 on Build GRCh38
rs767597659 Chr.11: 45805708-45805708 on Build GRCh38
rs756481902 Chr.11: 45805791-45805791 on Build GRCh38
rs544559319 Chr.11: 45805573-45805573 on Build GRCh38
rs924679340 Chr.11: 45805667-45805667 on Build GRCh38
rs888630125 Chr.11: 45805339-45805339 on Build GRCh38
rs955620712 Chr.11: 45805415-45805415 on Build GRCh38
rs886048312 Chr.11: 45805490-45805490 on Build GRCh38
rs893170637 Chr.11: 45805682-45805682 on Build GRCh38
rs911404277 Chr.11: 45805490-45805490 on Build GRCh38
rs1011415905 Chr.11: 45805660-45805660 on Build GRCh38
rs771383956 Chr.11: 45805485-45805485 on Build GRCh38
rs1029977935 Chr.11: 45805389-45805389 on Build GRCh38
rs181710036 Chr.11: 45805673-45805673 on Build GRCh38
rs1002648702 Chr.11: 45805710-45805710 on Build GRCh38
rs1018520778 Chr.11: 45805739-45805739 on Build GRCh38
rs1026032583 Chr.11: 45805549-45805549 on Build GRCh38
rs749571447 Chr.11: 45805801-45805802 on Build GRCh38
rs753045450 Chr.11: 45805789-45805789 on Build GRCh38
rs998523805 Chr.11: 45805322-45805322 on Build GRCh38
rs930024748 Chr.11: 45805540-45805540 on Build GRCh38
rs886048310 Chr.11: 45805321-45805321 on Build GRCh38
rs886048313 Chr.11: 45805568-45805568 on Build GRCh38
rs3808976 Chr.11: 45805799-45805799 on Build GRCh38
rs866028872 Chr.11: 45805589-45805589 on Build GRCh38
rs1048579789 Chr.11: 45805328-45805328 on Build GRCh38
rs528085855 Chr.11: 45805533-45805533 on Build GRCh38
rs755166873 Chr.11: 45805777-45805777 on Build GRCh38
rs540916963 Chr.11: 45805496-45805496 on Build GRCh38
rs974599944 Chr.11: 45805515-45805515 on Build GRCh38
rs934166780 Chr.11: 45805622-45805622 on Build GRCh38
rs987058897 Chr.11: 45805419-45805419 on Build GRCh38
rs921392592 Chr.11: 45805621-45805621 on Build GRCh38
rs902952952 Chr.11: 45805691-45805691 on Build GRCh38
rs558954435 Chr.11: 45805797-45805797 on Build GRCh38
rs564660234 Chr.11: 45805584-45805584 on Build GRCh38
rs999276653 Chr.11: 45805403-45805403 on Build GRCh38
rs566600488 Chr.11: 45805736-45805736 on Build GRCh38
rs1055710554 Chr.11: 45805591-45805591 on Build GRCh38
rs12289963 Chr.11: 45805499-45805499 on Build GRCh38
rs546865255 Chr.11: 45805714-45805714 on Build GRCh38
rs766594374 Chr.11: 45805758-45805758 on Build GRCh38
rs555128038 Chr.11: 45805363-45805363 on Build GRCh38
rs954343211 Chr.11: 45805721-45805721 on Build GRCh38
rs575057631 Chr.11: 45805428-45805428 on Build GRCh38
rs757212495 Chr.11: 45805753-45805753 on Build GRCh38
rs746314502 Chr.11: 45805513-45805513 on Build GRCh38
rs1013217184 Chr.11: 45805685-45805685 on Build GRCh38
rs1012875170 Chr.11: 45805422-45805422 on Build GRCh38
rs773132151 Chr.11: 45805725-45805725 on Build GRCh38
rs78455013 Chr.11: 45805629-45805629 on Build GRCh38
rs1006952025 Chr.11: 45805344-45805344 on Build GRCh38

View Full Product Details