Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56160184-56160682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8K5
Entrez Gene ID
219453
Gene Name
olfactory receptor family 8 subfamily K member 5
Gene Aliases
OR11-174
UniGene
Hs.553626
Gene Chromosome Location
Chr.11: 56159394-56160317 on Build GRCh38
Transcripts
NM_001004058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5759023 c.130A>C p.T44P Chr.11: 56160188-56160188 on Build GRCh38
332585 c.42G>T p.L14L Chr.11: 56160276-56160276 on Build GRCh38
5677065 c.4G>T p.G2C Chr.11: 56160314-56160314 on Build GRCh38
7078568 c.42G>A p.L14L Chr.11: 56160276-56160276 on Build GRCh38
1703968 c.85G>A p.G29R Chr.11: 56160233-56160233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371725621 Chr.11: 56160218-56160218 on Build GRCh38
rs756518580 Chr.11: 56160282-56160282 on Build GRCh38
rs137981851 Chr.11: 56160646-56160646 on Build GRCh38
rs1024207788 Chr.11: 56160537-56160537 on Build GRCh38
rs35566076 Chr.11: 56160559-56160559 on Build GRCh38
rs762622030 Chr.11: 56160525-56160525 on Build GRCh38
rs539541909 Chr.11: 56160596-56160596 on Build GRCh38
rs79004948 Chr.11: 56160287-56160287 on Build GRCh38
rs201944075 Chr.11: 56160316-56160316 on Build GRCh38
rs752433521 Chr.11: 56160512-56160512 on Build GRCh38
rs975730404 Chr.11: 56160303-56160303 on Build GRCh38
rs373198496 Chr.11: 56160350-56160350 on Build GRCh38
rs992707736 Chr.11: 56160254-56160254 on Build GRCh38
rs779016262 Chr.11: 56160306-56160306 on Build GRCh38
rs780992867 Chr.11: 56160286-56160286 on Build GRCh38
rs550593579 Chr.11: 56160250-56160250 on Build GRCh38
rs763878778 Chr.11: 56160235-56160235 on Build GRCh38
rs776381884 Chr.11: 56160333-56160333 on Build GRCh38
rs568904823 Chr.11: 56160259-56160259 on Build GRCh38
rs967876982 Chr.11: 56160618-56160618 on Build GRCh38
rs986255747 Chr.11: 56160590-56160590 on Build GRCh38
rs534535341 Chr.11: 56160363-56160363 on Build GRCh38
rs545742657 Chr.11: 56160479-56160479 on Build GRCh38
rs1049321011 Chr.11: 56160633-56160633 on Build GRCh38
rs200707839 Chr.11: 56160273-56160273 on Build GRCh38
rs755656601 Chr.11: 56160305-56160305 on Build GRCh38
rs575315207 Chr.11: 56160435-56160435 on Build GRCh38
rs772919481 Chr.11: 56160358-56160358 on Build GRCh38
rs200434144 Chr.11: 56160274-56160274 on Build GRCh38
rs753165753 Chr.11: 56160279-56160279 on Build GRCh38
rs894268696 Chr.11: 56160513-56160513 on Build GRCh38
rs749836268 Chr.11: 56160293-56160293 on Build GRCh38
rs993447544 Chr.11: 56160389-56160389 on Build GRCh38
rs557042442 Chr.11: 56160430-56160430 on Build GRCh38
rs910321134 Chr.11: 56160594-56160594 on Build GRCh38
rs746060339 Chr.11: 56160307-56160307 on Build GRCh38
rs145701231 Chr.11: 56160222-56160222 on Build GRCh38
rs145476036 Chr.11: 56160260-56160260 on Build GRCh38
rs1002660606 Chr.11: 56160441-56160441 on Build GRCh38
rs956442223 Chr.11: 56160647-56160647 on Build GRCh38
rs371957978 Chr.11: 56160242-56160242 on Build GRCh38
rs967323893 Chr.11: 56160558-56160558 on Build GRCh38
rs910679837 Chr.11: 56160323-56160323 on Build GRCh38
rs1033775292 Chr.11: 56160490-56160490 on Build GRCh38
rs775702535 Chr.11: 56160312-56160312 on Build GRCh38
rs200015077 Chr.11: 56160221-56160221 on Build GRCh38
rs370116389 Chr.11: 56160613-56160613 on Build GRCh38
rs568392891 Chr.11: 56160390-56160390 on Build GRCh38
rs976251565 Chr.11: 56160602-56160602 on Build GRCh38
rs761856106 Chr.11: 56160349-56160349 on Build GRCh38
rs977106909 Chr.11: 56160297-56160297 on Build GRCh38
rs533149542 Chr.11: 56160573-56160573 on Build GRCh38
rs1002868369 Chr.11: 56160501-56160501 on Build GRCh38
rs574184863 Chr.11: 56160372-56160372 on Build GRCh38
rs199988292 Chr.11: 56160301-56160301 on Build GRCh38
rs540496588 Chr.11: 56160611-56160611 on Build GRCh38
rs73472395 Chr.11: 56160574-56160574 on Build GRCh38
rs181891621 Chr.11: 56160419-56160419 on Build GRCh38
rs897900805 Chr.11: 56160498-56160498 on Build GRCh38
rs887740446 Chr.11: 56160645-56160645 on Build GRCh38

View Full Product Details