Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57695813-57696323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC5
Entrez Gene ID
25921
Gene Name
zinc finger DHHC-type containing 5
Gene Aliases
DHHC5, ZNF375
UniGene
Hs.27239
Gene Chromosome Location
Chr.11: 57667836-57701187 on Build GRCh38
Transcripts
NM_015457.2 XM_011544899.1 XM_011544900.1 XM_011544901.1 XM_017017498.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838841 c.955G>A p.D319N Chr.11: 57695989-57695989 on Build GRCh38
6826167 c.901G>A p.E301K Chr.11: 57695935-57695935 on Build GRCh38
5565582 c.965G>A p.R322H Chr.11: 57695999-57695999 on Build GRCh38
3752510 c.994T>C p.L332L Chr.11: 57696028-57696028 on Build GRCh38
270581 c.976T>G p.L326V Chr.11: 57696010-57696010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749964183 Chr.11: 57695949-57695949 on Build GRCh38
rs753762468 Chr.11: 57695936-57695936 on Build GRCh38
rs765267810 Chr.11: 57696025-57696025 on Build GRCh38
rs779621062 Chr.11: 57696296-57696296 on Build GRCh38
rs180891496 Chr.11: 57695865-57695865 on Build GRCh38
rs925392329 Chr.11: 57696139-57696139 on Build GRCh38
rs541057816 Chr.11: 57696221-57696221 on Build GRCh38
rs12804972 Chr.11: 57696295-57696295 on Build GRCh38
rs371229111 Chr.11: 57696066-57696066 on Build GRCh38
rs1057370704 Chr.11: 57696142-57696142 on Build GRCh38
rs965310064 Chr.11: 57695996-57695996 on Build GRCh38
rs760600324 Chr.11: 57696038-57696038 on Build GRCh38
rs185301102 Chr.11: 57695870-57695870 on Build GRCh38
rs760923152 Chr.11: 57696008-57696008 on Build GRCh38
rs746355368 Chr.11: 57696091-57696091 on Build GRCh38
rs377509146 Chr.11: 57695902-57695902 on Build GRCh38
rs149169388 Chr.11: 57695974-57695974 on Build GRCh38
rs1783978 Chr.11: 57696028-57696028 on Build GRCh38
rs771877390 Chr.11: 57695973-57695973 on Build GRCh38
rs779421288 Chr.11: 57696086-57696086 on Build GRCh38
rs376993238 Chr.11: 57696078-57696078 on Build GRCh38
rs894655249 Chr.11: 57696158-57696158 on Build GRCh38
rs778330860 Chr.11: 57696035-57696035 on Build GRCh38
rs377749811 Chr.11: 57695964-57695964 on Build GRCh38
rs373354979 Chr.11: 57695878-57695878 on Build GRCh38
rs776010427 Chr.11: 57695875-57695877 on Build GRCh38
rs191198611 Chr.11: 57696101-57696101 on Build GRCh38
rs779602654 Chr.11: 57695969-57695969 on Build GRCh38
rs143307006 Chr.11: 57695991-57695991 on Build GRCh38
rs537204462 Chr.11: 57696113-57696113 on Build GRCh38
rs751210699 Chr.11: 57696058-57696058 on Build GRCh38
rs1015639946 Chr.11: 57695909-57695909 on Build GRCh38
rs759645860 Chr.11: 57695998-57695998 on Build GRCh38
rs772093043 Chr.11: 57696002-57696002 on Build GRCh38
rs1043202813 Chr.11: 57696203-57696203 on Build GRCh38
rs763930944 Chr.11: 57695924-57695924 on Build GRCh38
rs771442798 Chr.11: 57695988-57695988 on Build GRCh38
rs147331802 Chr.11: 57696014-57696014 on Build GRCh38
rs755076437 Chr.11: 57695852-57695852 on Build GRCh38
rs932482670 Chr.11: 57696167-57696167 on Build GRCh38
rs115730307 Chr.11: 57696206-57696206 on Build GRCh38
rs778169032 Chr.11: 57695975-57695975 on Build GRCh38
rs552673847 Chr.11: 57696246-57696246 on Build GRCh38
rs778619518 Chr.11: 57695947-57695947 on Build GRCh38
rs1003482938 Chr.11: 57695846-57695846 on Build GRCh38
rs759556135 Chr.11: 57695882-57695882 on Build GRCh38
rs775626241 Chr.11: 57696005-57696005 on Build GRCh38
rs547457280 Chr.11: 57695982-57695982 on Build GRCh38
rs1035843689 Chr.11: 57695850-57695850 on Build GRCh38
rs765889349 Chr.11: 57696043-57696043 on Build GRCh38
rs1007883807 Chr.11: 57695908-57695908 on Build GRCh38
rs983840202 Chr.11: 57696053-57696053 on Build GRCh38
rs992424381 Chr.11: 57696055-57696055 on Build GRCh38
rs369629295 Chr.11: 57695904-57695904 on Build GRCh38
rs774774294 Chr.11: 57695992-57695992 on Build GRCh38
rs1049581439 Chr.11: 57696185-57696185 on Build GRCh38
rs112252367 Chr.11: 57695951-57695951 on Build GRCh38
rs373313939 Chr.11: 57696075-57696075 on Build GRCh38
rs753649128 Chr.11: 57696019-57696019 on Build GRCh38
rs374103668 Chr.11: 57695941-57695941 on Build GRCh38
rs969125169 Chr.11: 57695934-57695934 on Build GRCh38
rs140343860 Chr.11: 57696024-57696024 on Build GRCh38
rs558845736 Chr.11: 57696157-57696157 on Build GRCh38
rs1013049472 Chr.11: 57696181-57696181 on Build GRCh38
rs865882231 Chr.11: 57696111-57696111 on Build GRCh38
rs903666793 Chr.11: 57695838-57695838 on Build GRCh38
rs938965962 Chr.11: 57696127-57696127 on Build GRCh38
rs780651593 Chr.11: 57696074-57696074 on Build GRCh38
rs1036971212 Chr.11: 57696288-57696288 on Build GRCh38

View Full Product Details