Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57743458-57744000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD18
Entrez Gene ID
643376
Gene Name
BTB domain containing 18
Gene Aliases
-
UniGene
Hs.606355
Gene Chromosome Location
Chr.11: 57743514-57753176 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NM_001321335.1 NM_170746.3 NR_037646.1

Gene Symbol
C11orf31
Entrez Gene ID
280636
Gene Name
chromosome 11 open reading frame 31
Gene Aliases
C17orf10, SELH
UniGene
Hs.655373
Gene Chromosome Location
Chr.11: 57741250-57743554 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NM_001321335.1 NM_170746.3 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NM_001321335.1 NM_170746.3 NR_037646.1

SNP Detail

SNP ID Chromosome Location
rs189094269 Chr.11: 57743699-57743699 on Build GRCh38
rs1043117198 Chr.11: 57743517-57743517 on Build GRCh38
rs569713973 Chr.11: 57743550-57743551 on Build GRCh38
rs947552596 Chr.11: 57743584-57743584 on Build GRCh38
rs543279451 Chr.11: 57743881-57743881 on Build GRCh38
rs370927769 Chr.11: 57743969-57743969 on Build GRCh38
rs577860701 Chr.11: 57743554-57743554 on Build GRCh38
rs549890818 Chr.11: 57743515-57743515 on Build GRCh38
rs537987220 Chr.11: 57743503-57743503 on Build GRCh38
rs764483153 Chr.11: 57743611-57743611 on Build GRCh38
rs1053131522 Chr.11: 57743958-57743958 on Build GRCh38
rs960281278 Chr.11: 57743863-57743863 on Build GRCh38
rs1053482727 Chr.11: 57743556-57743556 on Build GRCh38
rs913411149 Chr.11: 57743569-57743569 on Build GRCh38
rs372784044 Chr.11: 57743558-57743558 on Build GRCh38
rs899000468 Chr.11: 57743797-57743797 on Build GRCh38
rs377220984 Chr.11: 57743855-57743855 on Build GRCh38
rs993056410 Chr.11: 57743876-57743877 on Build GRCh38
rs889590435 Chr.11: 57743610-57743610 on Build GRCh38
rs1005289624 Chr.11: 57743644-57743644 on Build GRCh38
rs948851810 Chr.11: 57743886-57743886 on Build GRCh38
rs994631381 Chr.11: 57743811-57743811 on Build GRCh38
rs535500319 Chr.11: 57743589-57743590 on Build GRCh38
rs545203585 Chr.11: 57743663-57743663 on Build GRCh38
rs972419187 Chr.11: 57743854-57743854 on Build GRCh38
rs182359529 Chr.11: 57743507-57743507 on Build GRCh38
rs772926563 Chr.11: 57743684-57743684 on Build GRCh38
rs947545452 Chr.11: 57743950-57743950 on Build GRCh38
rs916041570 Chr.11: 57743494-57743494 on Build GRCh38
rs761931807 Chr.11: 57743773-57743773 on Build GRCh38
rs934658481 Chr.11: 57743556-57743557 on Build GRCh38
rs1016667931 Chr.11: 57743685-57743685 on Build GRCh38
rs1023468719 Chr.11: 57743825-57743825 on Build GRCh38
rs764301105 Chr.11: 57743567-57743567 on Build GRCh38
rs969326263 Chr.11: 57743862-57743862 on Build GRCh38
rs770747825 Chr.11: 57743946-57743949 on Build GRCh38
rs978940012 Chr.11: 57743961-57743961 on Build GRCh38
rs914964934 Chr.11: 57743942-57743942 on Build GRCh38
rs900724861 Chr.11: 57743682-57743682 on Build GRCh38
rs760299808 Chr.11: 57743905-57743905 on Build GRCh38
rs555352298 Chr.11: 57743911-57743911 on Build GRCh38
rs982009625 Chr.11: 57743879-57743879 on Build GRCh38
rs1051749160 Chr.11: 57743696-57743696 on Build GRCh38
rs1005395513 Chr.11: 57743710-57743710 on Build GRCh38
rs935101442 Chr.11: 57743519-57743519 on Build GRCh38
rs1014980180 Chr.11: 57743755-57743755 on Build GRCh38

View Full Product Details