Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57795485-57795992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNND1
Entrez Gene ID
1500
Gene Name
catenin delta 1
Gene Aliases
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57761762-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001085458.1 NM_001085459.1 NM_001085460.1 NM_001085461.1 NM_001085462.1 NM_001085463.1 NM_001085464.1 NM_001085465.1 NM_001085466.1 NM_001085467.1 NM_001085468.1 NM_001085469.1 NM_001206883.1 NM_001206884.1 NM_001206885.1 NM_001206886.1 NM_001206887.1 NM_001206888.1 NM_001206889.1 NM_001206890.1 NM_001206891.1 NM_001331.2 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637603 c.407G>A p.R136H Chr.11: 57795716-57795716 on Build GRCh38
6069509 c.302G>C p.R101T Chr.11: 57795611-57795611 on Build GRCh38
1561337 c.420G>A p.T140T Chr.11: 57795729-57795729 on Build GRCh38
6637604 c.407G>A p.R136H Chr.11: 57795716-57795716 on Build GRCh38
467006 c.286T>G p.Y96D Chr.11: 57795595-57795595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570657052 Chr.11: 57795914-57795914 on Build GRCh38
rs201712948 Chr.11: 57795644-57795644 on Build GRCh38
rs750877877 Chr.11: 57795625-57795625 on Build GRCh38
rs548036079 Chr.11: 57795781-57795781 on Build GRCh38
rs1055430896 Chr.11: 57795709-57795709 on Build GRCh38
rs1045472661 Chr.11: 57795667-57795667 on Build GRCh38
rs61754546 Chr.11: 57795577-57795577 on Build GRCh38
rs1013176954 Chr.11: 57795764-57795764 on Build GRCh38
rs770019209 Chr.11: 57795584-57795584 on Build GRCh38
rs189718729 Chr.11: 57795712-57795712 on Build GRCh38
rs776719539 Chr.11: 57795573-57795573 on Build GRCh38
rs767448387 Chr.11: 57795713-57795713 on Build GRCh38
rs759450868 Chr.11: 57795617-57795617 on Build GRCh38
rs867108561 Chr.11: 57795716-57795716 on Build GRCh38
rs747155752 Chr.11: 57795548-57795548 on Build GRCh38
rs773407759 Chr.11: 57795647-57795647 on Build GRCh38
rs764307328 Chr.11: 57795741-57795741 on Build GRCh38
rs923218246 Chr.11: 57795963-57795963 on Build GRCh38
rs966060880 Chr.11: 57795846-57795846 on Build GRCh38
rs200594061 Chr.11: 57795728-57795728 on Build GRCh38
rs762711712 Chr.11: 57795599-57795599 on Build GRCh38
rs60763743 Chr.11: 57795955-57795955 on Build GRCh38
rs757802088 Chr.11: 57795893-57795893 on Build GRCh38
rs760562678 Chr.11: 57795949-57795949 on Build GRCh38
rs756912729 Chr.11: 57795742-57795742 on Build GRCh38
rs201815246 Chr.11: 57795646-57795646 on Build GRCh38
rs758832847 Chr.11: 57795626-57795626 on Build GRCh38
rs147401852 Chr.11: 57795631-57795631 on Build GRCh38
rs981640392 Chr.11: 57795929-57795929 on Build GRCh38
rs978181150 Chr.11: 57795537-57795537 on Build GRCh38
rs749144617 Chr.11: 57795670-57795670 on Build GRCh38
rs773033117 Chr.11: 57795586-57795586 on Build GRCh38
rs1019390064 Chr.11: 57795545-57795545 on Build GRCh38
rs974593994 Chr.11: 57795671-57795671 on Build GRCh38
rs932957523 Chr.11: 57795710-57795710 on Build GRCh38
rs774901816 Chr.11: 57795715-57795715 on Build GRCh38
rs367954206 Chr.11: 57795627-57795627 on Build GRCh38
rs755130916 Chr.11: 57795557-57795558 on Build GRCh38
rs774265957 Chr.11: 57795610-57795610 on Build GRCh38
rs199743391 Chr.11: 57795623-57795623 on Build GRCh38
rs376747640 Chr.11: 57795876-57795876 on Build GRCh38
rs967213262 Chr.11: 57795533-57795533 on Build GRCh38
rs964488408 Chr.11: 57795933-57795933 on Build GRCh38
rs11570193 Chr.11: 57795824-57795824 on Build GRCh38
rs374433581 Chr.11: 57795719-57795719 on Build GRCh38
rs974949339 Chr.11: 57795664-57795664 on Build GRCh38
rs770713347 Chr.11: 57795690-57795690 on Build GRCh38
rs112509051 Chr.11: 57795848-57795848 on Build GRCh38
rs766873356 Chr.11: 57795753-57795753 on Build GRCh38
rs200896049 Chr.11: 57795769-57795769 on Build GRCh38
rs751663613 Chr.11: 57795832-57795832 on Build GRCh38
rs1032088946 Chr.11: 57795865-57795865 on Build GRCh38
rs187946222 Chr.11: 57795919-57795919 on Build GRCh38
rs755749778 Chr.11: 57795624-57795624 on Build GRCh38
rs922956469 Chr.11: 57795686-57795686 on Build GRCh38
rs768626826 Chr.11: 57795565-57795565 on Build GRCh38
rs780696161 Chr.11: 57795539-57795539 on Build GRCh38
rs188146375 Chr.11: 57795693-57795693 on Build GRCh38
rs375424851 Chr.11: 57795640-57795640 on Build GRCh38
rs371716100 Chr.11: 57795602-57795602 on Build GRCh38
rs1000050338 Chr.11: 57795807-57795807 on Build GRCh38
rs937041315 Chr.11: 57795954-57795954 on Build GRCh38
rs367658814 Chr.11: 57795535-57795535 on Build GRCh38

View Full Product Details