Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58440084-58440581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5B12
Entrez Gene ID
390191
Gene Name
olfactory receptor family 5 subfamily B member 12
Gene Aliases
OR11-241, OR5B12P, OR5B16, OST743
UniGene
Hs.553754
Gene Chromosome Location
Chr.11: 58439120-58440173 on Build GRCh38
Transcripts
NM_001004733.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6069429 c.40G>T p.G14W Chr.11: 58440112-58440112 on Build GRCh38
5632498 c.56C>A p.P19Q Chr.11: 58440096-58440096 on Build GRCh38
369413 c.46A>C p.T16P Chr.11: 58440106-58440106 on Build GRCh38
4489598 c.34C>T p.L12F Chr.11: 58440118-58440118 on Build GRCh38
6069428 c.35T>A p.L12H Chr.11: 58440117-58440117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199719285 Chr.11: 58440155-58440155 on Build GRCh38
rs552473613 Chr.11: 58440417-58440417 on Build GRCh38
rs889666408 Chr.11: 58440409-58440409 on Build GRCh38
rs536644165 Chr.11: 58440123-58440123 on Build GRCh38
rs888517336 Chr.11: 58440529-58440529 on Build GRCh38
rs193192986 Chr.11: 58440385-58440385 on Build GRCh38
rs367865017 Chr.11: 58440187-58440187 on Build GRCh38
rs1001125599 Chr.11: 58440131-58440131 on Build GRCh38
rs976805559 Chr.11: 58440192-58440192 on Build GRCh38
rs960564125 Chr.11: 58440303-58440303 on Build GRCh38
rs58320648 Chr.11: 58440427-58440427 on Build GRCh38
rs778661189 Chr.11: 58440152-58440152 on Build GRCh38
rs575186963 Chr.11: 58440540-58440540 on Build GRCh38
rs369935408 Chr.11: 58440119-58440119 on Build GRCh38
rs1022478899 Chr.11: 58440451-58440451 on Build GRCh38
rs150484733 Chr.11: 58440311-58440311 on Build GRCh38
rs529863747 Chr.11: 58440174-58440174 on Build GRCh38
rs188705980 Chr.11: 58440208-58440208 on Build GRCh38
rs763525761 Chr.11: 58440312-58440312 on Build GRCh38
rs879320070 Chr.11: 58440118-58440118 on Build GRCh38
rs532161365 Chr.11: 58440286-58440286 on Build GRCh38
rs753771655 Chr.11: 58440274-58440274 on Build GRCh38
rs866401486 Chr.11: 58440148-58440148 on Build GRCh38
rs753933076 Chr.11: 58440139-58440139 on Build GRCh38
rs746856039 Chr.11: 58440172-58440172 on Build GRCh38
rs116983618 Chr.11: 58440137-58440137 on Build GRCh38
rs766728375 Chr.11: 58440110-58440110 on Build GRCh38
rs760017399 Chr.11: 58440120-58440120 on Build GRCh38
rs941191661 Chr.11: 58440377-58440377 on Build GRCh38
rs745807055 Chr.11: 58440154-58440154 on Build GRCh38
rs200547513 Chr.11: 58440130-58440130 on Build GRCh38
rs1027031144 Chr.11: 58440233-58440233 on Build GRCh38
rs889688169 Chr.11: 58440168-58440168 on Build GRCh38
rs11229459 Chr.11: 58440552-58440552 on Build GRCh38
rs370094120 Chr.11: 58440194-58440194 on Build GRCh38
rs553030778 Chr.11: 58440408-58440408 on Build GRCh38
rs952889595 Chr.11: 58440258-58440258 on Build GRCh38
rs909687949 Chr.11: 58440318-58440318 on Build GRCh38
rs369914220 Chr.11: 58440169-58440169 on Build GRCh38
rs796843893 Chr.11: 58440402-58440402 on Build GRCh38
rs111706434 Chr.11: 58440399-58440399 on Build GRCh38
rs556868332 Chr.11: 58440491-58440491 on Build GRCh38
rs994178914 Chr.11: 58440230-58440230 on Build GRCh38
rs1027044508 Chr.11: 58440508-58440508 on Build GRCh38
rs774934108 Chr.11: 58440112-58440112 on Build GRCh38
rs1040949094 Chr.11: 58440445-58440445 on Build GRCh38
rs945025077 Chr.11: 58440373-58440373 on Build GRCh38
rs757366343 Chr.11: 58440151-58440151 on Build GRCh38
rs897123551 Chr.11: 58440452-58440452 on Build GRCh38
rs112694361 Chr.11: 58440462-58440462 on Build GRCh38
rs751191177 Chr.11: 58440384-58440384 on Build GRCh38

View Full Product Details