Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60927554-60928051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132A
Entrez Gene ID
54972
Gene Name
transmembrane protein 132A
Gene Aliases
GBP, HSPA5BP1
UniGene
Hs.118552
Gene Chromosome Location
Chr.11: 60924441-60937159 on Build GRCh38
Transcripts
NM_017870.3 NM_178031.2 XM_017017951.1 XM_017017952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5959080 c.488delA p.H163fs*94 Chr.11: 60927813-60927813 on Build GRCh38
2036662 c.530T>G p.F177C Chr.11: 60927855-60927855 on Build GRCh38
352400 c.420G>T p.R140R Chr.11: 60927745-60927745 on Build GRCh38
4034485 c.473C>T p.P158L Chr.11: 60927798-60927798 on Build GRCh38
6069748 c.474C>T p.P158P Chr.11: 60927799-60927799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775143525 Chr.11: 60927901-60927901 on Build GRCh38
rs576747366 Chr.11: 60927923-60927923 on Build GRCh38
rs754247987 Chr.11: 60927617-60927617 on Build GRCh38
rs751022386 Chr.11: 60927596-60927596 on Build GRCh38
rs572475620 Chr.11: 60928018-60928018 on Build GRCh38
rs745563168 Chr.11: 60927863-60927863 on Build GRCh38
rs781189570 Chr.11: 60927830-60927830 on Build GRCh38
rs772408410 Chr.11: 60927649-60927649 on Build GRCh38
rs762563313 Chr.11: 60927590-60927590 on Build GRCh38
rs770668657 Chr.11: 60927851-60927851 on Build GRCh38
rs778078821 Chr.11: 60927611-60927611 on Build GRCh38
rs202235322 Chr.11: 60927824-60927824 on Build GRCh38
rs775123362 Chr.11: 60927798-60927798 on Build GRCh38
rs748152203 Chr.11: 60927739-60927739 on Build GRCh38
rs767027606 Chr.11: 60927675-60927675 on Build GRCh38
rs1020654105 Chr.11: 60927728-60927728 on Build GRCh38
rs531170475 Chr.11: 60928031-60928031 on Build GRCh38
rs576494449 Chr.11: 60927925-60927925 on Build GRCh38
rs144441365 Chr.11: 60927852-60927852 on Build GRCh38
rs373031720 Chr.11: 60927695-60927695 on Build GRCh38
rs545657824 Chr.11: 60927924-60927924 on Build GRCh38
rs757741829 Chr.11: 60927620-60927620 on Build GRCh38
rs756758518 Chr.11: 60927608-60927609 on Build GRCh38
rs1011811778 Chr.11: 60927823-60927823 on Build GRCh38
rs773742701 Chr.11: 60927710-60927710 on Build GRCh38
rs1052927250 Chr.11: 60927788-60927788 on Build GRCh38
rs997783537 Chr.11: 60927879-60927879 on Build GRCh38
rs755894631 Chr.11: 60927967-60927967 on Build GRCh38
rs572038760 Chr.11: 60927719-60927719 on Build GRCh38
rs970556780 Chr.11: 60927942-60927942 on Build GRCh38
rs368067544 Chr.11: 60927849-60927849 on Build GRCh38
rs754114057 Chr.11: 60927801-60927801 on Build GRCh38
rs753749618 Chr.11: 60927932-60927932 on Build GRCh38
rs139246153 Chr.11: 60927744-60927744 on Build GRCh38
rs903604983 Chr.11: 60927734-60927734 on Build GRCh38
rs771695533 Chr.11: 60927893-60927893 on Build GRCh38
rs980119298 Chr.11: 60927944-60927946 on Build GRCh38
rs374191784 Chr.11: 60927806-60927806 on Build GRCh38
rs891640438 Chr.11: 60927800-60927800 on Build GRCh38
rs765943930 Chr.11: 60927592-60927592 on Build GRCh38
rs942427455 Chr.11: 60927994-60927994 on Build GRCh38
rs766999372 Chr.11: 60927793-60927793 on Build GRCh38
rs750847310 Chr.11: 60927821-60927821 on Build GRCh38
rs373484312 Chr.11: 60927700-60927700 on Build GRCh38
rs919298396 Chr.11: 60927816-60927816 on Build GRCh38
rs769618210 Chr.11: 60927660-60927660 on Build GRCh38
rs765770473 Chr.11: 60927807-60927807 on Build GRCh38
rs748170489 Chr.11: 60927834-60927834 on Build GRCh38
rs538667454 Chr.11: 60927651-60927651 on Build GRCh38
rs1019617213 Chr.11: 60927761-60927761 on Build GRCh38
rs758786158 Chr.11: 60927822-60927822 on Build GRCh38
rs986376500 Chr.11: 60927757-60927757 on Build GRCh38
rs756662250 Chr.11: 60927598-60927598 on Build GRCh38
rs770652755 Chr.11: 60927745-60927745 on Build GRCh38
rs144704995 Chr.11: 60927654-60927654 on Build GRCh38
rs777651889 Chr.11: 60927846-60927846 on Build GRCh38
rs114893240 Chr.11: 60927691-60927691 on Build GRCh38
rs762654207 Chr.11: 60927668-60927668 on Build GRCh38
rs1000589341 Chr.11: 60927990-60927990 on Build GRCh38
rs1049524616 Chr.11: 60927996-60927996 on Build GRCh38
rs559330435 Chr.11: 60927956-60927956 on Build GRCh38
rs772988359 Chr.11: 60927661-60927661 on Build GRCh38
rs145457758 Chr.11: 60927715-60927715 on Build GRCh38
rs766006282 Chr.11: 60927579-60927579 on Build GRCh38
rs1016386518 Chr.11: 60927938-60927938 on Build GRCh38
rs751089802 Chr.11: 60927671-60927671 on Build GRCh38
rs370154055 Chr.11: 60927779-60927779 on Build GRCh38
rs889249211 Chr.11: 60927998-60927998 on Build GRCh38
rs369462368 Chr.11: 60927837-60927837 on Build GRCh38
rs746155825 Chr.11: 60927648-60927648 on Build GRCh38
rs575955 Chr.11: 60927903-60927903 on Build GRCh38
rs928658470 Chr.11: 60927955-60927955 on Build GRCh38
rs1008147554 Chr.11: 60928009-60928009 on Build GRCh38
rs201027775 Chr.11: 60927962-60927963 on Build GRCh38
rs554324495 Chr.11: 60927803-60927803 on Build GRCh38
rs758698043 Chr.11: 60927701-60927701 on Build GRCh38
rs965002361 Chr.11: 60927865-60927865 on Build GRCh38
rs367962515 Chr.11: 60927975-60927975 on Build GRCh38
rs769708258 Chr.11: 60927740-60927740 on Build GRCh38
rs944724062 Chr.11: 60927610-60927610 on Build GRCh38
rs768669264 Chr.11: 60927653-60927653 on Build GRCh38
rs180968764 Chr.11: 60927957-60927957 on Build GRCh38
rs780546391 Chr.11: 60927872-60927873 on Build GRCh38
rs575123 Chr.11: 60927829-60927829 on Build GRCh38
rs773919866 Chr.11: 60927772-60927772 on Build GRCh38
rs765892046 Chr.11: 60927670-60927670 on Build GRCh38
rs758917002 Chr.11: 60927674-60927674 on Build GRCh38
rs376179392 Chr.11: 60927714-60927714 on Build GRCh38
rs79330054 Chr.11: 60927962-60927962 on Build GRCh38
rs757832131 Chr.11: 60927692-60927692 on Build GRCh38
rs779387851 Chr.11: 60927624-60927624 on Build GRCh38
rs374551089 Chr.11: 60927743-60927743 on Build GRCh38

View Full Product Details