Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64258388-64258891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCB3
Entrez Gene ID
5331
Gene Name
phospholipase C beta 3
Gene Aliases
-
UniGene
Hs.523761
Gene Chromosome Location
Chr.11: 64251523-64269452 on Build GRCh38
Transcripts
NM_000932.2 NM_001184883.1 NM_001316314.1 XM_011545101.2 XM_017017925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4715014 c.1092C>T p.D364D Chr.11: 64258552-64258552 on Build GRCh38
4715015 c.1093G>A p.V365M Chr.11: 64258553-64258553 on Build GRCh38
1355653 c.1125C>A p.P375P Chr.11: 64258585-64258585 on Build GRCh38
4035077 c.1080C>T p.C360C Chr.11: 64258540-64258540 on Build GRCh38
7100218 c.1114G>A p.E372K Chr.11: 64258574-64258574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747343590 Chr.11: 64258492-64258492 on Build GRCh38
rs765514797 Chr.11: 64258623-64258623 on Build GRCh38
rs760504379 Chr.11: 64258459-64258459 on Build GRCh38
rs759420179 Chr.11: 64258708-64258708 on Build GRCh38
rs139714714 Chr.11: 64258519-64258519 on Build GRCh38
rs372855408 Chr.11: 64258753-64258753 on Build GRCh38
rs770882924 Chr.11: 64258697-64258697 on Build GRCh38
rs980340371 Chr.11: 64258798-64258798 on Build GRCh38
rs779837194 Chr.11: 64258497-64258497 on Build GRCh38
rs145872781 Chr.11: 64258682-64258682 on Build GRCh38
rs202112716 Chr.11: 64258569-64258569 on Build GRCh38
rs752334609 Chr.11: 64258848-64258848 on Build GRCh38
rs775319705 Chr.11: 64258428-64258428 on Build GRCh38
rs2244625 Chr.11: 64258672-64258672 on Build GRCh38
rs771668873 Chr.11: 64258680-64258680 on Build GRCh38
rs750203129 Chr.11: 64258826-64258826 on Build GRCh38
rs762545833 Chr.11: 64258440-64258440 on Build GRCh38
rs530167219 Chr.11: 64258654-64258654 on Build GRCh38
rs757989609 Chr.11: 64258850-64258850 on Build GRCh38
rs758787013 Chr.11: 64258474-64258474 on Build GRCh38
rs762931747 Chr.11: 64258725-64258725 on Build GRCh38
rs1039380142 Chr.11: 64258652-64258652 on Build GRCh38
rs773924319 Chr.11: 64258443-64258443 on Build GRCh38
rs941694561 Chr.11: 64258540-64258540 on Build GRCh38
rs200287886 Chr.11: 64258448-64258448 on Build GRCh38
rs910387920 Chr.11: 64258534-64258534 on Build GRCh38
rs777221475 Chr.11: 64258853-64258853 on Build GRCh38
rs2244621 Chr.11: 64258747-64258747 on Build GRCh38
rs778415930 Chr.11: 64258847-64258847 on Build GRCh38
rs750785811 Chr.11: 64258751-64258751 on Build GRCh38
rs561312676 Chr.11: 64258630-64258630 on Build GRCh38
rs776358095 Chr.11: 64258704-64258704 on Build GRCh38
rs143268643 Chr.11: 64258531-64258531 on Build GRCh38
rs200850537 Chr.11: 64258765-64258765 on Build GRCh38
rs755062861 Chr.11: 64258761-64258761 on Build GRCh38
rs151101135 Chr.11: 64258597-64258597 on Build GRCh38
rs972126620 Chr.11: 64258803-64258803 on Build GRCh38
rs757760404 Chr.11: 64258495-64258495 on Build GRCh38
rs754101491 Chr.11: 64258509-64258510 on Build GRCh38
rs759711349 Chr.11: 64258622-64258622 on Build GRCh38
rs371015701 Chr.11: 64258713-64258713 on Build GRCh38
rs369922076 Chr.11: 64258442-64258442 on Build GRCh38
rs193263245 Chr.11: 64258804-64258804 on Build GRCh38
rs149771242 Chr.11: 64258748-64258748 on Build GRCh38
rs766584939 Chr.11: 64258462-64258462 on Build GRCh38
rs758548996 Chr.11: 64258631-64258631 on Build GRCh38
rs563411880 Chr.11: 64258739-64258739 on Build GRCh38
rs958982043 Chr.11: 64258446-64258446 on Build GRCh38
rs943269665 Chr.11: 64258639-64258639 on Build GRCh38
rs963067556 Chr.11: 64258819-64258819 on Build GRCh38
rs765350654 Chr.11: 64258712-64258712 on Build GRCh38
rs113115779 Chr.11: 64258466-64258466 on Build GRCh38
rs748140537 Chr.11: 64258857-64258858 on Build GRCh38
rs373432207 Chr.11: 64258570-64258570 on Build GRCh38
rs375154491 Chr.11: 64258465-64258465 on Build GRCh38
rs779019666 Chr.11: 64258849-64258850 on Build GRCh38
rs747606768 Chr.11: 64258677-64258677 on Build GRCh38
rs141037173 Chr.11: 64258651-64258651 on Build GRCh38
rs2532580 Chr.11: 64258412-64258412 on Build GRCh38
rs910358213 Chr.11: 64258855-64258855 on Build GRCh38
rs766103292 Chr.11: 64258585-64258585 on Build GRCh38
rs746878875 Chr.11: 64258696-64258696 on Build GRCh38
rs182275817 Chr.11: 64258414-64258414 on Build GRCh38
rs535308048 Chr.11: 64258808-64258808 on Build GRCh38
rs929120823 Chr.11: 64258584-64258584 on Build GRCh38
rs766776483 Chr.11: 64258423-64258425 on Build GRCh38
rs754118757 Chr.11: 64258463-64258463 on Build GRCh38
rs760447479 Chr.11: 64258571-64258571 on Build GRCh38
rs547914518 Chr.11: 64258809-64258809 on Build GRCh38
rs933143441 Chr.11: 64258481-64258481 on Build GRCh38
rs377028452 Chr.11: 64258633-64258633 on Build GRCh38
rs185738787 Chr.11: 64258830-64258830 on Build GRCh38
rs748890795 Chr.11: 64258511-64258511 on Build GRCh38
rs778667804 Chr.11: 64258673-64258673 on Build GRCh38
rs746266616 Chr.11: 64258671-64258671 on Build GRCh38
rs868377071 Chr.11: 64258805-64258805 on Build GRCh38
rs751696241 Chr.11: 64258740-64258740 on Build GRCh38
rs1034475605 Chr.11: 64258724-64258724 on Build GRCh38
rs188590056 Chr.11: 64258525-64258525 on Build GRCh38
rs747742492 Chr.11: 64258552-64258552 on Build GRCh38
rs918989536 Chr.11: 64258831-64258831 on Build GRCh38
rs748457177 Chr.11: 64258868-64258868 on Build GRCh38
rs750163336 Chr.11: 64258449-64258449 on Build GRCh38

View Full Product Details