Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64789623-64790119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP4K2
Entrez Gene ID
5871
Gene Name
mitogen-activated protein kinase kinase kinase kinase 2
Gene Aliases
BL44, GCK, RAB8IP
UniGene
Hs.534341
Gene Chromosome Location
Chr.11: 64786914-64803254 on Build GRCh38
Transcripts
NM_001307990.1 NM_004579.4 XM_011545203.2 XM_011545204.2 XM_017018093.1 XM_017018094.1 XM_017018095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3809978 c.2263A>T p.S755C Chr.11: 64789945-64789945 on Build GRCh38
6437343 c.2299C>T p.R767* Chr.11: 64789909-64789909 on Build GRCh38
3770208 c.2324C>T p.T775I Chr.11: 64789781-64789781 on Build GRCh38
930199 c.2289G>T p.G763G Chr.11: 64789919-64789919 on Build GRCh38
1298389 c.2357G>A p.R786Q Chr.11: 64789748-64789748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757700736 Chr.11: 64790000-64790000 on Build GRCh38
rs766290220 Chr.11: 64789794-64789796 on Build GRCh38
rs767485134 Chr.11: 64789795-64789795 on Build GRCh38
rs769304982 Chr.11: 64789828-64789828 on Build GRCh38
rs779146993 Chr.11: 64789829-64789829 on Build GRCh38
rs144455485 Chr.11: 64789937-64789937 on Build GRCh38
rs773457675 Chr.11: 64789748-64789748 on Build GRCh38
rs750165069 Chr.11: 64789665-64789665 on Build GRCh38
rs559023055 Chr.11: 64790026-64790026 on Build GRCh38
rs929299451 Chr.11: 64789956-64789956 on Build GRCh38
rs755948934 Chr.11: 64789813-64789813 on Build GRCh38
rs760539615 Chr.11: 64789887-64789887 on Build GRCh38
rs565609247 Chr.11: 64789848-64789848 on Build GRCh38
rs534640096 Chr.11: 64789849-64789849 on Build GRCh38
rs763613582 Chr.11: 64789785-64789785 on Build GRCh38
rs773799222 Chr.11: 64789872-64789872 on Build GRCh38
rs539008803 Chr.11: 64790023-64790023 on Build GRCh38
rs541401417 Chr.11: 64790054-64790054 on Build GRCh38
rs771313952 Chr.11: 64789860-64789860 on Build GRCh38
rs761373464 Chr.11: 64789874-64789874 on Build GRCh38
rs773169513 Chr.11: 64789893-64789893 on Build GRCh38
rs748624939 Chr.11: 64789729-64789729 on Build GRCh38
rs755204338 Chr.11: 64789825-64789825 on Build GRCh38
rs758485508 Chr.11: 64789985-64789985 on Build GRCh38
rs761656511 Chr.11: 64789793-64789793 on Build GRCh38
rs577927685 Chr.11: 64790010-64790020 on Build GRCh38
rs752781827 Chr.11: 64789979-64789979 on Build GRCh38
rs775105122 Chr.11: 64789766-64789766 on Build GRCh38
rs200966022 Chr.11: 64789814-64789814 on Build GRCh38
rs753702381 Chr.11: 64789794-64789794 on Build GRCh38
rs373345505 Chr.11: 64789966-64789966 on Build GRCh38
rs535931498 Chr.11: 64789942-64789942 on Build GRCh38
rs982593684 Chr.11: 64789730-64789730 on Build GRCh38
rs935494939 Chr.11: 64789908-64789908 on Build GRCh38
rs1052564481 Chr.11: 64789943-64789943 on Build GRCh38
rs943749513 Chr.11: 64789840-64789840 on Build GRCh38
rs758898376 Chr.11: 64789839-64789839 on Build GRCh38
rs756019998 Chr.11: 64789788-64789788 on Build GRCh38
rs750243003 Chr.11: 64789806-64789806 on Build GRCh38
rs1039260843 Chr.11: 64790018-64790018 on Build GRCh38
rs536745553 Chr.11: 64789996-64789996 on Build GRCh38
rs201824736 Chr.11: 64789711-64789711 on Build GRCh38
rs905162631 Chr.11: 64790019-64790019 on Build GRCh38
rs11231869 Chr.11: 64789660-64789660 on Build GRCh38
rs374475841 Chr.11: 64789653-64789653 on Build GRCh38
rs951281725 Chr.11: 64790060-64790060 on Build GRCh38
rs747399938 Chr.11: 64789749-64789749 on Build GRCh38
rs555576594 Chr.11: 64790064-64790064 on Build GRCh38
rs753629166 Chr.11: 64789957-64789957 on Build GRCh38
rs568199989 Chr.11: 64789989-64789989 on Build GRCh38
rs780408137 Chr.11: 64789707-64789707 on Build GRCh38
rs1009197536 Chr.11: 64789703-64789703 on Build GRCh38
rs201615249 Chr.11: 64790015-64790015 on Build GRCh38
rs779097889 Chr.11: 64789724-64789724 on Build GRCh38
rs1009660615 Chr.11: 64790092-64790092 on Build GRCh38
rs756357562 Chr.11: 64789683-64789683 on Build GRCh38
rs1039326487 Chr.11: 64789850-64789850 on Build GRCh38
rs762549213 Chr.11: 64789783-64789783 on Build GRCh38
rs892140684 Chr.11: 64789685-64789685 on Build GRCh38
rs531938938 Chr.11: 64789714-64789714 on Build GRCh38
rs772573845 Chr.11: 64789733-64789733 on Build GRCh38
rs778583333 Chr.11: 64789839-64789842 on Build GRCh38
rs368874953 Chr.11: 64789654-64789654 on Build GRCh38
rs767225981 Chr.11: 64789662-64789662 on Build GRCh38
rs371939807 Chr.11: 64789658-64789658 on Build GRCh38
rs201087163 Chr.11: 64789868-64789868 on Build GRCh38
rs781484334 Chr.11: 64789864-64789864 on Build GRCh38
rs772981205 Chr.11: 64789836-64789836 on Build GRCh38
rs936138573 Chr.11: 64790042-64790042 on Build GRCh38
rs754738273 Chr.11: 64789796-64789799 on Build GRCh38
rs773848954 Chr.11: 64789789-64789789 on Build GRCh38
rs769496570 Chr.11: 64789755-64789755 on Build GRCh38
rs1025329975 Chr.11: 64789705-64789705 on Build GRCh38
rs748949129 Chr.11: 64789866-64789866 on Build GRCh38
rs71581722 Chr.11: 64790053-64790053 on Build GRCh38
rs145768386 Chr.11: 64790016-64790016 on Build GRCh38
rs759843368 Chr.11: 64789960-64789960 on Build GRCh38
rs750453367 Chr.11: 64789697-64789697 on Build GRCh38
rs775050295 Chr.11: 64789650-64789650 on Build GRCh38
rs371801593 Chr.11: 64789948-64789948 on Build GRCh38
rs896548407 Chr.11: 64789958-64789958 on Build GRCh38
rs764212651 Chr.11: 64790089-64790089 on Build GRCh38
rs372194535 Chr.11: 64789822-64789822 on Build GRCh38

View Full Product Details