Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42246431-42246956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRERF1
Entrez Gene ID
55809
Gene Name
transcriptional regulating factor 1
Gene Aliases
BCAR2, HSA277276, RAPA, TREP132, TReP-132, dJ139D8.5
UniGene
Hs.485392
Gene Chromosome Location
Chr.6: 42224933-42452216 on Build GRCh38
Transcripts
NM_001297573.1 NM_033502.3 XM_011514741.2 XM_011514742.2 XM_011514743.2 XM_011514744.1 XM_011514745.1 XM_017011034.1 XM_017011035.1 XM_017011036.1 XM_017011037.1 XM_017011038.1 XM_017011039.1 XM_017011040.1 XM_017011041.1 XM_017011042.1 XM_017011043.1 XM_017011044.1 XM_017011045.1 XM_017011046.1 XM_017011047.1 XM_017011048.1 XM_017011049.1 XM_017011050.1 XM_017011051.1 XM_017011052.1 XM_017011053.1 XM_017011054.1 XM_017011055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7308109 c.2691G>T p.L897L Chr.6: 42246510-42246510 on Build GRCh38
7293542 c.2739G>T p.Q913H Chr.6: 42246462-42246462 on Build GRCh38
7308111 c.2442G>T p.L814L Chr.6: 42246510-42246510 on Build GRCh38
742956 c.2662G>T p.D888Y Chr.6: 42246539-42246539 on Build GRCh38
3662548 c.2657-2A>G p.? Chr.6: 42246546-42246546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62416670 Chr.6: 42246516-42246516 on Build GRCh38
rs939505060 Chr.6: 42246888-42246888 on Build GRCh38
rs375341976 Chr.6: 42246770-42246770 on Build GRCh38
rs371036609 Chr.6: 42246659-42246659 on Build GRCh38
rs535538645 Chr.6: 42246592-42246592 on Build GRCh38
rs965883591 Chr.6: 42246744-42246744 on Build GRCh38
rs887710851 Chr.6: 42246710-42246710 on Build GRCh38
rs772563023 Chr.6: 42246490-42246490 on Build GRCh38
rs371207024 Chr.6: 42246573-42246573 on Build GRCh38
rs367561480 Chr.6: 42246562-42246562 on Build GRCh38
rs747341498 Chr.6: 42246585-42246585 on Build GRCh38
rs139783030 Chr.6: 42246540-42246540 on Build GRCh38
rs765549717 Chr.6: 42246524-42246524 on Build GRCh38
rs755566392 Chr.6: 42246568-42246568 on Build GRCh38
rs527499127 Chr.6: 42246818-42246818 on Build GRCh38
rs752855299 Chr.6: 42246534-42246534 on Build GRCh38
rs1056530278 Chr.6: 42246903-42246903 on Build GRCh38
rs141876724 Chr.6: 42246930-42246930 on Build GRCh38
rs980228122 Chr.6: 42246733-42246733 on Build GRCh38
rs567432435 Chr.6: 42246874-42246874 on Build GRCh38
rs367948820 Chr.6: 42246811-42246811 on Build GRCh38
rs1015158900 Chr.6: 42246730-42246730 on Build GRCh38
rs773898785 Chr.6: 42246493-42246493 on Build GRCh38
rs915471204 Chr.6: 42246853-42246853 on Build GRCh38
rs143675535 Chr.6: 42246601-42246601 on Build GRCh38
rs938768099 Chr.6: 42246791-42246791 on Build GRCh38
rs1004826565 Chr.6: 42246715-42246715 on Build GRCh38
rs779825165 Chr.6: 42246570-42246570 on Build GRCh38
rs761244152 Chr.6: 42246495-42246495 on Build GRCh38
rs117301162 Chr.6: 42246553-42246553 on Build GRCh38
rs537698342 Chr.6: 42246661-42246661 on Build GRCh38
rs759471906 Chr.6: 42246523-42246523 on Build GRCh38
rs749962758 Chr.6: 42246567-42246567 on Build GRCh38
rs531560106 Chr.6: 42246640-42246640 on Build GRCh38
rs751305767 Chr.6: 42246556-42246556 on Build GRCh38
rs771519986 Chr.6: 42246497-42246497 on Build GRCh38
rs778306955 Chr.6: 42246583-42246583 on Build GRCh38
rs536201004 Chr.6: 42246919-42246919 on Build GRCh38
rs551465866 Chr.6: 42246664-42246664 on Build GRCh38
rs924103817 Chr.6: 42246877-42246877 on Build GRCh38
rs781040997 Chr.6: 42246566-42246566 on Build GRCh38
rs751302520 Chr.6: 42246633-42246633 on Build GRCh38
rs189612433 Chr.6: 42246610-42246610 on Build GRCh38
rs750114759 Chr.6: 42246754-42246754 on Build GRCh38
rs953013971 Chr.6: 42246796-42246796 on Build GRCh38
rs554754643 Chr.6: 42246789-42246789 on Build GRCh38
rs895246687 Chr.6: 42246918-42246918 on Build GRCh38
rs185101021 Chr.6: 42246483-42246483 on Build GRCh38
rs148264189 Chr.6: 42246872-42246872 on Build GRCh38
rs780811993 Chr.6: 42246697-42246697 on Build GRCh38
rs946937773 Chr.6: 42246855-42246855 on Build GRCh38
rs73422437 Chr.6: 42246673-42246673 on Build GRCh38
rs763970470 Chr.6: 42246605-42246605 on Build GRCh38

View Full Product Details