Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89635522-89636068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYRM2
Entrez Gene ID
57226
Gene Name
LYR motif containing 2
Gene Aliases
DJ122O8.2
UniGene
Hs.177275
Gene Chromosome Location
Chr.6: 89632224-89638755 on Build GRCh38
Transcripts
NM_020466.4 NR_028493.1 NR_028494.1 NR_028495.1

SNP Detail

SNP ID Chromosome Location
rs577043917 Chr.6: 89635619-89635619 on Build GRCh38
rs985485843 Chr.6: 89635947-89635947 on Build GRCh38
rs750459190 Chr.6: 89636040-89636040 on Build GRCh38
rs960326924 Chr.6: 89635625-89635625 on Build GRCh38
rs11754238 Chr.6: 89635817-89635817 on Build GRCh38
rs116040148 Chr.6: 89635633-89635633 on Build GRCh38
rs1023750953 Chr.6: 89635908-89635908 on Build GRCh38
rs776161101 Chr.6: 89635670-89635670 on Build GRCh38
rs59902144 Chr.6: 89635744-89635744 on Build GRCh38
rs772740658 Chr.6: 89635595-89635595 on Build GRCh38
rs531781926 Chr.6: 89635695-89635695 on Build GRCh38
rs1050366510 Chr.6: 89636008-89636008 on Build GRCh38
rs776487332 Chr.6: 89635727-89635729 on Build GRCh38
rs960461658 Chr.6: 89635875-89635875 on Build GRCh38
rs763922260 Chr.6: 89635952-89635952 on Build GRCh38
rs571152358 Chr.6: 89636037-89636037 on Build GRCh38
rs761696378 Chr.6: 89635984-89635984 on Build GRCh38
rs568185061 Chr.6: 89635752-89635752 on Build GRCh38
rs568404391 Chr.6: 89635951-89635951 on Build GRCh38
rs1034727225 Chr.6: 89635860-89635860 on Build GRCh38
rs539714803 Chr.6: 89636033-89636033 on Build GRCh38
rs546227829 Chr.6: 89635567-89635567 on Build GRCh38
rs946644140 Chr.6: 89636039-89636039 on Build GRCh38
rs112398023 Chr.6: 89635800-89635800 on Build GRCh38
rs902378166 Chr.6: 89635826-89635826 on Build GRCh38
rs901945037 Chr.6: 89635781-89635781 on Build GRCh38
rs990457714 Chr.6: 89635878-89635878 on Build GRCh38
rs550542844 Chr.6: 89635978-89635978 on Build GRCh38
rs889695650 Chr.6: 89635798-89635802 on Build GRCh38
rs111676769 Chr.6: 89635799-89635799 on Build GRCh38
rs952312370 Chr.6: 89635928-89635928 on Build GRCh38
rs977872205 Chr.6: 89635576-89635578 on Build GRCh38
rs1011262215 Chr.6: 89635801-89635802 on Build GRCh38
rs908059183 Chr.6: 89635705-89635706 on Build GRCh38
rs544681815 Chr.6: 89635686-89635686 on Build GRCh38
rs564694512 Chr.6: 89635613-89635613 on Build GRCh38
rs932095216 Chr.6: 89635999-89635999 on Build GRCh38
rs769425555 Chr.6: 89635755-89635755 on Build GRCh38
rs759660054 Chr.6: 89635913-89635917 on Build GRCh38
rs370308472 Chr.6: 89635722-89635722 on Build GRCh38
rs1050397264 Chr.6: 89635798-89635801 on Build GRCh38
rs540481732 Chr.6: 89635798-89635799 on Build GRCh38
rs547178513 Chr.6: 89635664-89635664 on Build GRCh38
rs67506370 Chr.6: 89635979-89635979 on Build GRCh38
rs183605208 Chr.6: 89636034-89636034 on Build GRCh38
rs999367297 Chr.6: 89635849-89635849 on Build GRCh38
rs12211 Chr.6: 89635810-89635810 on Build GRCh38

View Full Product Details