Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114407680-114408195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM7
Entrez Gene ID
10179
Gene Name
RNA binding motif protein 7
Gene Aliases
-
UniGene
Hs.7527
Gene Chromosome Location
Chr.11: 114400529-114410610 on Build GRCh38
Transcripts
NM_001286045.1 NM_001286046.1 NM_001286047.1 NM_001286048.1 NM_016090.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4840454 c.780A>C p.K260N Chr.11: 114407786-114407786 on Build GRCh38
5970009 c.698A>G p.D233G Chr.11: 114407704-114407704 on Build GRCh38
4597096 c.781T>G p.W261G Chr.11: 114407787-114407787 on Build GRCh38
4018001 c.720G>T p.R240S Chr.11: 114407726-114407726 on Build GRCh38
923298 c.794G>A p.R265Q Chr.11: 114407800-114407800 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1055710318 Chr.11: 114408131-114408131 on Build GRCh38
rs757365374 Chr.11: 114407733-114407733 on Build GRCh38
rs780640128 Chr.11: 114407807-114407808 on Build GRCh38
rs945435056 Chr.11: 114408090-114408090 on Build GRCh38
rs190463336 Chr.11: 114407792-114407792 on Build GRCh38
rs753101100 Chr.11: 114407716-114407716 on Build GRCh38
rs906997644 Chr.11: 114408094-114408094 on Build GRCh38
rs757663369 Chr.11: 114407969-114407969 on Build GRCh38
rs372287494 Chr.11: 114408132-114408132 on Build GRCh38
rs532340098 Chr.11: 114408064-114408064 on Build GRCh38
rs758645695 Chr.11: 114407761-114407761 on Build GRCh38
rs936682145 Chr.11: 114408063-114408063 on Build GRCh38
rs764638996 Chr.11: 114407731-114407731 on Build GRCh38
rs764390303 Chr.11: 114407844-114407844 on Build GRCh38
rs879029123 Chr.11: 114407720-114407720 on Build GRCh38
rs749242719 Chr.11: 114407791-114407791 on Build GRCh38
rs1046484303 Chr.11: 114408093-114408093 on Build GRCh38
rs759112731 Chr.11: 114407814-114407814 on Build GRCh38
rs921545795 Chr.11: 114408042-114408042 on Build GRCh38
rs771544792 Chr.11: 114407915-114407915 on Build GRCh38
rs747561804 Chr.11: 114407855-114407860 on Build GRCh38
rs541868847 Chr.11: 114407797-114407797 on Build GRCh38
rs373571346 Chr.11: 114407790-114407790 on Build GRCh38
rs563511786 Chr.11: 114407930-114407930 on Build GRCh38
rs376509824 Chr.11: 114407833-114407833 on Build GRCh38
rs982540562 Chr.11: 114407955-114407955 on Build GRCh38
rs754434748 Chr.11: 114407732-114407732 on Build GRCh38
rs779148823 Chr.11: 114407754-114407754 on Build GRCh38
rs747955530 Chr.11: 114407772-114407772 on Build GRCh38
rs777222275 Chr.11: 114407859-114407859 on Build GRCh38
rs10790007 Chr.11: 114408169-114408169 on Build GRCh38
rs1003128640 Chr.11: 114408114-114408114 on Build GRCh38
rs780202100 Chr.11: 114407765-114407765 on Build GRCh38
rs777999213 Chr.11: 114407721-114407721 on Build GRCh38
rs575674158 Chr.11: 114407888-114407888 on Build GRCh38
rs965929785 Chr.11: 114407975-114407975 on Build GRCh38
rs773851467 Chr.11: 114407801-114407801 on Build GRCh38
rs199909173 Chr.11: 114407800-114407800 on Build GRCh38
rs544674673 Chr.11: 114407921-114407921 on Build GRCh38
rs758762577 Chr.11: 114407731-114407733 on Build GRCh38
rs769519996 Chr.11: 114407787-114407787 on Build GRCh38
rs754113563 Chr.11: 114407821-114407821 on Build GRCh38
rs771742271 Chr.11: 114407816-114407816 on Build GRCh38
rs555698382 Chr.11: 114407841-114407841 on Build GRCh38
rs559692817 Chr.11: 114408087-114408087 on Build GRCh38
rs975656501 Chr.11: 114407985-114407986 on Build GRCh38
rs750607699 Chr.11: 114407756-114407756 on Build GRCh38
rs757075095 Chr.11: 114408066-114408067 on Build GRCh38
rs754006158 Chr.11: 114408060-114408064 on Build GRCh38
rs545972700 Chr.11: 114408073-114408073 on Build GRCh38

View Full Product Details