Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119084752-119085283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMBS
Entrez Gene ID
3145
Gene Name
hydroxymethylbilane synthase
Gene Aliases
PBG-D, PBGD, PORC, UPS
UniGene
Hs.82609
Gene Chromosome Location
Chr.11: 119084871-119093549 on Build GRCh38
Transcripts
NM_000190.3 NM_001258208.1 NM_001258209.1 XM_005271531.1 XM_005271532.1 XM_005271533.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4884032 c.26C>A p.A9E Chr.11: 119085059-119085059 on Build GRCh38
2106357 c.26C>T p.A9V Chr.11: 119085059-119085059 on Build GRCh38
4935035 c.33+1G>A p.? Chr.11: 119085067-119085067 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782528817 Chr.11: 119085040-119085040 on Build GRCh38
rs1054295505 Chr.11: 119085101-119085101 on Build GRCh38
rs938203141 Chr.11: 119084985-119084985 on Build GRCh38
rs782106780 Chr.11: 119085185-119085186 on Build GRCh38
rs1000008323 Chr.11: 119084903-119084903 on Build GRCh38
rs869088008 Chr.11: 119085199-119085201 on Build GRCh38
rs781938663 Chr.11: 119085021-119085021 on Build GRCh38
rs781849318 Chr.11: 119085005-119085006 on Build GRCh38
rs796367961 Chr.11: 119085200-119085201 on Build GRCh38
rs782231650 Chr.11: 119085138-119085138 on Build GRCh38
rs782646347 Chr.11: 119085154-119085154 on Build GRCh38
rs782312612 Chr.11: 119085070-119085070 on Build GRCh38
rs549270240 Chr.11: 119085172-119085173 on Build GRCh38
rs574906308 Chr.11: 119085173-119085175 on Build GRCh38
rs782499908 Chr.11: 119084983-119084983 on Build GRCh38
rs535621227 Chr.11: 119084827-119084827 on Build GRCh38
rs782013094 Chr.11: 119085172-119085172 on Build GRCh38
rs782360978 Chr.11: 119085173-119085184 on Build GRCh38
rs781985872 Chr.11: 119085173-119085187 on Build GRCh38
rs958142961 Chr.11: 119084909-119084909 on Build GRCh38
rs367703545 Chr.11: 119085005-119085005 on Build GRCh38
rs782123841 Chr.11: 119085028-119085028 on Build GRCh38
rs782654123 Chr.11: 119085107-119085108 on Build GRCh38
rs199862927 Chr.11: 119085086-119085086 on Build GRCh38
rs782036309 Chr.11: 119085173-119085188 on Build GRCh38
rs782589841 Chr.11: 119085166-119085166 on Build GRCh38
rs1012441521 Chr.11: 119084918-119084918 on Build GRCh38
rs782319054 Chr.11: 119084993-119084993 on Build GRCh38
rs988589029 Chr.11: 119084911-119084911 on Build GRCh38
rs782521497 Chr.11: 119085024-119085024 on Build GRCh38
rs782679152 Chr.11: 119084990-119084990 on Build GRCh38
rs782772607 Chr.11: 119085205-119085213 on Build GRCh38
rs939710565 Chr.11: 119084811-119084811 on Build GRCh38
rs914502417 Chr.11: 119084826-119084826 on Build GRCh38
rs896619146 Chr.11: 119084912-119084912 on Build GRCh38
rs781818939 Chr.11: 119085027-119085027 on Build GRCh38
rs782408982 Chr.11: 119085009-119085009 on Build GRCh38
rs782082681 Chr.11: 119085080-119085080 on Build GRCh38
rs782243560 Chr.11: 119085063-119085063 on Build GRCh38
rs781880577 Chr.11: 119085109-119085109 on Build GRCh38
rs373965531 Chr.11: 119085022-119085022 on Build GRCh38
rs532957570 Chr.11: 119085025-119085025 on Build GRCh38
rs782571483 Chr.11: 119085135-119085135 on Build GRCh38
rs914471864 Chr.11: 119085061-119085061 on Build GRCh38
rs782474151 Chr.11: 119085147-119085147 on Build GRCh38
rs782647894 Chr.11: 119085062-119085062 on Build GRCh38
rs1032870688 Chr.11: 119084904-119084904 on Build GRCh38
rs1020972364 Chr.11: 119084920-119084920 on Build GRCh38
rs142812375 Chr.11: 119085064-119085064 on Build GRCh38
rs938480653 Chr.11: 119084907-119084907 on Build GRCh38
rs891586774 Chr.11: 119085223-119085223 on Build GRCh38
rs200691284 Chr.11: 119085023-119085023 on Build GRCh38
rs781964094 Chr.11: 119085201-119085202 on Build GRCh38
rs796315620 Chr.11: 119085202-119085203 on Build GRCh38
rs781889757 Chr.11: 119085130-119085130 on Build GRCh38
rs782461751 Chr.11: 119084855-119084855 on Build GRCh38
rs782465908 Chr.11: 119085132-119085132 on Build GRCh38
rs782184714 Chr.11: 119085206-119085206 on Build GRCh38
rs866577852 Chr.11: 119084929-119084929 on Build GRCh38
rs782573719 Chr.11: 119084858-119084858 on Build GRCh38
rs782239877 Chr.11: 119084865-119084865 on Build GRCh38
rs368445287 Chr.11: 119085033-119085033 on Build GRCh38
rs782044774 Chr.11: 119085094-119085094 on Build GRCh38
rs781981242 Chr.11: 119085082-119085082 on Build GRCh38
rs34104555 Chr.11: 119085204-119085204 on Build GRCh38
rs782577135 Chr.11: 119085041-119085041 on Build GRCh38
rs372520245 Chr.11: 119085081-119085081 on Build GRCh38
rs782155487 Chr.11: 119085084-119085084 on Build GRCh38
rs782474855 Chr.11: 119085173-119085185 on Build GRCh38
rs199772277 Chr.11: 119085018-119085018 on Build GRCh38
rs567436321 Chr.11: 119085099-119085099 on Build GRCh38
rs72997366 Chr.11: 119084931-119084931 on Build GRCh38
rs782304835 Chr.11: 119085120-119085136 on Build GRCh38
rs1009664830 Chr.11: 119085227-119085227 on Build GRCh38
rs753765657 Chr.11: 119085100-119085100 on Build GRCh38
rs782284518 Chr.11: 119085108-119085109 on Build GRCh38
rs782581394 Chr.11: 119085162-119085162 on Build GRCh38
rs782692751 Chr.11: 119085110-119085110 on Build GRCh38
rs782487780 Chr.11: 119085060-119085060 on Build GRCh38
rs686624 Chr.11: 119084799-119084799 on Build GRCh38
rs782411017 Chr.11: 119085164-119085164 on Build GRCh38
rs782631409 Chr.11: 119085000-119085000 on Build GRCh38
rs1041407117 Chr.11: 119084841-119084841 on Build GRCh38
rs889628179 Chr.11: 119084877-119084877 on Build GRCh38
rs1048637240 Chr.11: 119084790-119084790 on Build GRCh38
rs1056880185 Chr.11: 119085197-119085197 on Build GRCh38
rs148084355 Chr.11: 119085059-119085059 on Build GRCh38
rs1014958802 Chr.11: 119084943-119084943 on Build GRCh38
rs982374074 Chr.11: 119084967-119084967 on Build GRCh38
rs782214327 Chr.11: 119085168-119085171 on Build GRCh38
rs568398887 Chr.11: 119084824-119084824 on Build GRCh38
rs571419402 Chr.11: 119085257-119085257 on Build GRCh38
rs1006630002 Chr.11: 119084940-119084940 on Build GRCh38
rs938351818 Chr.11: 119085098-119085098 on Build GRCh38
rs782240825 Chr.11: 119085161-119085161 on Build GRCh38
rs569415770 Chr.11: 119085075-119085075 on Build GRCh38
rs782055615 Chr.11: 119085202-119085203 on Build GRCh38
rs118204118 Chr.11: 119085034-119085034 on Build GRCh38
rs589925 Chr.11: 119084969-119084969 on Build GRCh38
rs782213223 Chr.11: 119085002-119085002 on Build GRCh38
rs371065891 Chr.11: 119085065-119085066 on Build GRCh38
rs782719815 Chr.11: 119085202-119085213 on Build GRCh38
rs550114087 Chr.11: 119084821-119084821 on Build GRCh38
rs553977615 Chr.11: 119084956-119084956 on Build GRCh38
rs781901249 Chr.11: 119085036-119085036 on Build GRCh38
rs188582005 Chr.11: 119085243-119085243 on Build GRCh38
rs782201645 Chr.11: 119085068-119085068 on Build GRCh38
rs371380824 Chr.11: 119085091-119085091 on Build GRCh38
rs201349602 Chr.11: 119085006-119085006 on Build GRCh38
rs782130094 Chr.11: 119085173-119085186 on Build GRCh38
rs534638201 Chr.11: 119085217-119085217 on Build GRCh38

View Full Product Details