Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119639773-119640282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NECTIN1
Entrez Gene ID
5818
Gene Name
nectin cell adhesion molecule 1
Gene Aliases
CD111, CLPED1, ED4, HIgR, HV1S, HVEC, OFC7, PRR, PRR1, PVRL1, PVRR, PVRR1, SK-12, nectin-1
UniGene
Hs.334846
Gene Chromosome Location
Chr.11: 119638098-119728725 on Build GRCh38
Transcripts
NM_203285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6315402 c.1151+6C>A p.? Chr.11: 119639859-119639859 on Build GRCh38
4521889 c.1135G>A p.E379K Chr.11: 119639881-119639881 on Build GRCh38
5371816 c.1092C>T p.L364L Chr.11: 119639924-119639924 on Build GRCh38
3786994 c.1127G>T p.S376I Chr.11: 119639889-119639889 on Build GRCh38
7348625 c.1052C>T p.A351V Chr.11: 119639964-119639964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771549215 Chr.11: 119639873-119639873 on Build GRCh38
rs889132874 Chr.11: 119639804-119639804 on Build GRCh38
rs554942877 Chr.11: 119640065-119640065 on Build GRCh38
rs1018629929 Chr.11: 119640194-119640194 on Build GRCh38
rs1006163119 Chr.11: 119640139-119640139 on Build GRCh38
rs200519777 Chr.11: 119639882-119639882 on Build GRCh38
rs545782858 Chr.11: 119639877-119639877 on Build GRCh38
rs73571279 Chr.11: 119640200-119640200 on Build GRCh38
rs185577694 Chr.11: 119640084-119640084 on Build GRCh38
rs897209565 Chr.11: 119640176-119640176 on Build GRCh38
rs769877855 Chr.11: 119639906-119639906 on Build GRCh38
rs999167062 Chr.11: 119640086-119640086 on Build GRCh38
rs745988281 Chr.11: 119639818-119639818 on Build GRCh38
rs761064885 Chr.11: 119640222-119640222 on Build GRCh38
rs137960191 Chr.11: 119639876-119639876 on Build GRCh38
rs376901395 Chr.11: 119639825-119639825 on Build GRCh38
rs199613491 Chr.11: 119639982-119639982 on Build GRCh38
rs745463793 Chr.11: 119640016-119640016 on Build GRCh38
rs7940667 Chr.11: 119639934-119639934 on Build GRCh38
rs983790922 Chr.11: 119640135-119640135 on Build GRCh38
rs746238394 Chr.11: 119640041-119640041 on Build GRCh38
rs945724469 Chr.11: 119639810-119639810 on Build GRCh38
rs1003768754 Chr.11: 119639894-119639894 on Build GRCh38
rs750594983 Chr.11: 119639964-119639964 on Build GRCh38
rs374812698 Chr.11: 119639900-119639900 on Build GRCh38
rs573059971 Chr.11: 119640131-119640131 on Build GRCh38
rs1006807226 Chr.11: 119640191-119640191 on Build GRCh38
rs770519718 Chr.11: 119640175-119640175 on Build GRCh38
rs761699695 Chr.11: 119639851-119639851 on Build GRCh38
rs990358571 Chr.11: 119640202-119640202 on Build GRCh38
rs748044781 Chr.11: 119639902-119639902 on Build GRCh38
rs373455164 Chr.11: 119639814-119639814 on Build GRCh38
rs747582783 Chr.11: 119640050-119640050 on Build GRCh38
rs557773990 Chr.11: 119639879-119639879 on Build GRCh38
rs535276280 Chr.11: 119640208-119640208 on Build GRCh38
rs776755803 Chr.11: 119639835-119639835 on Build GRCh38
rs969771421 Chr.11: 119640133-119640133 on Build GRCh38
rs780236235 Chr.11: 119639973-119639973 on Build GRCh38
rs774091006 Chr.11: 119640180-119640180 on Build GRCh38
rs939571168 Chr.11: 119640100-119640100 on Build GRCh38
rs1022678320 Chr.11: 119640123-119640123 on Build GRCh38
rs149487173 Chr.11: 119639883-119639883 on Build GRCh38
rs762467761 Chr.11: 119639865-119639865 on Build GRCh38
rs766088548 Chr.11: 119639866-119639866 on Build GRCh38
rs1045347801 Chr.11: 119640074-119640074 on Build GRCh38
rs777148734 Chr.11: 119640121-119640121 on Build GRCh38
rs957469463 Chr.11: 119640025-119640025 on Build GRCh38
rs889163843 Chr.11: 119640036-119640036 on Build GRCh38
rs933486722 Chr.11: 119640183-119640183 on Build GRCh38
rs1031560055 Chr.11: 119640104-119640104 on Build GRCh38
rs4938699 Chr.11: 119640227-119640227 on Build GRCh38
rs549778498 Chr.11: 119640177-119640177 on Build GRCh38
rs1051865284 Chr.11: 119639999-119639999 on Build GRCh38
rs963689709 Chr.11: 119640169-119640169 on Build GRCh38
rs960164679 Chr.11: 119640105-119640105 on Build GRCh38
rs141063530 Chr.11: 119639963-119639963 on Build GRCh38
rs755894063 Chr.11: 119639832-119639832 on Build GRCh38
rs891457125 Chr.11: 119639809-119639809 on Build GRCh38
rs769824361 Chr.11: 119639990-119639990 on Build GRCh38
rs762616169 Chr.11: 119640144-119640144 on Build GRCh38
rs771196798 Chr.11: 119639816-119639816 on Build GRCh38
rs771733323 Chr.11: 119640027-119640027 on Build GRCh38
rs779080266 Chr.11: 119639977-119639978 on Build GRCh38
rs773103055 Chr.11: 119639909-119639909 on Build GRCh38
rs368086348 Chr.11: 119639957-119639957 on Build GRCh38
rs1029638165 Chr.11: 119640178-119640178 on Build GRCh38
rs774939942 Chr.11: 119640037-119640037 on Build GRCh38
rs977323860 Chr.11: 119639843-119639843 on Build GRCh38
rs1054646067 Chr.11: 119639803-119639803 on Build GRCh38
rs761121601 Chr.11: 119639956-119639956 on Build GRCh38
rs201902191 Chr.11: 119639862-119639862 on Build GRCh38
rs749025183 Chr.11: 119640003-119640003 on Build GRCh38
rs766944808 Chr.11: 119639950-119639950 on Build GRCh38
rs772241031 Chr.11: 119640006-119640006 on Build GRCh38
rs909534101 Chr.11: 119640166-119640166 on Build GRCh38
rs536151466 Chr.11: 119639980-119639980 on Build GRCh38
rs774111612 Chr.11: 119639937-119639937 on Build GRCh38
rs773158554 Chr.11: 119639864-119639864 on Build GRCh38
rs777615421 Chr.11: 119639867-119639867 on Build GRCh38
rs555397459 Chr.11: 119640004-119640004 on Build GRCh38
rs780287456 Chr.11: 119639901-119639901 on Build GRCh38
rs140342742 Chr.11: 119639998-119639998 on Build GRCh38
rs943417776 Chr.11: 119640212-119640212 on Build GRCh38
rs562428156 Chr.11: 119639899-119639899 on Build GRCh38
rs758962540 Chr.11: 119639948-119639948 on Build GRCh38
rs1007602070 Chr.11: 119640062-119640062 on Build GRCh38
rs760214425 Chr.11: 119640041-119640041 on Build GRCh38
rs762847279 Chr.11: 119639915-119639915 on Build GRCh38
rs752216575 Chr.11: 119639954-119639954 on Build GRCh38
rs200132006 Chr.11: 119639898-119639898 on Build GRCh38
rs989896206 Chr.11: 119640029-119640029 on Build GRCh38
rs112109871 Chr.11: 119640011-119640011 on Build GRCh38
rs758710390 Chr.11: 119639966-119639966 on Build GRCh38
rs1038548865 Chr.11: 119640158-119640158 on Build GRCh38
rs780690195 Chr.11: 119640046-119640046 on Build GRCh38
rs531353778 Chr.11: 119640108-119640108 on Build GRCh38
rs935731557 Chr.11: 119639802-119639802 on Build GRCh38

View Full Product Details