Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 39640625-39641143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD2
Entrez Gene ID
225
Gene Name
ATP binding cassette subfamily D member 2
Gene Aliases
ABC39, ALDL1, ALDR, ALDRP, hALDR
UniGene
Hs.117852
Gene Chromosome Location
Chr.12: 39530966-39651333 on Build GRCh38
Transcripts
XM_011538027.2 NM_001031748.3 NM_001319247.1 NR_135051.1 XM_005268806.3

Gene Symbol
C12orf40
Entrez Gene ID
283461
Gene Name
chromosome 12 open reading frame 40
Gene Aliases
HEL-206, HEL-S-94
UniGene
Hs.648205
Gene Chromosome Location
Chr.12: 39626167-39908300 on Build GRCh38
Transcripts
XM_011538027.2 NM_001031748.3 NM_001319247.1 NR_135051.1 XM_005268806.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5515472 c.63+7A>T p.? Chr.12: 39641001-39641001 on Build GRCh38
431057 c.36G>C p.L12L Chr.12: 39640967-39640967 on Build GRCh38
5445229 c.63+5A>T p.? Chr.12: 39640999-39640999 on Build GRCh38
938970 c.63+1G>A p.? Chr.12: 39640995-39640995 on Build GRCh38
3739630 c.27C>A p.S9S Chr.12: 39640958-39640958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888609416 Chr.12: 39640666-39640666 on Build GRCh38
rs1015852351 Chr.12: 39640725-39640725 on Build GRCh38
rs373907993 Chr.12: 39640959-39640959 on Build GRCh38
rs965603574 Chr.12: 39640864-39640864 on Build GRCh38
rs752634721 Chr.12: 39640963-39640963 on Build GRCh38
rs769109704 Chr.12: 39641026-39641026 on Build GRCh38
rs888148581 Chr.12: 39641054-39641054 on Build GRCh38
rs544997727 Chr.12: 39640673-39640673 on Build GRCh38
rs769250495 Chr.12: 39641014-39641014 on Build GRCh38
rs888472646 Chr.12: 39640794-39640794 on Build GRCh38
rs901318926 Chr.12: 39641098-39641098 on Build GRCh38
rs560090783 Chr.12: 39640760-39640760 on Build GRCh38
rs1004457313 Chr.12: 39641074-39641074 on Build GRCh38
rs550100187 Chr.12: 39640916-39640916 on Build GRCh38
rs998330242 Chr.12: 39640885-39640885 on Build GRCh38
rs376486007 Chr.12: 39641048-39641048 on Build GRCh38
rs777719718 Chr.12: 39641043-39641051 on Build GRCh38
rs1007083973 Chr.12: 39640830-39640830 on Build GRCh38
rs968405291 Chr.12: 39640997-39640997 on Build GRCh38
rs751175225 Chr.12: 39640970-39640970 on Build GRCh38
rs749859741 Chr.12: 39641023-39641023 on Build GRCh38
rs374037420 Chr.12: 39640859-39640859 on Build GRCh38
rs1057369024 Chr.12: 39640678-39640678 on Build GRCh38
rs897205775 Chr.12: 39640680-39640684 on Build GRCh38
rs927158412 Chr.12: 39641011-39641011 on Build GRCh38
rs998774418 Chr.12: 39641121-39641121 on Build GRCh38
rs547672824 Chr.12: 39640846-39640846 on Build GRCh38
rs568771293 Chr.12: 39640921-39640921 on Build GRCh38
rs527799633 Chr.12: 39640788-39640788 on Build GRCh38
rs771803154 Chr.12: 39641067-39641067 on Build GRCh38
rs370616425 Chr.12: 39641049-39641049 on Build GRCh38
rs566440596 Chr.12: 39640975-39640975 on Build GRCh38
rs780596997 Chr.12: 39641020-39641020 on Build GRCh38
rs994674921 Chr.12: 39640688-39640688 on Build GRCh38
rs190930538 Chr.12: 39640831-39640831 on Build GRCh38
rs1037714187 Chr.12: 39641095-39641095 on Build GRCh38
rs976796656 Chr.12: 39640801-39640801 on Build GRCh38
rs201513060 Chr.12: 39641034-39641034 on Build GRCh38
rs374734474 Chr.12: 39641008-39641008 on Build GRCh38
rs758170992 Chr.12: 39640965-39640965 on Build GRCh38
rs926385812 Chr.12: 39641025-39641025 on Build GRCh38
rs113706234 Chr.12: 39640662-39640662 on Build GRCh38
rs373154824 Chr.12: 39641089-39641089 on Build GRCh38
rs1018516266 Chr.12: 39640857-39640857 on Build GRCh38
rs201615505 Chr.12: 39640928-39640928 on Build GRCh38
rs376045147 Chr.12: 39640911-39640911 on Build GRCh38
rs368791441 Chr.12: 39641036-39641036 on Build GRCh38
rs755996662 Chr.12: 39641009-39641009 on Build GRCh38
rs533883894 Chr.12: 39641007-39641007 on Build GRCh38
rs577974644 Chr.12: 39640670-39640670 on Build GRCh38
rs780508496 Chr.12: 39641006-39641006 on Build GRCh38
rs927101696 Chr.12: 39640669-39640670 on Build GRCh38
rs749805581 Chr.12: 39640994-39640994 on Build GRCh38
rs58302581 Chr.12: 39640968-39640968 on Build GRCh38
rs558575211 Chr.12: 39641113-39641113 on Build GRCh38
rs371797285 Chr.12: 39640995-39640995 on Build GRCh38
rs978833437 Chr.12: 39640905-39640905 on Build GRCh38
rs759326618 Chr.12: 39640945-39640945 on Build GRCh38
rs11836770 Chr.12: 39640897-39640897 on Build GRCh38

View Full Product Details