Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120495889-120496426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLL1
Entrez Gene ID
8655
Gene Name
dynein light chain LC8-type 1
Gene Aliases
DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, PIN, hdlc1
UniGene
Hs.5120
Gene Chromosome Location
Chr.12: 120469857-120498495 on Build GRCh38
Transcripts
NM_001037494.1 NM_001037495.1 NM_003746.2 NR_038854.1

Gene Symbol
NRAV
Entrez Gene ID
100506668
Gene Name
negative regulator of antiviral response (non-protein coding)
Gene Aliases
DYNLL1-AS1, DYNLL1AS1
UniGene
Hs.660080
Gene Chromosome Location
Chr.12: 120490338-120495946 on Build GRCh38
Transcripts
NM_001037494.1 NM_001037495.1 NM_003746.2 NR_038854.1

SNP Detail

SNP ID Chromosome Location
rs554496255 Chr.12: 120496151-120496151 on Build GRCh38
rs909928679 Chr.12: 120496175-120496175 on Build GRCh38
rs1029813606 Chr.12: 120496130-120496130 on Build GRCh38
rs957914588 Chr.12: 120496333-120496333 on Build GRCh38
rs368159020 Chr.12: 120496196-120496196 on Build GRCh38
rs572693672 Chr.12: 120496347-120496347 on Build GRCh38
rs573257955 Chr.12: 120496368-120496368 on Build GRCh38
rs565441080 Chr.12: 120495982-120495982 on Build GRCh38
rs1002433250 Chr.12: 120496325-120496325 on Build GRCh38
rs541588936 Chr.12: 120496365-120496365 on Build GRCh38
rs1025141046 Chr.12: 120496282-120496282 on Build GRCh38
rs981099574 Chr.12: 120496104-120496104 on Build GRCh38
rs946124658 Chr.12: 120496115-120496115 on Build GRCh38
rs879746850 Chr.12: 120496236-120496237 on Build GRCh38
rs917117920 Chr.12: 120496349-120496349 on Build GRCh38
rs957068696 Chr.12: 120496136-120496136 on Build GRCh38
rs988340931 Chr.12: 120496137-120496137 on Build GRCh38
rs888549866 Chr.12: 120496101-120496101 on Build GRCh38
rs755419676 Chr.12: 120496398-120496398 on Build GRCh38
rs561428419 Chr.12: 120496055-120496055 on Build GRCh38
rs773657591 Chr.12: 120495923-120495923 on Build GRCh38
rs774600543 Chr.12: 120496032-120496032 on Build GRCh38
rs1039670398 Chr.12: 120496187-120496187 on Build GRCh38
rs570039998 Chr.12: 120496119-120496119 on Build GRCh38
rs374019342 Chr.12: 120496392-120496392 on Build GRCh38
rs914593387 Chr.12: 120496178-120496178 on Build GRCh38
rs183162883 Chr.12: 120496399-120496399 on Build GRCh38
rs561294020 Chr.12: 120496384-120496384 on Build GRCh38
rs375465709 Chr.12: 120496391-120496391 on Build GRCh38
rs970976242 Chr.12: 120496153-120496153 on Build GRCh38
rs1020421379 Chr.12: 120496138-120496138 on Build GRCh38
rs749696461 Chr.12: 120495936-120495936 on Build GRCh38
rs781259831 Chr.12: 120496380-120496380 on Build GRCh38
rs893263436 Chr.12: 120495917-120495917 on Build GRCh38
rs530243574 Chr.12: 120496107-120496107 on Build GRCh38
rs34457759 Chr.12: 120496376-120496377 on Build GRCh38
rs1048026650 Chr.12: 120496261-120496261 on Build GRCh38
rs576380944 Chr.12: 120496251-120496251 on Build GRCh38
rs2233677 Chr.12: 120496328-120496328 on Build GRCh38
rs580016 Chr.12: 120496174-120496174 on Build GRCh38
rs768713675 Chr.12: 120496375-120496375 on Build GRCh38
rs976850146 Chr.12: 120495958-120495958 on Build GRCh38
rs999976970 Chr.12: 120496304-120496304 on Build GRCh38
rs748980731 Chr.12: 120496388-120496388 on Build GRCh38
rs1010390828 Chr.12: 120495943-120495943 on Build GRCh38
rs550304010 Chr.12: 120496160-120496160 on Build GRCh38
rs899867631 Chr.12: 120496197-120496197 on Build GRCh38
rs982566275 Chr.12: 120496084-120496085 on Build GRCh38
rs1710132 Chr.12: 120495989-120495989 on Build GRCh38
rs192438612 Chr.12: 120496249-120496249 on Build GRCh38
rs992518928 Chr.12: 120496338-120496338 on Build GRCh38
rs926594124 Chr.12: 120496105-120496105 on Build GRCh38
rs1042199703 Chr.12: 120496203-120496203 on Build GRCh38
rs941487497 Chr.12: 120496180-120496180 on Build GRCh38
rs949309481 Chr.12: 120496100-120496100 on Build GRCh38
rs1056981205 Chr.12: 120496051-120496051 on Build GRCh38
rs998426477 Chr.12: 120496120-120496120 on Build GRCh38
rs929768692 Chr.12: 120496166-120496166 on Build GRCh38
rs764321380 Chr.12: 120496271-120496271 on Build GRCh38
rs1018232764 Chr.12: 120496117-120496117 on Build GRCh38
rs11544058 Chr.12: 120496214-120496214 on Build GRCh38
rs370300291 Chr.12: 120495961-120495961 on Build GRCh38
rs1008913324 Chr.12: 120496010-120496010 on Build GRCh38
rs12857 Chr.12: 120496143-120496143 on Build GRCh38
rs913000526 Chr.12: 120496020-120496020 on Build GRCh38
rs1033995733 Chr.12: 120496326-120496326 on Build GRCh38
rs895665543 Chr.12: 120496065-120496065 on Build GRCh38
rs185914041 Chr.12: 120495909-120495909 on Build GRCh38
rs748007480 Chr.12: 120496382-120496382 on Build GRCh38
rs1051756759 Chr.12: 120496233-120496233 on Build GRCh38
rs552461487 Chr.12: 120496146-120496146 on Build GRCh38
rs998159574 Chr.12: 120496227-120496227 on Build GRCh38
rs947008322 Chr.12: 120496031-120496031 on Build GRCh38
rs912683612 Chr.12: 120496207-120496207 on Build GRCh38
rs1047565273 Chr.12: 120496094-120496094 on Build GRCh38
rs771524418 Chr.12: 120495965-120495965 on Build GRCh38
rs559192703 Chr.12: 120496277-120496277 on Build GRCh38
rs113359923 Chr.12: 120495954-120495954 on Build GRCh38
rs545403435 Chr.12: 120496270-120496270 on Build GRCh38
rs117015363 Chr.12: 120496149-120496149 on Build GRCh38
rs368008301 Chr.12: 120496373-120496373 on Build GRCh38
rs550315954 Chr.12: 120496111-120496111 on Build GRCh38
rs980667715 Chr.12: 120496156-120496156 on Build GRCh38
rs1167704 Chr.12: 120496043-120496043 on Build GRCh38
rs753590933 Chr.12: 120496280-120496280 on Build GRCh38
rs754295556 Chr.12: 120496286-120496286 on Build GRCh38
rs374987411 Chr.12: 120496275-120496275 on Build GRCh38
rs995728851 Chr.12: 120496254-120496254 on Build GRCh38
rs545267200 Chr.12: 120495916-120495916 on Build GRCh38
rs937320993 Chr.12: 120496106-120496106 on Build GRCh38
rs951160229 Chr.12: 120496078-120496078 on Build GRCh38
rs774146121 Chr.12: 120496171-120496172 on Build GRCh38
rs1026691412 Chr.12: 120496070-120496070 on Build GRCh38
rs988990971 Chr.12: 120495992-120495992 on Build GRCh38
rs993174939 Chr.12: 120496154-120496154 on Build GRCh38
rs991683291 Chr.12: 120496062-120496062 on Build GRCh38
rs772836331 Chr.12: 120496386-120496386 on Build GRCh38
rs937283225 Chr.12: 120496050-120496050 on Build GRCh38
rs995117957 Chr.12: 120496140-120496140 on Build GRCh38
rs930056437 Chr.12: 120496090-120496090 on Build GRCh38
rs914540855 Chr.12: 120496385-120496385 on Build GRCh38

View Full Product Details