Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121160851-121161355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RX7
Entrez Gene ID
5027
Gene Name
purinergic receptor P2X 7
Gene Aliases
P2X7
UniGene
Hs.729169
Gene Chromosome Location
Chr.12: 121132819-121189478 on Build GRCh38
Transcripts
NM_002562.5 NR_033948.1 NR_033949.1 NR_033950.1 NR_033951.1 NR_033952.1 NR_033953.1 NR_033954.1 NR_033955.1 NR_033956.1 XM_011538419.2 XM_011538420.2 XM_017019364.1 XM_017019365.1 XM_017019366.1 XM_017019367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1359427 c.380C>T p.T127M Chr.12: 121160918-121160918 on Build GRCh38
935995 c.381G>A p.T127T Chr.12: 121160919-121160919 on Build GRCh38
5840329 c.406delA p.K137fs*17 Chr.12: 121160944-121160944 on Build GRCh38
4710660 c.398G>A p.R133Q Chr.12: 121160936-121160936 on Build GRCh38
7303206 c.422A>G p.D141G Chr.12: 121160960-121160960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374149653 Chr.12: 121161001-121161001 on Build GRCh38
rs914023555 Chr.12: 121161140-121161140 on Build GRCh38
rs1002490878 Chr.12: 121161108-121161108 on Build GRCh38
rs893413238 Chr.12: 121161187-121161187 on Build GRCh38
rs1054524391 Chr.12: 121161177-121161177 on Build GRCh38
rs897218261 Chr.12: 121161322-121161322 on Build GRCh38
rs201680771 Chr.12: 121160885-121160885 on Build GRCh38
rs1027589442 Chr.12: 121161063-121161063 on Build GRCh38
rs555587636 Chr.12: 121160891-121160891 on Build GRCh38
rs779103436 Chr.12: 121160886-121160886 on Build GRCh38
rs112278868 Chr.12: 121160988-121160988 on Build GRCh38
rs1044826582 Chr.12: 121161293-121161293 on Build GRCh38
rs886230823 Chr.12: 121161253-121161253 on Build GRCh38
rs1047562910 Chr.12: 121161239-121161239 on Build GRCh38
rs745474845 Chr.12: 121160883-121160883 on Build GRCh38
rs920543144 Chr.12: 121161208-121161208 on Build GRCh38
rs36075646 Chr.12: 121161320-121161320 on Build GRCh38
rs200743882 Chr.12: 121160979-121160979 on Build GRCh38
rs558369761 Chr.12: 121160950-121160950 on Build GRCh38
rs562546486 Chr.12: 121161302-121161302 on Build GRCh38
rs200430009 Chr.12: 121160935-121160935 on Build GRCh38
rs765837924 Chr.12: 121161160-121161160 on Build GRCh38
rs944486797 Chr.12: 121161142-121161142 on Build GRCh38
rs200822907 Chr.12: 121160911-121160911 on Build GRCh38
rs200332151 Chr.12: 121160991-121160991 on Build GRCh38
rs572114518 Chr.12: 121161161-121161161 on Build GRCh38
rs746057828 Chr.12: 121160977-121160977 on Build GRCh38
rs1054637409 Chr.12: 121160919-121160919 on Build GRCh38
rs201296551 Chr.12: 121160995-121160995 on Build GRCh38
rs764669349 Chr.12: 121161008-121161008 on Build GRCh38
rs778967979 Chr.12: 121160975-121160975 on Build GRCh38
rs558413927 Chr.12: 121161133-121161133 on Build GRCh38
rs200025457 Chr.12: 121160918-121160918 on Build GRCh38
rs940124196 Chr.12: 121161277-121161277 on Build GRCh38
rs143722015 Chr.12: 121160963-121160963 on Build GRCh38
rs993347154 Chr.12: 121161328-121161328 on Build GRCh38
rs534018162 Chr.12: 121161122-121161122 on Build GRCh38
rs762447689 Chr.12: 121161123-121161123 on Build GRCh38
rs201136772 Chr.12: 121160921-121160921 on Build GRCh38
rs948960393 Chr.12: 121161224-121161224 on Build GRCh38
rs368041937 Chr.12: 121160915-121160915 on Build GRCh38
rs140927967 Chr.12: 121160909-121160909 on Build GRCh38
rs202212713 Chr.12: 121160936-121160936 on Build GRCh38
rs951917195 Chr.12: 121161093-121161093 on Build GRCh38
rs34702297 Chr.12: 121161319-121161320 on Build GRCh38
rs780044922 Chr.12: 121160978-121160978 on Build GRCh38
rs182118655 Chr.12: 121161298-121161298 on Build GRCh38
rs749523524 Chr.12: 121160884-121160884 on Build GRCh38
rs199972027 Chr.12: 121160898-121160898 on Build GRCh38
rs201668926 Chr.12: 121160912-121160912 on Build GRCh38
rs375852722 Chr.12: 121160993-121160993 on Build GRCh38
rs745908932 Chr.12: 121160890-121160890 on Build GRCh38
rs762607910 Chr.12: 121160996-121160996 on Build GRCh38
rs200426192 Chr.12: 121161007-121161007 on Build GRCh38
rs746135200 Chr.12: 121160879-121160880 on Build GRCh38
rs762400464 Chr.12: 121160938-121160938 on Build GRCh38
rs754238799 Chr.12: 121160964-121160964 on Build GRCh38
rs1015688598 Chr.12: 121161303-121161303 on Build GRCh38
rs201964517 Chr.12: 121161000-121161000 on Build GRCh38
rs966490107 Chr.12: 121161154-121161154 on Build GRCh38
rs1038920265 Chr.12: 121160968-121160968 on Build GRCh38
rs970711839 Chr.12: 121161089-121161089 on Build GRCh38
rs769160925 Chr.12: 121160969-121160969 on Build GRCh38
rs757589091 Chr.12: 121161013-121161013 on Build GRCh38
rs750618459 Chr.12: 121161016-121161016 on Build GRCh38
rs375894120 Chr.12: 121160957-121160957 on Build GRCh38
rs568948806 Chr.12: 121160956-121160956 on Build GRCh38
rs765863461 Chr.12: 121160949-121160949 on Build GRCh38
rs758645477 Chr.12: 121161020-121161020 on Build GRCh38
rs761319746 Chr.12: 121161005-121161005 on Build GRCh38
rs934837137 Chr.12: 121161163-121161163 on Build GRCh38
rs765584241 Chr.12: 121161015-121161015 on Build GRCh38
rs754768143 Chr.12: 121160889-121160891 on Build GRCh38
rs924324739 Chr.12: 121161162-121161162 on Build GRCh38
rs1024981813 Chr.12: 121161075-121161075 on Build GRCh38

View Full Product Details