Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122481430-122481937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC8
Entrez Gene ID
55596
Gene Name
zinc finger CCHC-type containing 8
Gene Aliases
-
UniGene
Hs.37706
Gene Chromosome Location
Chr.12: 122471599-122501073 on Build GRCh38
Transcripts
NM_017612.4 XM_006719491.2 XM_011538555.2 XM_017019606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5935992 c.1013G>A p.G338E Chr.12: 122481527-122481527 on Build GRCh38
264778 c.1005C>A p.L335L Chr.12: 122481535-122481535 on Build GRCh38
4645836 c.957A>G p.P319P Chr.12: 122481583-122481583 on Build GRCh38
692577 c.945G>C p.Q315H Chr.12: 122481595-122481595 on Build GRCh38
379022 c.935G>A p.R312Q Chr.12: 122481605-122481605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534897461 Chr.12: 122481705-122481705 on Build GRCh38
rs763983823 Chr.12: 122481911-122481911 on Build GRCh38
rs759186171 Chr.12: 122481694-122481694 on Build GRCh38
rs754640355 Chr.12: 122481599-122481599 on Build GRCh38
rs528243881 Chr.12: 122481698-122481698 on Build GRCh38
rs941953795 Chr.12: 122481799-122481799 on Build GRCh38
rs751699624 Chr.12: 122481487-122481496 on Build GRCh38
rs565612846 Chr.12: 122481537-122481537 on Build GRCh38
rs919056320 Chr.12: 122481818-122481818 on Build GRCh38
rs57144421 Chr.12: 122481469-122481470 on Build GRCh38
rs869291377 Chr.12: 122481481-122481482 on Build GRCh38
rs747085021 Chr.12: 122481607-122481607 on Build GRCh38
rs776006897 Chr.12: 122481565-122481565 on Build GRCh38
rs770142506 Chr.12: 122481652-122481652 on Build GRCh38
rs4758692 Chr.12: 122481826-122481826 on Build GRCh38
rs777624068 Chr.12: 122481606-122481606 on Build GRCh38
rs1006576157 Chr.12: 122481884-122481884 on Build GRCh38
rs753447389 Chr.12: 122481589-122481589 on Build GRCh38
rs758171659 Chr.12: 122481895-122481895 on Build GRCh38
rs759871715 Chr.12: 122481558-122481558 on Build GRCh38
rs777905526 Chr.12: 122481488-122481488 on Build GRCh38
rs765756885 Chr.12: 122481669-122481669 on Build GRCh38
rs918499269 Chr.12: 122481816-122481816 on Build GRCh38
rs373244714 Chr.12: 122481630-122481630 on Build GRCh38
rs397713843 Chr.12: 122481480-122481481 on Build GRCh38
rs1057245773 Chr.12: 122481465-122481465 on Build GRCh38
rs772906844 Chr.12: 122481604-122481604 on Build GRCh38
rs775988353 Chr.12: 122481654-122481654 on Build GRCh38
rs1048037564 Chr.12: 122481869-122481869 on Build GRCh38
rs1012935616 Chr.12: 122481474-122481475 on Build GRCh38
rs749724899 Chr.12: 122481660-122481660 on Build GRCh38
rs1041155507 Chr.12: 122481890-122481890 on Build GRCh38
rs938739765 Chr.12: 122481462-122481462 on Build GRCh38
rs771165963 Chr.12: 122481538-122481538 on Build GRCh38
rs11058341 Chr.12: 122481729-122481729 on Build GRCh38
rs747116385 Chr.12: 122481500-122481500 on Build GRCh38
rs758343951 Chr.12: 122481605-122481605 on Build GRCh38
rs985802982 Chr.12: 122481772-122481772 on Build GRCh38
rs748023211 Chr.12: 122481473-122481473 on Build GRCh38
rs776761359 Chr.12: 122481547-122481547 on Build GRCh38
rs755170585 Chr.12: 122481488-122481489 on Build GRCh38
rs369253915 Chr.12: 122481614-122481614 on Build GRCh38
rs760150601 Chr.12: 122481586-122481586 on Build GRCh38
rs771909561 Chr.12: 122481487-122481487 on Build GRCh38
rs762411563 Chr.12: 122481668-122481668 on Build GRCh38
rs970057297 Chr.12: 122481732-122481732 on Build GRCh38
rs202179974 Chr.12: 122481469-122481469 on Build GRCh38
rs769184093 Chr.12: 122481661-122481661 on Build GRCh38
rs928738728 Chr.12: 122481749-122481749 on Build GRCh38
rs764571945 Chr.12: 122481583-122481583 on Build GRCh38
rs1036900774 Chr.12: 122481771-122481771 on Build GRCh38
rs764928051 Chr.12: 122481701-122481701 on Build GRCh38
rs965526469 Chr.12: 122481725-122481725 on Build GRCh38
rs377015965 Chr.12: 122481493-122481493 on Build GRCh38
rs77652707 Chr.12: 122481746-122481746 on Build GRCh38
rs776123904 Chr.12: 122481670-122481670 on Build GRCh38
rs756730899 Chr.12: 122481872-122481872 on Build GRCh38
rs972008012 Chr.12: 122481809-122481809 on Build GRCh38
rs781446496 Chr.12: 122481629-122481629 on Build GRCh38
rs766023074 Chr.12: 122481587-122481587 on Build GRCh38
rs878870842 Chr.12: 122481470-122481470 on Build GRCh38
rs914603337 Chr.12: 122481722-122481722 on Build GRCh38
rs566817171 Chr.12: 122481470-122481470 on Build GRCh38
rs763483581 Chr.12: 122481578-122481578 on Build GRCh38
rs201072310 Chr.12: 122481467-122481467 on Build GRCh38
rs186788133 Chr.12: 122481765-122481765 on Build GRCh38
rs565145884 Chr.12: 122481853-122481853 on Build GRCh38
rs900930247 Chr.12: 122481909-122481909 on Build GRCh38

View Full Product Details