Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112647476-112647971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11AUN
Entrez Gene ID
400165
Gene Name
ATP11A upstream neighbor
Gene Aliases
C13orf35, SMABLO1
UniGene
Hs.591213
Gene Chromosome Location
Chr.13: 112647044-112684497 on Build GRCh38
Transcripts
NM_207440.2

SNP Detail

SNP ID Chromosome Location
rs183426838 Chr.13: 112647935-112647935 on Build GRCh38
rs747969897 Chr.13: 112647801-112647801 on Build GRCh38
rs550403775 Chr.13: 112647929-112647929 on Build GRCh38
rs641148 Chr.13: 112647596-112647596 on Build GRCh38
rs970365401 Chr.13: 112647739-112647739 on Build GRCh38
rs928284807 Chr.13: 112647791-112647791 on Build GRCh38
rs902639779 Chr.13: 112647643-112647643 on Build GRCh38
rs1039540131 Chr.13: 112647687-112647687 on Build GRCh38
rs146849601 Chr.13: 112647576-112647576 on Build GRCh38
rs9324317 Chr.13: 112647619-112647619 on Build GRCh38
rs997957014 Chr.13: 112647775-112647775 on Build GRCh38
rs889804586 Chr.13: 112647611-112647611 on Build GRCh38
rs986015692 Chr.13: 112647883-112647883 on Build GRCh38
rs35800964 Chr.13: 112647504-112647505 on Build GRCh38
rs552891051 Chr.13: 112647670-112647670 on Build GRCh38
rs191552609 Chr.13: 112647578-112647578 on Build GRCh38
rs531915346 Chr.13: 112647703-112647703 on Build GRCh38
rs945948811 Chr.13: 112647621-112647621 on Build GRCh38
rs779647658 Chr.13: 112647505-112647505 on Build GRCh38
rs931560788 Chr.13: 112647591-112647591 on Build GRCh38
rs143762483 Chr.13: 112647575-112647575 on Build GRCh38
rs1015220000 Chr.13: 112647849-112647849 on Build GRCh38
rs576594338 Chr.13: 112647551-112647551 on Build GRCh38
rs981495298 Chr.13: 112647543-112647543 on Build GRCh38
rs963632563 Chr.13: 112647882-112647882 on Build GRCh38
rs972441313 Chr.13: 112647817-112647817 on Build GRCh38
rs935426817 Chr.13: 112647940-112647940 on Build GRCh38
rs981165227 Chr.13: 112647753-112647753 on Build GRCh38
rs140007433 Chr.13: 112647518-112647518 on Build GRCh38
rs754914272 Chr.13: 112647617-112647617 on Build GRCh38
rs547358832 Chr.13: 112647910-112647910 on Build GRCh38
rs931447444 Chr.13: 112647854-112647854 on Build GRCh38
rs1031469120 Chr.13: 112647669-112647669 on Build GRCh38
rs1011925975 Chr.13: 112647675-112647675 on Build GRCh38
rs370235335 Chr.13: 112647915-112647915 on Build GRCh38
rs185323007 Chr.13: 112647517-112647517 on Build GRCh38
rs527319137 Chr.13: 112647667-112647667 on Build GRCh38
rs557890083 Chr.13: 112647893-112647893 on Build GRCh38
rs957323545 Chr.13: 112647500-112647500 on Build GRCh38
rs564789113 Chr.13: 112647683-112647683 on Build GRCh38
rs1041432761 Chr.13: 112647635-112647635 on Build GRCh38
rs992956207 Chr.13: 112647937-112647937 on Build GRCh38
rs928432790 Chr.13: 112647573-112647573 on Build GRCh38
rs540113438 Chr.13: 112647504-112647504 on Build GRCh38
rs920102478 Chr.13: 112647834-112647834 on Build GRCh38
rs367940138 Chr.13: 112647589-112647589 on Build GRCh38
rs902483547 Chr.13: 112647936-112647936 on Build GRCh38

View Full Product Details