Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113480657-113481167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D2
Entrez Gene ID
55208
Gene Name
defective in cullin neddylation 1 domain containing 2
Gene Aliases
C13orf17
UniGene
Hs.682987
Gene Chromosome Location
Chr.13: 113455819-113492047 on Build GRCh38
Transcripts
NM_001014283.1 XM_017020643.1 XM_017020644.1 XM_017020645.1 XM_017020646.1 XM_017020647.1 XM_017020648.1 XM_017020649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6641163 c.249C>T p.V83V Chr.13: 113480715-113480715 on Build GRCh38
7148001 c.229G>A p.D77N Chr.13: 113480735-113480735 on Build GRCh38
3955419 c.264G>A p.Q88Q Chr.13: 113480700-113480700 on Build GRCh38
3955420 c.264G>A p.Q88Q Chr.13: 113480700-113480700 on Build GRCh38
7150971 c.223C>T p.P75S Chr.13: 113480741-113480741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935552136 Chr.13: 113481010-113481010 on Build GRCh38
rs11352484 Chr.13: 113480975-113480975 on Build GRCh38
rs765516110 Chr.13: 113480751-113480751 on Build GRCh38
rs34270770 Chr.13: 113480975-113480976 on Build GRCh38
rs985990199 Chr.13: 113481142-113481142 on Build GRCh38
rs940298468 Chr.13: 113480906-113480906 on Build GRCh38
rs758814846 Chr.13: 113480759-113480759 on Build GRCh38
rs193265693 Chr.13: 113480850-113480850 on Build GRCh38
rs529520008 Chr.13: 113480775-113480775 on Build GRCh38
rs1041080270 Chr.13: 113480975-113480975 on Build GRCh38
rs760731911 Chr.13: 113480703-113480703 on Build GRCh38
rs148536121 Chr.13: 113481104-113481104 on Build GRCh38
rs961715297 Chr.13: 113481111-113481111 on Build GRCh38
rs78649302 Chr.13: 113481059-113481059 on Build GRCh38
rs1037270087 Chr.13: 113480957-113480957 on Build GRCh38
rs552952214 Chr.13: 113480925-113480925 on Build GRCh38
rs115138574 Chr.13: 113480819-113480819 on Build GRCh38
rs748732696 Chr.13: 113480789-113480789 on Build GRCh38
rs376665865 Chr.13: 113480756-113480756 on Build GRCh38
rs994705481 Chr.13: 113481118-113481118 on Build GRCh38
rs187267836 Chr.13: 113480771-113480771 on Build GRCh38
rs777359537 Chr.13: 113480787-113480787 on Build GRCh38
rs527587130 Chr.13: 113480974-113480975 on Build GRCh38
rs780467596 Chr.13: 113480764-113480764 on Build GRCh38
rs1031536061 Chr.13: 113481132-113481132 on Build GRCh38
rs764219723 Chr.13: 113480709-113480709 on Build GRCh38
rs953295037 Chr.13: 113481126-113481126 on Build GRCh38
rs906303794 Chr.13: 113481097-113481097 on Build GRCh38
rs911313348 Chr.13: 113480851-113480851 on Build GRCh38
rs145359153 Chr.13: 113480743-113480743 on Build GRCh38
rs185726654 Chr.13: 113480998-113480998 on Build GRCh38
rs397851515 Chr.13: 113480985-113480986 on Build GRCh38
rs759802517 Chr.13: 113481127-113481127 on Build GRCh38
rs767490161 Chr.13: 113480701-113480701 on Build GRCh38
rs77254342 Chr.13: 113481122-113481122 on Build GRCh38
rs759273925 Chr.13: 113480692-113480692 on Build GRCh38
rs990222568 Chr.13: 113480807-113480807 on Build GRCh38
rs760932843 Chr.13: 113480936-113480936 on Build GRCh38
rs755639467 Chr.13: 113480776-113480776 on Build GRCh38
rs931866994 Chr.13: 113480845-113480845 on Build GRCh38
rs537179667 Chr.13: 113480710-113480710 on Build GRCh38
rs757556195 Chr.13: 113480739-113480739 on Build GRCh38
rs957182957 Chr.13: 113480801-113480801 on Build GRCh38
rs7319843 Chr.13: 113481102-113481102 on Build GRCh38
rs920409423 Chr.13: 113480825-113480825 on Build GRCh38
rs924477644 Chr.13: 113480785-113480785 on Build GRCh38
rs1054073591 Chr.13: 113481030-113481030 on Build GRCh38
rs528467675 Chr.13: 113480714-113480714 on Build GRCh38
rs539423249 Chr.13: 113481055-113481055 on Build GRCh38
rs893961375 Chr.13: 113481045-113481045 on Build GRCh38
rs561941389 Chr.13: 113481128-113481128 on Build GRCh38
rs775463956 Chr.13: 113480702-113480702 on Build GRCh38

View Full Product Details