Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17393896-17394407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI6
Entrez Gene ID
353238
Gene Name
peptidyl arginine deiminase 6
Gene Aliases
hPADVI
UniGene
Hs.531598
Gene Chromosome Location
Chr.1: 17372196-17401699 on Build GRCh38
Transcripts
NM_207421.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5889559 c.1182C>T p.S394S Chr.1: 17394302-17394302 on Build GRCh38
212570 c.1089C>T p.C363C Chr.1: 17393992-17393992 on Build GRCh38
1626646 c.1072-5T>C p.? Chr.1: 17393970-17393970 on Build GRCh38
463477 c.1283G>A p.G428E Chr.1: 17394403-17394403 on Build GRCh38
5508917 c.1248G>T p.G416G Chr.1: 17394368-17394368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747256729 Chr.1: 17394373-17394373 on Build GRCh38
rs904235705 Chr.1: 17393967-17393967 on Build GRCh38
rs564180756 Chr.1: 17394204-17394204 on Build GRCh38
rs767987621 Chr.1: 17394291-17394291 on Build GRCh38
rs745997829 Chr.1: 17394341-17394341 on Build GRCh38
rs754388713 Chr.1: 17394093-17394093 on Build GRCh38
rs753183749 Chr.1: 17394068-17394068 on Build GRCh38
rs764265777 Chr.1: 17394131-17394132 on Build GRCh38
rs368222037 Chr.1: 17394287-17394287 on Build GRCh38
rs768000252 Chr.1: 17394026-17394026 on Build GRCh38
rs749170626 Chr.1: 17394332-17394332 on Build GRCh38
rs560246479 Chr.1: 17394050-17394050 on Build GRCh38
rs557389130 Chr.1: 17394061-17394061 on Build GRCh38
rs942852013 Chr.1: 17394162-17394162 on Build GRCh38
rs745414523 Chr.1: 17394038-17394038 on Build GRCh38
rs746928284 Chr.1: 17393927-17393927 on Build GRCh38
rs761741338 Chr.1: 17393972-17393972 on Build GRCh38
rs755798854 Chr.1: 17394329-17394329 on Build GRCh38
rs777638791 Chr.1: 17394331-17394331 on Build GRCh38
rs774649171 Chr.1: 17394058-17394058 on Build GRCh38
rs754193133 Chr.1: 17394056-17394056 on Build GRCh38
rs777532256 Chr.1: 17394127-17394127 on Build GRCh38
rs776875102 Chr.1: 17393933-17393933 on Build GRCh38
rs145873927 Chr.1: 17394091-17394091 on Build GRCh38
rs749254020 Chr.1: 17394132-17394132 on Build GRCh38
rs779925985 Chr.1: 17394335-17394335 on Build GRCh38
rs183994985 Chr.1: 17393928-17393928 on Build GRCh38
rs6695849 Chr.1: 17393943-17393943 on Build GRCh38
rs372065243 Chr.1: 17394364-17394364 on Build GRCh38
rs201076846 Chr.1: 17394260-17394260 on Build GRCh38
rs532914694 Chr.1: 17394317-17394317 on Build GRCh38
rs866964896 Chr.1: 17394294-17394294 on Build GRCh38
rs74848593 Chr.1: 17394196-17394196 on Build GRCh38
rs114841763 Chr.1: 17393969-17393969 on Build GRCh38
rs761218220 Chr.1: 17394084-17394084 on Build GRCh38
rs202067511 Chr.1: 17394309-17394309 on Build GRCh38
rs770181514 Chr.1: 17393990-17393990 on Build GRCh38
rs778301791 Chr.1: 17393949-17393949 on Build GRCh38
rs201857683 Chr.1: 17394019-17394019 on Build GRCh38
rs76886447 Chr.1: 17394197-17394197 on Build GRCh38
rs765300452 Chr.1: 17393985-17393985 on Build GRCh38
rs34075690 Chr.1: 17394191-17394191 on Build GRCh38
rs374238645 Chr.1: 17394286-17394286 on Build GRCh38
rs1057517681 Chr.1: 17394041-17394041 on Build GRCh38
rs768459705 Chr.1: 17393968-17393968 on Build GRCh38
rs762892891 Chr.1: 17394055-17394055 on Build GRCh38
rs763433125 Chr.1: 17394036-17394036 on Build GRCh38
rs756745193 Chr.1: 17394032-17394032 on Build GRCh38
rs751136067 Chr.1: 17394301-17394301 on Build GRCh38
rs148192373 Chr.1: 17394191-17394191 on Build GRCh38
rs775320795 Chr.1: 17393962-17393962 on Build GRCh38
rs753200384 Chr.1: 17394031-17394031 on Build GRCh38
rs758043338 Chr.1: 17394039-17394039 on Build GRCh38
rs10458537 Chr.1: 17394119-17394119 on Build GRCh38
rs35163797 Chr.1: 17393996-17393997 on Build GRCh38
rs193052775 Chr.1: 17394095-17394095 on Build GRCh38
rs759837073 Chr.1: 17394276-17394276 on Build GRCh38
rs766559038 Chr.1: 17394016-17394016 on Build GRCh38
rs1031826474 Chr.1: 17394192-17394192 on Build GRCh38
rs148575053 Chr.1: 17394163-17394163 on Build GRCh38
rs774632854 Chr.1: 17394271-17394271 on Build GRCh38
rs1017376877 Chr.1: 17393956-17393956 on Build GRCh38
rs775979898 Chr.1: 17394285-17394285 on Build GRCh38
rs758560675 Chr.1: 17393926-17393926 on Build GRCh38
rs955892669 Chr.1: 17394017-17394017 on Build GRCh38
rs919899754 Chr.1: 17394193-17394193 on Build GRCh38
rs1012035864 Chr.1: 17394224-17394224 on Build GRCh38
rs552781402 Chr.1: 17394117-17394117 on Build GRCh38
rs12073549 Chr.1: 17394049-17394049 on Build GRCh38
rs779520691 Chr.1: 17394042-17394042 on Build GRCh38
rs751152775 Chr.1: 17394103-17394103 on Build GRCh38
rs754593236 Chr.1: 17394304-17394304 on Build GRCh38
rs187517746 Chr.1: 17393944-17393944 on Build GRCh38
rs1035887289 Chr.1: 17394270-17394270 on Build GRCh38
rs889215189 Chr.1: 17394216-17394216 on Build GRCh38
rs767982192 Chr.1: 17394065-17394065 on Build GRCh38
rs903186139 Chr.1: 17394230-17394231 on Build GRCh38
rs915679231 Chr.1: 17394160-17394160 on Build GRCh38
rs745525036 Chr.1: 17393950-17393950 on Build GRCh38
rs890011290 Chr.1: 17394154-17394154 on Build GRCh38
rs768370024 Chr.1: 17394047-17394047 on Build GRCh38
rs142799058 Chr.1: 17394203-17394203 on Build GRCh38
rs768841667 Chr.1: 17394376-17394376 on Build GRCh38
rs575136493 Chr.1: 17394165-17394165 on Build GRCh38
rs1051364044 Chr.1: 17394135-17394135 on Build GRCh38
rs770970009 Chr.1: 17394338-17394338 on Build GRCh38
rs781536467 Chr.1: 17394121-17394122 on Build GRCh38
rs1043079252 Chr.1: 17394229-17394229 on Build GRCh38
rs772467361 Chr.1: 17394280-17394280 on Build GRCh38
rs779618404 Chr.1: 17394101-17394101 on Build GRCh38
rs751853751 Chr.1: 17394018-17394018 on Build GRCh38
rs541767432 Chr.1: 17394122-17394122 on Build GRCh38
rs746851853 Chr.1: 17393964-17393964 on Build GRCh38
rs999984564 Chr.1: 17394173-17394173 on Build GRCh38
rs770860771 Chr.1: 17394256-17394256 on Build GRCh38
rs374146973 Chr.1: 17393931-17393931 on Build GRCh38
rs777110271 Chr.1: 17394380-17394380 on Build GRCh38
rs374615037 Chr.1: 17394014-17394014 on Build GRCh38
rs749259717 Chr.1: 17394382-17394382 on Build GRCh38
rs750522486 Chr.1: 17394291-17394292 on Build GRCh38
rs771692673 Chr.1: 17393952-17393952 on Build GRCh38
rs569705862 Chr.1: 17393946-17393946 on Build GRCh38
rs999093972 Chr.1: 17394252-17394252 on Build GRCh38
rs977643624 Chr.1: 17394143-17394143 on Build GRCh38
rs756208047 Chr.1: 17394314-17394315 on Build GRCh38
rs754681383 Chr.1: 17394108-17394108 on Build GRCh38
rs960223209 Chr.1: 17394151-17394151 on Build GRCh38
rs112098336 Chr.1: 17394046-17394046 on Build GRCh38

View Full Product Details