Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17409884-17410377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCC2
Entrez Gene ID
55920
Gene Name
regulator of chromosome condensation 2
Gene Aliases
TD-60
UniGene
Hs.380857
Gene Chromosome Location
Chr.1: 17406755-17439754 on Build GRCh38
Transcripts
NM_001136204.2 NM_018715.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2094377 c.1387-2A>G p.? Chr.1: 17410053-17410053 on Build GRCh38
1473043 c.1455C>G p.F485L Chr.1: 17409983-17409983 on Build GRCh38
341828 c.1429G>A p.E477K Chr.1: 17410009-17410009 on Build GRCh38
1185345 c.1405C>G p.P469A Chr.1: 17410033-17410033 on Build GRCh38
5680298 c.1397A>G p.D466G Chr.1: 17410041-17410041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973032817 Chr.1: 17410162-17410162 on Build GRCh38
rs752583266 Chr.1: 17410033-17410033 on Build GRCh38
rs371533435 Chr.1: 17410001-17410001 on Build GRCh38
rs554054710 Chr.1: 17410046-17410046 on Build GRCh38
rs759400591 Chr.1: 17409926-17409926 on Build GRCh38
rs929196387 Chr.1: 17410184-17410184 on Build GRCh38
rs544643718 Chr.1: 17410175-17410175 on Build GRCh38
rs761231932 Chr.1: 17410090-17410090 on Build GRCh38
rs774817750 Chr.1: 17409947-17409947 on Build GRCh38
rs752129388 Chr.1: 17410272-17410272 on Build GRCh38
rs768015163 Chr.1: 17409950-17409950 on Build GRCh38
rs181669428 Chr.1: 17409935-17409935 on Build GRCh38
rs1000238836 Chr.1: 17410356-17410356 on Build GRCh38
rs749489054 Chr.1: 17410048-17410048 on Build GRCh38
rs759937850 Chr.1: 17409948-17409948 on Build GRCh38
rs754988042 Chr.1: 17409929-17409929 on Build GRCh38
rs940101144 Chr.1: 17410119-17410119 on Build GRCh38
rs3752351 Chr.1: 17409936-17409936 on Build GRCh38
rs779021248 Chr.1: 17410075-17410075 on Build GRCh38
rs926291179 Chr.1: 17410084-17410084 on Build GRCh38
rs775889569 Chr.1: 17409959-17409959 on Build GRCh38
rs757552588 Chr.1: 17410207-17410207 on Build GRCh38
rs868134384 Chr.1: 17410210-17410210 on Build GRCh38
rs368014486 Chr.1: 17409942-17409942 on Build GRCh38
rs2762890 Chr.1: 17410082-17410082 on Build GRCh38
rs143136960 Chr.1: 17410253-17410253 on Build GRCh38
rs763195391 Chr.1: 17410081-17410081 on Build GRCh38
rs542976508 Chr.1: 17410118-17410118 on Build GRCh38
rs750065624 Chr.1: 17410002-17410002 on Build GRCh38
rs959275070 Chr.1: 17409953-17409953 on Build GRCh38
rs771241567 Chr.1: 17410042-17410042 on Build GRCh38
rs370053181 Chr.1: 17410039-17410039 on Build GRCh38
rs781111347 Chr.1: 17409933-17409933 on Build GRCh38
rs1000458663 Chr.1: 17409991-17409991 on Build GRCh38
rs758117369 Chr.1: 17410007-17410007 on Build GRCh38
rs902251303 Chr.1: 17409939-17409939 on Build GRCh38
rs773504113 Chr.1: 17409938-17409938 on Build GRCh38
rs941664201 Chr.1: 17410141-17410141 on Build GRCh38
rs943469475 Chr.1: 17410274-17410274 on Build GRCh38
rs139019005 Chr.1: 17410010-17410010 on Build GRCh38
rs531357924 Chr.1: 17410129-17410129 on Build GRCh38
rs751423219 Chr.1: 17410019-17410019 on Build GRCh38
rs372433444 Chr.1: 17410056-17410056 on Build GRCh38
rs147488477 Chr.1: 17410283-17410283 on Build GRCh38
rs908656932 Chr.1: 17410117-17410117 on Build GRCh38
rs771327928 Chr.1: 17410074-17410074 on Build GRCh38
rs769292274 Chr.1: 17410093-17410093 on Build GRCh38
rs1050046473 Chr.1: 17410242-17410242 on Build GRCh38
rs754751310 Chr.1: 17410022-17410022 on Build GRCh38
rs375045285 Chr.1: 17409941-17409941 on Build GRCh38
rs761244162 Chr.1: 17409992-17409992 on Build GRCh38
rs368478824 Chr.1: 17409928-17409928 on Build GRCh38
rs904600233 Chr.1: 17410295-17410295 on Build GRCh38
rs781144883 Chr.1: 17410029-17410029 on Build GRCh38
rs1050395710 Chr.1: 17409912-17409912 on Build GRCh38
rs976810083 Chr.1: 17410170-17410170 on Build GRCh38
rs746199708 Chr.1: 17410077-17410077 on Build GRCh38
rs569263753 Chr.1: 17409927-17409927 on Build GRCh38
rs772324607 Chr.1: 17410078-17410078 on Build GRCh38
rs375043018 Chr.1: 17410006-17410006 on Build GRCh38
rs562748961 Chr.1: 17410121-17410121 on Build GRCh38

View Full Product Details