Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17412033-17412532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCC2
Entrez Gene ID
55920
Gene Name
regulator of chromosome condensation 2
Gene Aliases
TD-60
UniGene
Hs.380857
Gene Chromosome Location
Chr.1: 17406755-17439754 on Build GRCh38
Transcripts
NM_001136204.2 NM_018715.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2094379 c.1354A>G p.S452G Chr.1: 17412154-17412154 on Build GRCh38
1295534 c.1342G>A p.E448K Chr.1: 17412166-17412166 on Build GRCh38
5509279 c.1374C>A p.T458T Chr.1: 17412134-17412134 on Build GRCh38
7414306 c.1359G>T p.W453C Chr.1: 17412149-17412149 on Build GRCh38
4720878 c.1364C>T p.P455L Chr.1: 17412144-17412144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003175976 Chr.1: 17412386-17412386 on Build GRCh38
rs565993530 Chr.1: 17412068-17412068 on Build GRCh38
rs558106285 Chr.1: 17412140-17412140 on Build GRCh38
rs536783186 Chr.1: 17412104-17412104 on Build GRCh38
rs897800217 Chr.1: 17412369-17412369 on Build GRCh38
rs545548568 Chr.1: 17412418-17412418 on Build GRCh38
rs921402549 Chr.1: 17412298-17412298 on Build GRCh38
rs777149738 Chr.1: 17412242-17412242 on Build GRCh38
rs983182657 Chr.1: 17412167-17412167 on Build GRCh38
rs992765209 Chr.1: 17412507-17412507 on Build GRCh38
rs376312124 Chr.1: 17412134-17412134 on Build GRCh38
rs993132391 Chr.1: 17412297-17412297 on Build GRCh38
rs146457201 Chr.1: 17412355-17412355 on Build GRCh38
rs779121668 Chr.1: 17412185-17412185 on Build GRCh38
rs951541316 Chr.1: 17412446-17412446 on Build GRCh38
rs111826586 Chr.1: 17412120-17412120 on Build GRCh38
rs536139965 Chr.1: 17412216-17412216 on Build GRCh38
rs770352187 Chr.1: 17412245-17412245 on Build GRCh38
rs569779443 Chr.1: 17412223-17412223 on Build GRCh38
rs202008853 Chr.1: 17412453-17412453 on Build GRCh38
rs965668723 Chr.1: 17412469-17412469 on Build GRCh38
rs111465289 Chr.1: 17412276-17412277 on Build GRCh38
rs759309869 Chr.1: 17412137-17412137 on Build GRCh38
rs769204274 Chr.1: 17412237-17412237 on Build GRCh38
rs112439473 Chr.1: 17412456-17412456 on Build GRCh38
rs397979380 Chr.1: 17412077-17412077 on Build GRCh38
rs1041757968 Chr.1: 17412381-17412381 on Build GRCh38
rs41310395 Chr.1: 17412057-17412057 on Build GRCh38
rs995528026 Chr.1: 17412133-17412133 on Build GRCh38
rs571971967 Chr.1: 17412331-17412331 on Build GRCh38
rs890594848 Chr.1: 17412211-17412211 on Build GRCh38
rs561301226 Chr.1: 17412508-17412508 on Build GRCh38
rs756158591 Chr.1: 17412153-17412153 on Build GRCh38
rs541737417 Chr.1: 17412491-17412491 on Build GRCh38
rs907146690 Chr.1: 17412382-17412382 on Build GRCh38
rs534323678 Chr.1: 17412338-17412338 on Build GRCh38
rs757310152 Chr.1: 17412173-17412173 on Build GRCh38
rs189256697 Chr.1: 17412450-17412450 on Build GRCh38
rs772752363 Chr.1: 17412076-17412076 on Build GRCh38
rs370262803 Chr.1: 17412231-17412231 on Build GRCh38
rs71014950 Chr.1: 17412075-17412075 on Build GRCh38
rs1031580208 Chr.1: 17412249-17412249 on Build GRCh38
rs369388198 Chr.1: 17412088-17412088 on Build GRCh38
rs183344854 Chr.1: 17412391-17412391 on Build GRCh38
rs7522509 Chr.1: 17412248-17412248 on Build GRCh38
rs917093869 Chr.1: 17412300-17412300 on Build GRCh38
rs541247461 Chr.1: 17412359-17412359 on Build GRCh38
rs373162219 Chr.1: 17412101-17412101 on Build GRCh38
rs772467824 Chr.1: 17412205-17412205 on Build GRCh38
rs567458904 Chr.1: 17412143-17412143 on Build GRCh38
rs1006216004 Chr.1: 17412055-17412055 on Build GRCh38
rs777976805 Chr.1: 17412169-17412169 on Build GRCh38
rs770814086 Chr.1: 17412074-17412074 on Build GRCh38
rs1004239242 Chr.1: 17412451-17412451 on Build GRCh38
rs192299170 Chr.1: 17412366-17412366 on Build GRCh38
rs768245227 Chr.1: 17412345-17412345 on Build GRCh38
rs894275986 Chr.1: 17412393-17412393 on Build GRCh38
rs57188450 Chr.1: 17412412-17412412 on Build GRCh38
rs566232151 Chr.1: 17412070-17412070 on Build GRCh38
rs931737144 Chr.1: 17412323-17412323 on Build GRCh38
rs753920683 Chr.1: 17412170-17412170 on Build GRCh38
rs746088278 Chr.1: 17412204-17412204 on Build GRCh38
rs568999075 Chr.1: 17412273-17412274 on Build GRCh38
rs538800554 Chr.1: 17412244-17412244 on Build GRCh38
rs759310049 Chr.1: 17412321-17412321 on Build GRCh38
rs565586868 Chr.1: 17412421-17412421 on Build GRCh38
rs112978140 Chr.1: 17412365-17412365 on Build GRCh38
rs1027063487 Chr.1: 17412436-17412436 on Build GRCh38
rs890663550 Chr.1: 17412394-17412394 on Build GRCh38
rs904269705 Chr.1: 17412420-17412420 on Build GRCh38

View Full Product Details