Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 18634943-18635441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX7
Entrez Gene ID
5081
Gene Name
paired box 7
Gene Aliases
HUP1, PAX7B, RMS2
UniGene
Hs.113253
Gene Chromosome Location
Chr.1: 18631006-18748866 on Build GRCh38
Transcripts
NM_001135254.1 NM_002584.2 NM_013945.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6527520 c.351G>C p.K117N Chr.1: 18635140-18635140 on Build GRCh38
4395214 c.351G>A p.K117K Chr.1: 18635140-18635140 on Build GRCh38
6738282 c.345G>A p.E115E Chr.1: 18635134-18635134 on Build GRCh38
233046 c.370G>A p.E124K Chr.1: 18635159-18635159 on Build GRCh38
6527521 c.351G>C p.K117N Chr.1: 18635140-18635140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113911034 Chr.1: 18635259-18635259 on Build GRCh38
rs369367031 Chr.1: 18635248-18635248 on Build GRCh38
rs1050957153 Chr.1: 18635148-18635148 on Build GRCh38
rs746707682 Chr.1: 18635062-18635062 on Build GRCh38
rs988376122 Chr.1: 18634971-18634971 on Build GRCh38
rs774565338 Chr.1: 18635118-18635118 on Build GRCh38
rs532794942 Chr.1: 18635379-18635379 on Build GRCh38
rs747944975 Chr.1: 18635071-18635071 on Build GRCh38
rs762305801 Chr.1: 18635129-18635129 on Build GRCh38
rs183827251 Chr.1: 18635260-18635260 on Build GRCh38
rs577857072 Chr.1: 18635140-18635140 on Build GRCh38
rs770225389 Chr.1: 18635268-18635268 on Build GRCh38
rs72937278 Chr.1: 18635058-18635058 on Build GRCh38
rs1034002176 Chr.1: 18635324-18635324 on Build GRCh38
rs897598525 Chr.1: 18635332-18635332 on Build GRCh38
rs751059274 Chr.1: 18635136-18635136 on Build GRCh38
rs1038984689 Chr.1: 18635401-18635401 on Build GRCh38
rs956896288 Chr.1: 18635029-18635029 on Build GRCh38
rs951282858 Chr.1: 18635366-18635366 on Build GRCh38
rs750568811 Chr.1: 18635062-18635062 on Build GRCh38
rs773711564 Chr.1: 18635271-18635271 on Build GRCh38
rs557731665 Chr.1: 18634985-18634985 on Build GRCh38
rs575952185 Chr.1: 18634992-18634992 on Build GRCh38
rs751061934 Chr.1: 18635213-18635213 on Build GRCh38
rs780858389 Chr.1: 18635141-18635141 on Build GRCh38
rs760663197 Chr.1: 18635144-18635146 on Build GRCh38
rs1051639583 Chr.1: 18634989-18634989 on Build GRCh38
rs375962225 Chr.1: 18635249-18635249 on Build GRCh38
rs1031116612 Chr.1: 18635027-18635027 on Build GRCh38
rs993147367 Chr.1: 18635079-18635079 on Build GRCh38
rs546225863 Chr.1: 18635349-18635349 on Build GRCh38
rs775727650 Chr.1: 18635189-18635189 on Build GRCh38
rs368527949 Chr.1: 18635250-18635250 on Build GRCh38
rs770442530 Chr.1: 18635312-18635312 on Build GRCh38
rs372764200 Chr.1: 18635085-18635085 on Build GRCh38
rs539584877 Chr.1: 18634981-18634981 on Build GRCh38
rs759037398 Chr.1: 18635215-18635215 on Build GRCh38
rs904072815 Chr.1: 18635011-18635011 on Build GRCh38
rs547164637 Chr.1: 18635335-18635335 on Build GRCh38
rs1051875461 Chr.1: 18635373-18635373 on Build GRCh38
rs1024447151 Chr.1: 18635065-18635065 on Build GRCh38
rs1007887514 Chr.1: 18635385-18635385 on Build GRCh38
rs780899110 Chr.1: 18635067-18635067 on Build GRCh38
rs890382154 Chr.1: 18635096-18635096 on Build GRCh38
rs371257583 Chr.1: 18635151-18635151 on Build GRCh38
rs368444827 Chr.1: 18635410-18635410 on Build GRCh38
rs769651150 Chr.1: 18635073-18635073 on Build GRCh38
rs776969728 Chr.1: 18635202-18635202 on Build GRCh38
rs764601304 Chr.1: 18635126-18635126 on Build GRCh38
rs942768268 Chr.1: 18635404-18635404 on Build GRCh38
rs1007357826 Chr.1: 18635102-18635102 on Build GRCh38
rs754279860 Chr.1: 18635128-18635128 on Build GRCh38
rs373901126 Chr.1: 18635164-18635164 on Build GRCh38
rs758972552 Chr.1: 18635277-18635277 on Build GRCh38
rs187521955 Chr.1: 18635014-18635014 on Build GRCh38
rs72937282 Chr.1: 18635197-18635197 on Build GRCh38
rs539004358 Chr.1: 18634970-18634970 on Build GRCh38
rs540805473 Chr.1: 18635086-18635086 on Build GRCh38
rs137899505 Chr.1: 18635252-18635252 on Build GRCh38
rs768846421 Chr.1: 18635254-18635254 on Build GRCh38
rs949553842 Chr.1: 18634988-18634988 on Build GRCh38
rs768978188 Chr.1: 18634991-18634991 on Build GRCh38
rs573472332 Chr.1: 18635081-18635081 on Build GRCh38
rs754647717 Chr.1: 18635066-18635066 on Build GRCh38
rs758413235 Chr.1: 18635221-18635221 on Build GRCh38
rs376411971 Chr.1: 18635121-18635121 on Build GRCh38
rs141499812 Chr.1: 18635119-18635119 on Build GRCh38
rs745925674 Chr.1: 18635173-18635173 on Build GRCh38
rs1001327554 Chr.1: 18635314-18635317 on Build GRCh38
rs752326328 Chr.1: 18635222-18635222 on Build GRCh38
rs762358727 Chr.1: 18635211-18635211 on Build GRCh38
rs760954795 Chr.1: 18635190-18635190 on Build GRCh38
rs777766224 Chr.1: 18635243-18635243 on Build GRCh38
rs373323044 Chr.1: 18635100-18635100 on Build GRCh38
rs912601399 Chr.1: 18635149-18635149 on Build GRCh38
rs1037449423 Chr.1: 18635013-18635013 on Build GRCh38
rs770976315 Chr.1: 18635165-18635165 on Build GRCh38
rs971552899 Chr.1: 18635306-18635306 on Build GRCh38
rs77994984 Chr.1: 18635375-18635375 on Build GRCh38
rs528633995 Chr.1: 18635289-18635289 on Build GRCh38
rs778904746 Chr.1: 18635171-18635171 on Build GRCh38
rs371150760 Chr.1: 18635125-18635125 on Build GRCh38
rs374019969 Chr.1: 18635317-18635320 on Build GRCh38
rs192807434 Chr.1: 18635247-18635247 on Build GRCh38
rs368102853 Chr.1: 18635175-18635175 on Build GRCh38
rs767089117 Chr.1: 18635220-18635220 on Build GRCh38
rs755821765 Chr.1: 18635223-18635223 on Build GRCh38
rs765933792 Chr.1: 18635131-18635131 on Build GRCh38
rs777856039 Chr.1: 18635158-18635158 on Build GRCh38
rs765702601 Chr.1: 18635212-18635212 on Build GRCh38
rs941784032 Chr.1: 18635008-18635008 on Build GRCh38
rs1030183372 Chr.1: 18635084-18635084 on Build GRCh38
rs142754204 Chr.1: 18635124-18635124 on Build GRCh38
rs139680453 Chr.1: 18635154-18635154 on Build GRCh38

View Full Product Details