Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20070118-20070621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G5
Entrez Gene ID
5322
Gene Name
phospholipase A2 group V
Gene Aliases
FRFB, GV-PLA2, PLA2-10, hVPLA(2)
UniGene
Hs.319438
Gene Chromosome Location
Chr.1: 20028350-20091901 on Build GRCh38
Transcripts
NM_000929.2 XM_005245891.4 XM_005245892.4 XM_005245893.4 XM_011541586.2 XM_011541587.2 XM_011541588.2 XM_011541589.2 XM_011541590.2 XM_011541591.2 XM_011541592.2

SNP Detail

SNP ID Chromosome Location
rs3806311 Chr.1: 20070155-20070155 on Build GRCh38
rs911906243 Chr.1: 20070252-20070252 on Build GRCh38
rs559778671 Chr.1: 20070582-20070582 on Build GRCh38
rs533947062 Chr.1: 20070374-20070374 on Build GRCh38
rs1026807089 Chr.1: 20070511-20070511 on Build GRCh38
rs117321541 Chr.1: 20070409-20070409 on Build GRCh38
rs1016740267 Chr.1: 20070424-20070424 on Build GRCh38
rs961503592 Chr.1: 20070466-20070466 on Build GRCh38
rs989028417 Chr.1: 20070584-20070584 on Build GRCh38
rs35165482 Chr.1: 20070422-20070423 on Build GRCh38
rs184330725 Chr.1: 20070543-20070543 on Build GRCh38
rs949160435 Chr.1: 20070388-20070388 on Build GRCh38
rs1014141316 Chr.1: 20070246-20070246 on Build GRCh38
rs910718488 Chr.1: 20070330-20070330 on Build GRCh38
rs936429893 Chr.1: 20070220-20070220 on Build GRCh38
rs955000210 Chr.1: 20070513-20070513 on Build GRCh38
rs771337394 Chr.1: 20070258-20070258 on Build GRCh38
rs76363809 Chr.1: 20070221-20070221 on Build GRCh38
rs11573194 Chr.1: 20070512-20070512 on Build GRCh38
rs963332209 Chr.1: 20070282-20070282 on Build GRCh38
rs1044126197 Chr.1: 20070396-20070396 on Build GRCh38
rs558722783 Chr.1: 20070381-20070381 on Build GRCh38
rs993559711 Chr.1: 20070479-20070479 on Build GRCh38
rs993467299 Chr.1: 20070503-20070503 on Build GRCh38
rs566193771 Chr.1: 20070371-20070371 on Build GRCh38
rs558099222 Chr.1: 20070382-20070382 on Build GRCh38
rs549864801 Chr.1: 20070257-20070257 on Build GRCh38
rs668630 Chr.1: 20070181-20070181 on Build GRCh38
rs1018211424 Chr.1: 20070540-20070540 on Build GRCh38
rs3806313 Chr.1: 20070240-20070240 on Build GRCh38
rs1051967552 Chr.1: 20070187-20070187 on Build GRCh38
rs574071476 Chr.1: 20070554-20070554 on Build GRCh38
rs796364287 Chr.1: 20070380-20070380 on Build GRCh38
rs1052759460 Chr.1: 20070236-20070236 on Build GRCh38
rs370294820 Chr.1: 20070237-20070239 on Build GRCh38
rs954782801 Chr.1: 20070310-20070310 on Build GRCh38
rs905014434 Chr.1: 20070397-20070397 on Build GRCh38
rs667147 Chr.1: 20070557-20070557 on Build GRCh38
rs972574701 Chr.1: 20070299-20070299 on Build GRCh38
rs1005388883 Chr.1: 20070254-20070254 on Build GRCh38
rs1026370804 Chr.1: 20070309-20070309 on Build GRCh38
rs879519797 Chr.1: 20070278-20070278 on Build GRCh38
rs910804180 Chr.1: 20070188-20070188 on Build GRCh38
rs1043655678 Chr.1: 20070199-20070199 on Build GRCh38
rs1039810526 Chr.1: 20070198-20070198 on Build GRCh38
rs972016933 Chr.1: 20070307-20070307 on Build GRCh38
rs936335026 Chr.1: 20070379-20070379 on Build GRCh38
rs917002208 Chr.1: 20070589-20070589 on Build GRCh38
rs943595373 Chr.1: 20070197-20070197 on Build GRCh38
rs775723095 Chr.1: 20070333-20070333 on Build GRCh38
rs899058962 Chr.1: 20070433-20070433 on Build GRCh38
rs1008969781 Chr.1: 20070527-20070527 on Build GRCh38
rs978989385 Chr.1: 20070370-20070370 on Build GRCh38
rs537771779 Chr.1: 20070276-20070276 on Build GRCh38
rs1046691712 Chr.1: 20070389-20070389 on Build GRCh38
rs933510464 Chr.1: 20070162-20070162 on Build GRCh38
rs193295822 Chr.1: 20070361-20070361 on Build GRCh38
rs905146445 Chr.1: 20070269-20070269 on Build GRCh38
rs943647262 Chr.1: 20070348-20070348 on Build GRCh38
rs867697827 Chr.1: 20070413-20070413 on Build GRCh38
rs910800655 Chr.1: 20070337-20070337 on Build GRCh38
rs748427019 Chr.1: 20070544-20070544 on Build GRCh38
rs370339787 Chr.1: 20070235-20070237 on Build GRCh38
rs1037602471 Chr.1: 20070432-20070432 on Build GRCh38
rs2020885 Chr.1: 20070329-20070329 on Build GRCh38
rs541519005 Chr.1: 20070137-20070137 on Build GRCh38
rs866009083 Chr.1: 20070593-20070593 on Build GRCh38
rs934833165 Chr.1: 20070583-20070583 on Build GRCh38
rs903506750 Chr.1: 20070201-20070201 on Build GRCh38
rs1043217015 Chr.1: 20070368-20070368 on Build GRCh38
rs11573193 Chr.1: 20070458-20070458 on Build GRCh38
rs776813378 Chr.1: 20070275-20070275 on Build GRCh38
rs956234343 Chr.1: 20070573-20070573 on Build GRCh38
rs943646419 Chr.1: 20070331-20070331 on Build GRCh38
rs116287501 Chr.1: 20070248-20070248 on Build GRCh38

View Full Product Details