Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 79322551-79323045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMED3
Entrez Gene ID
23423
Gene Name
transmembrane p24 trafficking protein 3
Gene Aliases
C15orf22, P24B, p24g4, p26
UniGene
Hs.513058
Gene Chromosome Location
Chr.15: 79311062-79419879 on Build GRCh38
Transcripts
NM_001301203.1 NM_007364.3 NR_125394.1 XM_017022040.1

SNP Detail

SNP ID Chromosome Location
rs922455026 Chr.15: 79322724-79322724 on Build GRCh38
rs1045198404 Chr.15: 79323022-79323022 on Build GRCh38
rs1050111617 Chr.15: 79322735-79322735 on Build GRCh38
rs966072564 Chr.15: 79322910-79322910 on Build GRCh38
rs113557327 Chr.15: 79322794-79322794 on Build GRCh38
rs533585250 Chr.15: 79322837-79322837 on Build GRCh38
rs57004304 Chr.15: 79322619-79322619 on Build GRCh38
rs896638043 Chr.15: 79322713-79322713 on Build GRCh38
rs74326713 Chr.15: 79322826-79322826 on Build GRCh38
rs572368905 Chr.15: 79322902-79322902 on Build GRCh38
rs983840625 Chr.15: 79322905-79322905 on Build GRCh38
rs976148344 Chr.15: 79322911-79322911 on Build GRCh38
rs959533039 Chr.15: 79322832-79322832 on Build GRCh38
rs150656724 Chr.15: 79322811-79322811 on Build GRCh38
rs930083613 Chr.15: 79322889-79322889 on Build GRCh38
rs955809892 Chr.15: 79322801-79322802 on Build GRCh38
rs771632506 Chr.15: 79322640-79322640 on Build GRCh38
rs932552164 Chr.15: 79322726-79322726 on Build GRCh38
rs139739775 Chr.15: 79322696-79322696 on Build GRCh38
rs915131318 Chr.15: 79322975-79322975 on Build GRCh38
rs1024318235 Chr.15: 79322747-79322747 on Build GRCh38
rs941365819 Chr.15: 79322579-79322579 on Build GRCh38
rs139872152 Chr.15: 79322838-79322838 on Build GRCh38
rs112815175 Chr.15: 79322712-79322712 on Build GRCh38
rs764235745 Chr.15: 79322675-79322675 on Build GRCh38
rs946677354 Chr.15: 79322977-79322977 on Build GRCh38
rs542732928 Chr.15: 79322896-79322896 on Build GRCh38
rs907956248 Chr.15: 79322636-79322636 on Build GRCh38
rs1038810349 Chr.15: 79322873-79322873 on Build GRCh38
rs968296800 Chr.15: 79322781-79322781 on Build GRCh38
rs555576188 Chr.15: 79322970-79322970 on Build GRCh38
rs904143386 Chr.15: 79323023-79323023 on Build GRCh38
rs1012835143 Chr.15: 79323004-79323004 on Build GRCh38
rs368305819 Chr.15: 79322815-79322815 on Build GRCh38
rs988547693 Chr.15: 79322803-79322803 on Build GRCh38
rs897285014 Chr.15: 79322667-79322667 on Build GRCh38
rs932475305 Chr.15: 79322926-79322926 on Build GRCh38
rs918574280 Chr.15: 79322880-79322880 on Build GRCh38
rs907901843 Chr.15: 79322907-79322907 on Build GRCh38
rs896544770 Chr.15: 79322932-79322932 on Build GRCh38
rs893763995 Chr.15: 79322751-79322751 on Build GRCh38
rs112375220 Chr.15: 79322618-79322618 on Build GRCh38
rs149838342 Chr.15: 79322890-79322890 on Build GRCh38
rs985346317 Chr.15: 79322972-79322972 on Build GRCh38
rs775882779 Chr.15: 79322913-79322913 on Build GRCh38
rs567683343 Chr.15: 79322601-79322601 on Build GRCh38
rs547755411 Chr.15: 79322592-79322592 on Build GRCh38
rs994203162 Chr.15: 79322706-79322706 on Build GRCh38
rs1010982186 Chr.15: 79322758-79322758 on Build GRCh38
rs922452994 Chr.15: 79322811-79322812 on Build GRCh38
rs1032679525 Chr.15: 79322797-79322797 on Build GRCh38

View Full Product Details