Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 18807634-18808149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMG1
Entrez Gene ID
23049
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Gene Aliases
61E3.4, ATX, LIP
UniGene
Hs.460179
Gene Chromosome Location
Chr.16: 18804853-18926417 on Build GRCh38
Transcripts
NM_015092.4 XM_005255182.3 XM_005255183.3 XM_005255184.3 XM_011545769.2 XM_011545770.1 XM_017023066.1 XM_017023067.1

SNP Detail

SNP ID Chromosome Location
rs962890246 Chr.16: 18807896-18807896 on Build GRCh38
rs187443878 Chr.16: 18807686-18807686 on Build GRCh38
rs1046851645 Chr.16: 18807885-18807885 on Build GRCh38
rs577539517 Chr.16: 18807846-18807846 on Build GRCh38
rs527427337 Chr.16: 18807935-18807935 on Build GRCh38
rs867978896 Chr.16: 18807829-18807829 on Build GRCh38
rs761400093 Chr.16: 18808106-18808106 on Build GRCh38
rs1037378221 Chr.16: 18807721-18807722 on Build GRCh38
rs1020374296 Chr.16: 18807963-18807963 on Build GRCh38
rs1006878150 Chr.16: 18808050-18808050 on Build GRCh38
rs988944980 Chr.16: 18807800-18807800 on Build GRCh38
rs889806000 Chr.16: 18808042-18808042 on Build GRCh38
rs960107730 Chr.16: 18807926-18807926 on Build GRCh38
rs1006426711 Chr.16: 18807709-18807709 on Build GRCh38
rs1029865241 Chr.16: 18808020-18808021 on Build GRCh38
rs1048350946 Chr.16: 18807699-18807699 on Build GRCh38
rs1056364807 Chr.16: 18807927-18807927 on Build GRCh38
rs903590644 Chr.16: 18807983-18807983 on Build GRCh38
rs1025492802 Chr.16: 18808080-18808080 on Build GRCh38
rs904728812 Chr.16: 18807749-18807749 on Build GRCh38
rs559800644 Chr.16: 18807917-18807917 on Build GRCh38
rs985986110 Chr.16: 18807822-18807822 on Build GRCh38
rs1031913933 Chr.16: 18807750-18807750 on Build GRCh38
rs976732910 Chr.16: 18808004-18808004 on Build GRCh38
rs968540343 Chr.16: 18807819-18807819 on Build GRCh38
rs1040785306 Chr.16: 18807682-18807682 on Build GRCh38
rs1010695800 Chr.16: 18807960-18807960 on Build GRCh38
rs142305294 Chr.16: 18807984-18807984 on Build GRCh38
rs544852194 Chr.16: 18807765-18807765 on Build GRCh38
rs376376696 Chr.16: 18808112-18808112 on Build GRCh38
rs906238937 Chr.16: 18808001-18808001 on Build GRCh38
rs929660504 Chr.16: 18808041-18808041 on Build GRCh38
rs544635060 Chr.16: 18807905-18807905 on Build GRCh38
rs956308648 Chr.16: 18807766-18807766 on Build GRCh38
rs1016474126 Chr.16: 18807730-18807730 on Build GRCh38
rs556034753 Chr.16: 18807731-18807731 on Build GRCh38
rs1018145168 Chr.16: 18807992-18807992 on Build GRCh38
rs922323238 Chr.16: 18807868-18807868 on Build GRCh38
rs1056162195 Chr.16: 18807742-18807745 on Build GRCh38
rs913053396 Chr.16: 18807809-18807809 on Build GRCh38
rs1041092936 Chr.16: 18807861-18807861 on Build GRCh38
rs1017317384 Chr.16: 18808057-18808057 on Build GRCh38
rs1043476006 Chr.16: 18807748-18807749 on Build GRCh38
rs189836708 Chr.16: 18808069-18808069 on Build GRCh38
rs551622566 Chr.16: 18808013-18808013 on Build GRCh38
rs751919129 Chr.16: 18807742-18807743 on Build GRCh38
rs942317770 Chr.16: 18807852-18807852 on Build GRCh38
rs528261916 Chr.16: 18807929-18807929 on Build GRCh38
rs865886675 Chr.16: 18807916-18807916 on Build GRCh38
rs963946181 Chr.16: 18808003-18808003 on Build GRCh38
rs113629541 Chr.16: 18808079-18808079 on Build GRCh38
rs1000352555 Chr.16: 18807749-18807752 on Build GRCh38
rs1037889516 Chr.16: 18807908-18807908 on Build GRCh38
rs374458531 Chr.16: 18807687-18807687 on Build GRCh38
rs918492280 Chr.16: 18807725-18807725 on Build GRCh38
rs1036091693 Chr.16: 18807921-18807921 on Build GRCh38
rs886645533 Chr.16: 18807705-18807705 on Build GRCh38
rs556598323 Chr.16: 18807781-18807781 on Build GRCh38
rs922260852 Chr.16: 18808012-18808012 on Build GRCh38
rs929709969 Chr.16: 18807884-18807884 on Build GRCh38
rs982906196 Chr.16: 18808052-18808052 on Build GRCh38
rs993957052 Chr.16: 18807688-18807688 on Build GRCh38
rs1043890391 Chr.16: 18807974-18807974 on Build GRCh38
rs879873487 Chr.16: 18808065-18808065 on Build GRCh38
rs540446238 Chr.16: 18808099-18808100 on Build GRCh38
rs561091258 Chr.16: 18807991-18807991 on Build GRCh38
rs1029626332 Chr.16: 18807794-18807794 on Build GRCh38
rs886569812 Chr.16: 18807904-18807904 on Build GRCh38
rs148748468 Chr.16: 18807747-18807747 on Build GRCh38
rs942350200 Chr.16: 18807665-18807665 on Build GRCh38
rs941197996 Chr.16: 18807716-18807716 on Build GRCh38
rs1035717684 Chr.16: 18807793-18807793 on Build GRCh38
rs960385714 Chr.16: 18807796-18807796 on Build GRCh38

View Full Product Details