Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68294366-68294866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A6
Entrez Gene ID
9057
Gene Name
solute carrier family 7 member 6
Gene Aliases
LAT-2, LAT3, y+LAT-2
UniGene
Hs.679580
Gene Chromosome Location
Chr.16: 68263830-68301823 on Build GRCh38
Transcripts
NM_001076785.2 NM_003983.5 XM_011523433.1 XM_011523434.1 XM_011523438.1 XM_011523439.1 XM_017023851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4843383 c.1116C>T p.F372F Chr.16: 68294798-68294798 on Build GRCh38
145950 c.1043G>A p.R348Q Chr.16: 68294725-68294725 on Build GRCh38
471984 c.1071C>A p.S357S Chr.16: 68294753-68294753 on Build GRCh38
3511206 c.1034T>A p.V345E Chr.16: 68294716-68294716 on Build GRCh38
6783536 c.1058C>T p.P353L Chr.16: 68294740-68294740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750428902 Chr.16: 68294535-68294535 on Build GRCh38
rs763619923 Chr.16: 68294811-68294811 on Build GRCh38
rs769826112 Chr.16: 68294740-68294740 on Build GRCh38
rs1013914237 Chr.16: 68294651-68294651 on Build GRCh38
rs747984243 Chr.16: 68294800-68294800 on Build GRCh38
rs778214762 Chr.16: 68294704-68294704 on Build GRCh38
rs932179974 Chr.16: 68294429-68294430 on Build GRCh38
rs774914854 Chr.16: 68294460-68294460 on Build GRCh38
rs1019940398 Chr.16: 68294653-68294653 on Build GRCh38
rs759136369 Chr.16: 68294795-68294795 on Build GRCh38
rs753775956 Chr.16: 68294590-68294590 on Build GRCh38
rs774298366 Chr.16: 68294741-68294741 on Build GRCh38
rs367828592 Chr.16: 68294810-68294810 on Build GRCh38
rs753259739 Chr.16: 68294808-68294808 on Build GRCh38
rs767818578 Chr.16: 68294517-68294517 on Build GRCh38
rs565944717 Chr.16: 68294839-68294839 on Build GRCh38
rs929334291 Chr.16: 68294401-68294401 on Build GRCh38
rs141038680 Chr.16: 68294451-68294451 on Build GRCh38
rs199805922 Chr.16: 68294752-68294752 on Build GRCh38
rs115489206 Chr.16: 68294825-68294825 on Build GRCh38
rs747393940 Chr.16: 68294714-68294714 on Build GRCh38
rs113380153 Chr.16: 68294833-68294833 on Build GRCh38
rs753206328 Chr.16: 68294671-68294671 on Build GRCh38
rs781456783 Chr.16: 68294729-68294729 on Build GRCh38
rs570812538 Chr.16: 68294513-68294513 on Build GRCh38
rs978976459 Chr.16: 68294689-68294689 on Build GRCh38
rs55710496 Chr.16: 68294536-68294536 on Build GRCh38
rs139848493 Chr.16: 68294781-68294781 on Build GRCh38
rs757480348 Chr.16: 68294715-68294715 on Build GRCh38
rs771861891 Chr.16: 68294763-68294763 on Build GRCh38
rs190478975 Chr.16: 68294391-68294391 on Build GRCh38
rs776302205 Chr.16: 68294792-68294792 on Build GRCh38
rs372173191 Chr.16: 68294664-68294664 on Build GRCh38
rs369041629 Chr.16: 68294711-68294711 on Build GRCh38
rs758847260 Chr.16: 68294701-68294701 on Build GRCh38
rs748025271 Chr.16: 68294758-68294758 on Build GRCh38
rs944697931 Chr.16: 68294501-68294501 on Build GRCh38
rs374875068 Chr.16: 68294673-68294673 on Build GRCh38
rs772950452 Chr.16: 68294764-68294764 on Build GRCh38
rs926083396 Chr.16: 68294429-68294429 on Build GRCh38
rs745975132 Chr.16: 68294730-68294730 on Build GRCh38
rs912019475 Chr.16: 68294453-68294453 on Build GRCh38
rs147952669 Chr.16: 68294611-68294611 on Build GRCh38
rs778662864 Chr.16: 68294724-68294724 on Build GRCh38
rs996994347 Chr.16: 68294571-68294571 on Build GRCh38
rs751919247 Chr.16: 68294822-68294822 on Build GRCh38
rs959849068 Chr.16: 68294612-68294612 on Build GRCh38
rs1047741338 Chr.16: 68294402-68294402 on Build GRCh38
rs770527629 Chr.16: 68294787-68294787 on Build GRCh38
rs372458911 Chr.16: 68294821-68294821 on Build GRCh38
rs571654600 Chr.16: 68294807-68294807 on Build GRCh38

View Full Product Details