Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7484633-7485137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2A
Entrez Gene ID
5430
Gene Name
RNA polymerase II subunit A
Gene Aliases
POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
UniGene
Hs.270017
Gene Chromosome Location
Chr.17: 7484379-7514618 on Build GRCh38
Transcripts
NM_000937.4

SNP Detail

SNP ID Chromosome Location
rs750846594 Chr.17: 7484886-7484886 on Build GRCh38
rs777199783 Chr.17: 7484778-7484779 on Build GRCh38
rs1024560256 Chr.17: 7484792-7484792 on Build GRCh38
rs1001765651 Chr.17: 7484811-7484811 on Build GRCh38
rs942247212 Chr.17: 7484920-7484920 on Build GRCh38
rs985529131 Chr.17: 7485110-7485110 on Build GRCh38
rs190864669 Chr.17: 7484878-7484878 on Build GRCh38
rs534069867 Chr.17: 7484670-7484670 on Build GRCh38
rs1037313117 Chr.17: 7484680-7484680 on Build GRCh38
rs986345264 Chr.17: 7485084-7485084 on Build GRCh38
rs1011426513 Chr.17: 7484994-7484994 on Build GRCh38
rs774332582 Chr.17: 7484716-7484716 on Build GRCh38
rs769665843 Chr.17: 7484780-7484780 on Build GRCh38
rs976155358 Chr.17: 7484882-7484882 on Build GRCh38
rs928690914 Chr.17: 7484675-7484675 on Build GRCh38
rs1001397987 Chr.17: 7484980-7484980 on Build GRCh38
rs573945524 Chr.17: 7484699-7484699 on Build GRCh38
rs751215469 Chr.17: 7484820-7484820 on Build GRCh38
rs942262042 Chr.17: 7485095-7485095 on Build GRCh38
rs375984351 Chr.17: 7484932-7484932 on Build GRCh38
rs534299498 Chr.17: 7485004-7485004 on Build GRCh38
rs575099034 Chr.17: 7484707-7484707 on Build GRCh38
rs761341697 Chr.17: 7485052-7485052 on Build GRCh38
rs566796481 Chr.17: 7484989-7484989 on Build GRCh38
rs768926310 Chr.17: 7484876-7484876 on Build GRCh38
rs754306483 Chr.17: 7484747-7484747 on Build GRCh38
rs981496620 Chr.17: 7484674-7484674 on Build GRCh38
rs561861244 Chr.17: 7484885-7484885 on Build GRCh38
rs765732394 Chr.17: 7484798-7484798 on Build GRCh38
rs764846272 Chr.17: 7484668-7484668 on Build GRCh38
rs1034131013 Chr.17: 7484981-7484981 on Build GRCh38
rs754205577 Chr.17: 7484787-7484787 on Build GRCh38
rs751162326 Chr.17: 7484906-7484906 on Build GRCh38
rs569146357 Chr.17: 7484946-7484946 on Build GRCh38
rs967372871 Chr.17: 7485011-7485011 on Build GRCh38
rs775338936 Chr.17: 7484677-7484677 on Build GRCh38
rs4796424 Chr.17: 7484654-7484654 on Build GRCh38
rs775645262 Chr.17: 7484785-7484785 on Build GRCh38
rs754661696 Chr.17: 7484907-7484907 on Build GRCh38
rs749274276 Chr.17: 7484860-7484860 on Build GRCh38
rs929722809 Chr.17: 7484958-7484958 on Build GRCh38
rs1011949951 Chr.17: 7485016-7485016 on Build GRCh38
rs994214645 Chr.17: 7485068-7485068 on Build GRCh38
rs971220369 Chr.17: 7484802-7484802 on Build GRCh38
rs750944351 Chr.17: 7484800-7484800 on Build GRCh38
rs759598292 Chr.17: 7484782-7484782 on Build GRCh38
rs986702845 Chr.17: 7484903-7484903 on Build GRCh38
rs972274784 Chr.17: 7485108-7485108 on Build GRCh38
rs533144452 Chr.17: 7484957-7484957 on Build GRCh38
rs893592432 Chr.17: 7484996-7484996 on Build GRCh38
rs769761900 Chr.17: 7484833-7484833 on Build GRCh38
rs1036130754 Chr.17: 7484914-7484914 on Build GRCh38
rs906943076 Chr.17: 7484968-7484968 on Build GRCh38
rs967455777 Chr.17: 7484869-7484869 on Build GRCh38
rs766440685 Chr.17: 7484715-7484715 on Build GRCh38
rs867781905 Chr.17: 7484799-7484799 on Build GRCh38
rs1044567431 Chr.17: 7485031-7485031 on Build GRCh38
rs1020247307 Chr.17: 7485003-7485003 on Build GRCh38
rs1053955543 Chr.17: 7484995-7484995 on Build GRCh38
rs760731040 Chr.17: 7484779-7484779 on Build GRCh38
rs757789292 Chr.17: 7484805-7484805 on Build GRCh38
rs545806291 Chr.17: 7484713-7484713 on Build GRCh38
rs765892388 Chr.17: 7484880-7484881 on Build GRCh38
rs927293386 Chr.17: 7484665-7484665 on Build GRCh38
rs566937158 Chr.17: 7484669-7484669 on Build GRCh38
rs772103266 Chr.17: 7484861-7484861 on Build GRCh38
rs369995545 Chr.17: 7484761-7484761 on Build GRCh38
rs931252683 Chr.17: 7484693-7484693 on Build GRCh38
rs756417344 Chr.17: 7484733-7484733 on Build GRCh38
rs768521150 Chr.17: 7484770-7484770 on Build GRCh38
rs757771437 Chr.17: 7484788-7484788 on Build GRCh38
rs555534999 Chr.17: 7484697-7484697 on Build GRCh38
rs757746668 Chr.17: 7484750-7484750 on Build GRCh38
rs976483487 Chr.17: 7485034-7485034 on Build GRCh38
rs1005262668 Chr.17: 7484777-7484777 on Build GRCh38
rs753052351 Chr.17: 7484721-7484721 on Build GRCh38
rs759779251 Chr.17: 7484717-7484717 on Build GRCh38
rs139712421 Chr.17: 7484706-7484706 on Build GRCh38
rs745428887 Chr.17: 7484758-7484758 on Build GRCh38
rs919988510 Chr.17: 7484941-7484941 on Build GRCh38
rs978026455 Chr.17: 7484664-7484664 on Build GRCh38
rs777501152 Chr.17: 7484840-7484840 on Build GRCh38
rs778465664 Chr.17: 7484754-7484754 on Build GRCh38
rs762376289 Chr.17: 7484779-7484779 on Build GRCh38
rs561641273 Chr.17: 7484763-7484763 on Build GRCh38
rs764468304 Chr.17: 7484746-7484746 on Build GRCh38
rs550849994 Chr.17: 7484901-7484901 on Build GRCh38
rs1035923361 Chr.17: 7485051-7485051 on Build GRCh38
rs567479938 Chr.17: 7485063-7485063 on Build GRCh38
rs953569775 Chr.17: 7485044-7485044 on Build GRCh38
rs182661622 Chr.17: 7484897-7484897 on Build GRCh38
rs1049644721 Chr.17: 7484698-7484698 on Build GRCh38
rs369511293 Chr.17: 7484678-7484678 on Build GRCh38
rs948697519 Chr.17: 7484672-7484672 on Build GRCh38
rs909351902 Chr.17: 7485085-7485085 on Build GRCh38
rs961679313 Chr.17: 7485054-7485054 on Build GRCh38
rs774472822 Chr.17: 7484781-7484781 on Build GRCh38
rs946120133 Chr.17: 7484703-7484703 on Build GRCh38
rs980002642 Chr.17: 7484656-7484656 on Build GRCh38
rs141361156 Chr.17: 7484827-7484827 on Build GRCh38
rs898312322 Chr.17: 7484691-7484691 on Build GRCh38
rs527350355 Chr.17: 7484964-7484964 on Build GRCh38
rs1040952901 Chr.17: 7484977-7484977 on Build GRCh38
rs952795720 Chr.17: 7485100-7485100 on Build GRCh38
rs553761464 Chr.17: 7484864-7484864 on Build GRCh38
rs767686288 Chr.17: 7484719-7484719 on Build GRCh38
rs764536293 Chr.17: 7484737-7484749 on Build GRCh38
rs973247410 Chr.17: 7484938-7484938 on Build GRCh38
rs893295621 Chr.17: 7484883-7484883 on Build GRCh38
rs746673121 Chr.17: 7484765-7484765 on Build GRCh38
rs373497066 Chr.17: 7484810-7484810 on Build GRCh38
rs915922538 Chr.17: 7484671-7484671 on Build GRCh38
rs112473664 Chr.17: 7484795-7484795 on Build GRCh38
rs776597121 Chr.17: 7484771-7484771 on Build GRCh38
rs186456788 Chr.17: 7485042-7485042 on Build GRCh38
rs138249788 Chr.17: 7484784-7484784 on Build GRCh38
rs913208975 Chr.17: 7484865-7484865 on Build GRCh38
rs544457148 Chr.17: 7484704-7484704 on Build GRCh38
rs762124073 Chr.17: 7484749-7484749 on Build GRCh38

View Full Product Details