Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7924054-7924549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB3
Entrez Gene ID
9196
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 3
Gene Aliases
AKR6A9, KCNA3.1B, KCNA3B, KV-BETA-3
UniGene
Hs.435074
Gene Chromosome Location
Chr.17: 7921859-7929435 on Build GRCh38
Transcripts
NM_004732.3 XM_011524068.1 XM_017025304.1 XM_017025305.1 XM_017025306.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6998745 c.721T>A p.S241T Chr.17: 7924256-7924256 on Build GRCh38
4773236 c.833-5C>A p.? Chr.17: 7924067-7924067 on Build GRCh38
6690528 c.667G>A p.A223T Chr.17: 7924459-7924459 on Build GRCh38
6196478 c.732delA p.R244fs*5 Chr.17: 7924245-7924245 on Build GRCh38
4749525 c.634C>T p.R212* Chr.17: 7924492-7924492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035664410 Chr.17: 7924152-7924152 on Build GRCh38
rs373812385 Chr.17: 7924183-7924183 on Build GRCh38
rs775769080 Chr.17: 7924307-7924307 on Build GRCh38
rs747865575 Chr.17: 7924439-7924439 on Build GRCh38
rs371028022 Chr.17: 7924400-7924400 on Build GRCh38
rs1021351066 Chr.17: 7924361-7924361 on Build GRCh38
rs553511494 Chr.17: 7924311-7924311 on Build GRCh38
rs750357232 Chr.17: 7924379-7924379 on Build GRCh38
rs774817744 Chr.17: 7924102-7924102 on Build GRCh38
rs572025169 Chr.17: 7924389-7924389 on Build GRCh38
rs765330958 Chr.17: 7924499-7924499 on Build GRCh38
rs779768586 Chr.17: 7924263-7924263 on Build GRCh38
rs753762064 Chr.17: 7924155-7924155 on Build GRCh38
rs997344529 Chr.17: 7924242-7924242 on Build GRCh38
rs376678079 Chr.17: 7924505-7924505 on Build GRCh38
rs757234706 Chr.17: 7924174-7924174 on Build GRCh38
rs751428211 Chr.17: 7924397-7924397 on Build GRCh38
rs752088911 Chr.17: 7924135-7924135 on Build GRCh38
rs746364868 Chr.17: 7924417-7924417 on Build GRCh38
rs746997800 Chr.17: 7924415-7924415 on Build GRCh38
rs780427394 Chr.17: 7924425-7924425 on Build GRCh38
rs746429614 Chr.17: 7924294-7924294 on Build GRCh38
rs765247026 Chr.17: 7924244-7924244 on Build GRCh38
rs758893161 Chr.17: 7924500-7924500 on Build GRCh38
rs772471642 Chr.17: 7924295-7924295 on Build GRCh38
rs778918865 Chr.17: 7924410-7924410 on Build GRCh38
rs778153217 Chr.17: 7924402-7924402 on Build GRCh38
rs143227345 Chr.17: 7924445-7924445 on Build GRCh38
rs753388080 Chr.17: 7924233-7924233 on Build GRCh38
rs780554724 Chr.17: 7924503-7924503 on Build GRCh38
rs1025440535 Chr.17: 7924433-7924433 on Build GRCh38
rs763662504 Chr.17: 7924149-7924149 on Build GRCh38
rs766824648 Chr.17: 7924089-7924089 on Build GRCh38
rs117731997 Chr.17: 7924237-7924237 on Build GRCh38
rs199631499 Chr.17: 7924181-7924181 on Build GRCh38
rs771556030 Chr.17: 7924221-7924221 on Build GRCh38
rs371618530 Chr.17: 7924438-7924438 on Build GRCh38
rs769978059 Chr.17: 7924205-7924205 on Build GRCh38
rs763609969 Chr.17: 7924088-7924088 on Build GRCh38
rs749399173 Chr.17: 7924279-7924279 on Build GRCh38
rs778909909 Chr.17: 7924428-7924429 on Build GRCh38
rs747399495 Chr.17: 7924429-7924429 on Build GRCh38
rs755627791 Chr.17: 7924147-7924147 on Build GRCh38
rs979728882 Chr.17: 7924399-7924399 on Build GRCh38
rs372782303 Chr.17: 7924488-7924488 on Build GRCh38
rs755336061 Chr.17: 7924527-7924527 on Build GRCh38
rs753902409 Chr.17: 7924404-7924404 on Build GRCh38
rs553249244 Chr.17: 7924222-7924222 on Build GRCh38
rs1048292728 Chr.17: 7924479-7924479 on Build GRCh38
rs752939665 Chr.17: 7924266-7924266 on Build GRCh38
rs763573059 Chr.17: 7924232-7924232 on Build GRCh38
rs542357066 Chr.17: 7924448-7924448 on Build GRCh38
rs981849977 Chr.17: 7924103-7924103 on Build GRCh38
rs775205848 Chr.17: 7924223-7924223 on Build GRCh38
rs770369504 Chr.17: 7924080-7924080 on Build GRCh38
rs765021670 Chr.17: 7924378-7924378 on Build GRCh38
rs781493717 Chr.17: 7924191-7924191 on Build GRCh38
rs769321398 Chr.17: 7924440-7924440 on Build GRCh38
rs1056605213 Chr.17: 7924113-7924113 on Build GRCh38
rs764359053 Chr.17: 7924226-7924227 on Build GRCh38
rs777958416 Chr.17: 7924278-7924278 on Build GRCh38
rs35825894 Chr.17: 7924088-7924089 on Build GRCh38
rs959229040 Chr.17: 7924200-7924200 on Build GRCh38
rs779479251 Chr.17: 7924289-7924289 on Build GRCh38
rs759957113 Chr.17: 7924219-7924219 on Build GRCh38
rs774228071 Chr.17: 7924460-7924460 on Build GRCh38
rs749778107 Chr.17: 7924210-7924210 on Build GRCh38
rs757116777 Chr.17: 7924375-7924375 on Build GRCh38
rs145774275 Chr.17: 7924418-7924418 on Build GRCh38
rs760379968 Chr.17: 7924229-7924229 on Build GRCh38
rs767486615 Chr.17: 7924491-7924491 on Build GRCh38
rs368626433 Chr.17: 7924193-7924193 on Build GRCh38
rs371141502 Chr.17: 7924209-7924209 on Build GRCh38
rs778650825 Chr.17: 7924180-7924180 on Build GRCh38
rs370095308 Chr.17: 7924123-7924123 on Build GRCh38
rs991887174 Chr.17: 7924466-7924466 on Build GRCh38
rs762843750 Chr.17: 7924387-7924387 on Build GRCh38
rs556025090 Chr.17: 7924218-7924218 on Build GRCh38
rs368190219 Chr.17: 7924257-7924257 on Build GRCh38
rs745672481 Chr.17: 7924429-7924429 on Build GRCh38
rs1021867975 Chr.17: 7924482-7924482 on Build GRCh38
rs766247391 Chr.17: 7924453-7924453 on Build GRCh38
rs771507651 Chr.17: 7924285-7924285 on Build GRCh38
rs754099814 Chr.17: 7924492-7924492 on Build GRCh38
rs567178691 Chr.17: 7924107-7924107 on Build GRCh38
rs34360617 Chr.17: 7924498-7924498 on Build GRCh38
rs756440646 Chr.17: 7924277-7924277 on Build GRCh38
rs113982727 Chr.17: 7924097-7924097 on Build GRCh38
rs1017484081 Chr.17: 7924226-7924226 on Build GRCh38
rs754157041 Chr.17: 7924273-7924274 on Build GRCh38
rs759409428 Chr.17: 7924487-7924487 on Build GRCh38
rs761330286 Chr.17: 7924368-7924368 on Build GRCh38
rs111968052 Chr.17: 7924437-7924437 on Build GRCh38
rs988948514 Chr.17: 7924336-7924336 on Build GRCh38
rs757352314 Chr.17: 7924405-7924405 on Build GRCh38
rs758221210 Chr.17: 7924259-7924259 on Build GRCh38
rs372067112 Chr.17: 7924310-7924310 on Build GRCh38

View Full Product Details