Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8828005-8828495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R6
Entrez Gene ID
146850
Gene Name
phosphoinositide-3-kinase regulatory subunit 6
Gene Aliases
C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP
UniGene
Hs.255809
Gene Chromosome Location
Chr.17: 8802723-8870003 on Build GRCh38
Transcripts
NM_001010855.3 NM_001290211.1 NR_110865.1 XM_011523677.2 XM_011523678.2 XM_011523680.2 XM_011523681.2 XM_011523683.2 XM_011523686.2 XM_017024228.1 XM_017024229.1 XM_017024230.1 XM_017024231.1

SNP Detail

SNP ID Chromosome Location
rs370275678 Chr.17: 8828084-8828084 on Build GRCh38
rs748223491 Chr.17: 8828341-8828341 on Build GRCh38
rs764260800 Chr.17: 8828210-8828210 on Build GRCh38
rs550947334 Chr.17: 8828430-8828430 on Build GRCh38
rs762897524 Chr.17: 8828178-8828178 on Build GRCh38
rs752956813 Chr.17: 8828122-8828122 on Build GRCh38
rs376771141 Chr.17: 8828144-8828144 on Build GRCh38
rs1057330203 Chr.17: 8828278-8828278 on Build GRCh38
rs904471050 Chr.17: 8828400-8828400 on Build GRCh38
rs767916377 Chr.17: 8828114-8828114 on Build GRCh38
rs531891787 Chr.17: 8828395-8828395 on Build GRCh38
rs768516686 Chr.17: 8828431-8828431 on Build GRCh38
rs578103057 Chr.17: 8828203-8828203 on Build GRCh38
rs376286933 Chr.17: 8828290-8828290 on Build GRCh38
rs775095103 Chr.17: 8828070-8828070 on Build GRCh38
rs779466054 Chr.17: 8828224-8828224 on Build GRCh38
rs975669906 Chr.17: 8828199-8828199 on Build GRCh38
rs1010849222 Chr.17: 8828413-8828413 on Build GRCh38
rs780352365 Chr.17: 8828229-8828229 on Build GRCh38
rs903858081 Chr.17: 8828045-8828045 on Build GRCh38
rs923166643 Chr.17: 8828031-8828031 on Build GRCh38
rs573753611 Chr.17: 8828120-8828120 on Build GRCh38
rs776631238 Chr.17: 8828091-8828091 on Build GRCh38
rs777648571 Chr.17: 8828129-8828129 on Build GRCh38
rs762809880 Chr.17: 8828113-8828113 on Build GRCh38
rs906707290 Chr.17: 8828385-8828385 on Build GRCh38
rs774471857 Chr.17: 8828276-8828276 on Build GRCh38
rs199556046 Chr.17: 8828094-8828094 on Build GRCh38
rs540455106 Chr.17: 8828050-8828050 on Build GRCh38
rs1031482686 Chr.17: 8828105-8828105 on Build GRCh38
rs779693308 Chr.17: 8828155-8828155 on Build GRCh38
rs994086849 Chr.17: 8828127-8828127 on Build GRCh38
rs796881138 Chr.17: 8828393-8828395 on Build GRCh38
rs768240219 Chr.17: 8828163-8828163 on Build GRCh38
rs757299790 Chr.17: 8828222-8828222 on Build GRCh38
rs938464559 Chr.17: 8828249-8828249 on Build GRCh38
rs964249392 Chr.17: 8828194-8828194 on Build GRCh38
rs915755347 Chr.17: 8828302-8828302 on Build GRCh38
rs373060073 Chr.17: 8828226-8828226 on Build GRCh38
rs143782173 Chr.17: 8828393-8828393 on Build GRCh38
rs888818474 Chr.17: 8828081-8828081 on Build GRCh38
rs757660738 Chr.17: 8828137-8828137 on Build GRCh38
rs1030545960 Chr.17: 8828451-8828451 on Build GRCh38
rs1042953877 Chr.17: 8828366-8828366 on Build GRCh38
rs964198956 Chr.17: 8828455-8828455 on Build GRCh38
rs754400988 Chr.17: 8828136-8828136 on Build GRCh38
rs918990657 Chr.17: 8828305-8828305 on Build GRCh38
rs549623642 Chr.17: 8828260-8828260 on Build GRCh38
rs73973230 Chr.17: 8828424-8828424 on Build GRCh38
rs561323992 Chr.17: 8828394-8828394 on Build GRCh38
rs956839459 Chr.17: 8828124-8828124 on Build GRCh38
rs769892050 Chr.17: 8828176-8828176 on Build GRCh38
rs374913677 Chr.17: 8828200-8828200 on Build GRCh38
rs527989281 Chr.17: 8828250-8828250 on Build GRCh38
rs1050086455 Chr.17: 8828042-8828042 on Build GRCh38
rs760288236 Chr.17: 8828072-8828072 on Build GRCh38
rs1045012265 Chr.17: 8828039-8828039 on Build GRCh38
rs12949852 Chr.17: 8828428-8828428 on Build GRCh38
rs59253413 Chr.17: 8828215-8828215 on Build GRCh38
rs544724196 Chr.17: 8828140-8828140 on Build GRCh38
rs374771571 Chr.17: 8828227-8828227 on Build GRCh38
rs948591804 Chr.17: 8828329-8828329 on Build GRCh38
rs746693980 Chr.17: 8828230-8828230 on Build GRCh38
rs139107090 Chr.17: 8828024-8828024 on Build GRCh38
rs166435 Chr.17: 8828169-8828169 on Build GRCh38
rs368266052 Chr.17: 8828154-8828154 on Build GRCh38
rs575690601 Chr.17: 8828214-8828214 on Build GRCh38
rs765145493 Chr.17: 8828096-8828096 on Build GRCh38
rs201665615 Chr.17: 8828180-8828180 on Build GRCh38
rs867403544 Chr.17: 8828185-8828185 on Build GRCh38
rs756232603 Chr.17: 8828126-8828126 on Build GRCh38
rs762407425 Chr.17: 8828221-8828221 on Build GRCh38
rs566428797 Chr.17: 8828439-8828439 on Build GRCh38
rs550300824 Chr.17: 8828401-8828401 on Build GRCh38
rs750291229 Chr.17: 8828100-8828100 on Build GRCh38
rs562931068 Chr.17: 8828156-8828156 on Build GRCh38

View Full Product Details