Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 33015867-33016374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC2
Entrez Gene ID
40
Gene Name
acid sensing ion channel subunit 2
Gene Aliases
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1
UniGene
Hs.368417
Gene Chromosome Location
Chr.17: 33013087-34156806 on Build GRCh38
Transcripts
NM_001094.4 NM_183377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4065448 c.1422C>T p.S474S Chr.17: 33015986-33015986 on Build GRCh38
3516022 c.1411G>A p.D471N Chr.17: 33015997-33015997 on Build GRCh38
5587109 c.1576C>T p.H526Y Chr.17: 33015985-33015985 on Build GRCh38
4065447 c.1575C>T p.S525S Chr.17: 33015986-33015986 on Build GRCh38
6365183 c.1417G>T p.G473W Chr.17: 33015991-33015991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748308827 Chr.17: 33016026-33016026 on Build GRCh38
rs902175396 Chr.17: 33015991-33015991 on Build GRCh38
rs368009672 Chr.17: 33015920-33015920 on Build GRCh38
rs1016367517 Chr.17: 33016110-33016110 on Build GRCh38
rs781384406 Chr.17: 33015923-33015923 on Build GRCh38
rs563229762 Chr.17: 33016223-33016223 on Build GRCh38
rs577780157 Chr.17: 33016220-33016220 on Build GRCh38
rs543881385 Chr.17: 33015917-33015917 on Build GRCh38
rs751122738 Chr.17: 33015972-33015972 on Build GRCh38
rs780233002 Chr.17: 33016060-33016060 on Build GRCh38
rs1042050278 Chr.17: 33015973-33015973 on Build GRCh38
rs780407455 Chr.17: 33015997-33015997 on Build GRCh38
rs974235757 Chr.17: 33016172-33016172 on Build GRCh38
rs755208807 Chr.17: 33016156-33016157 on Build GRCh38
rs961112865 Chr.17: 33016158-33016158 on Build GRCh38
rs945453427 Chr.17: 33016299-33016299 on Build GRCh38
rs904005812 Chr.17: 33016308-33016308 on Build GRCh38
rs755819913 Chr.17: 33016090-33016090 on Build GRCh38
rs766937728 Chr.17: 33016075-33016075 on Build GRCh38
rs371904950 Chr.17: 33016086-33016086 on Build GRCh38
rs144775533 Chr.17: 33016218-33016218 on Build GRCh38
rs775495791 Chr.17: 33016055-33016055 on Build GRCh38
rs975335581 Chr.17: 33016199-33016199 on Build GRCh38
rs778825062 Chr.17: 33016010-33016010 on Build GRCh38
rs770163274 Chr.17: 33015926-33015926 on Build GRCh38
rs960381080 Chr.17: 33015918-33015918 on Build GRCh38
rs752625870 Chr.17: 33015960-33015960 on Build GRCh38
rs763043983 Chr.17: 33015965-33015965 on Build GRCh38
rs374194349 Chr.17: 33016072-33016072 on Build GRCh38
rs994082887 Chr.17: 33016087-33016087 on Build GRCh38
rs1033956735 Chr.17: 33016117-33016117 on Build GRCh38
rs763695729 Chr.17: 33015966-33015966 on Build GRCh38
rs3025251 Chr.17: 33016157-33016157 on Build GRCh38
rs747247725 Chr.17: 33016042-33016042 on Build GRCh38
rs746018164 Chr.17: 33015936-33015936 on Build GRCh38
rs749779467 Chr.17: 33016001-33016001 on Build GRCh38
rs766724049 Chr.17: 33016091-33016091 on Build GRCh38
rs28929 Chr.17: 33015931-33015931 on Build GRCh38
rs878950784 Chr.17: 33016099-33016099 on Build GRCh38
rs537068900 Chr.17: 33015949-33015949 on Build GRCh38
rs757840701 Chr.17: 33015923-33015923 on Build GRCh38
rs756841726 Chr.17: 33015982-33015982 on Build GRCh38
rs758542088 Chr.17: 33016059-33016059 on Build GRCh38
rs944701546 Chr.17: 33016261-33016261 on Build GRCh38
rs776266524 Chr.17: 33015952-33015952 on Build GRCh38
rs750032103 Chr.17: 33016081-33016081 on Build GRCh38
rs543452925 Chr.17: 33016221-33016221 on Build GRCh38
rs762954958 Chr.17: 33016067-33016067 on Build GRCh38
rs146766167 Chr.17: 33016277-33016277 on Build GRCh38
rs769700834 Chr.17: 33016045-33016045 on Build GRCh38
rs1002466260 Chr.17: 33015895-33015895 on Build GRCh38
rs773417540 Chr.17: 33016036-33016036 on Build GRCh38
rs545253585 Chr.17: 33016175-33016175 on Build GRCh38
rs200924687 Chr.17: 33015983-33015983 on Build GRCh38
rs112529854 Chr.17: 33015924-33015924 on Build GRCh38
rs141054977 Chr.17: 33015957-33015957 on Build GRCh38
rs960348605 Chr.17: 33016122-33016122 on Build GRCh38
rs1044218263 Chr.17: 33016307-33016307 on Build GRCh38
rs761934426 Chr.17: 33015944-33015944 on Build GRCh38
rs768662417 Chr.17: 33015934-33015934 on Build GRCh38
rs536901099 Chr.17: 33016002-33016002 on Build GRCh38
rs1013294637 Chr.17: 33016148-33016148 on Build GRCh38
rs909422013 Chr.17: 33015894-33015894 on Build GRCh38
rs368500636 Chr.17: 33015893-33015893 on Build GRCh38
rs551885482 Chr.17: 33015929-33015929 on Build GRCh38
rs984909947 Chr.17: 33016101-33016101 on Build GRCh38
rs3025250 Chr.17: 33016327-33016327 on Build GRCh38
rs923522820 Chr.17: 33015925-33015925 on Build GRCh38
rs919739291 Chr.17: 33016174-33016174 on Build GRCh38
rs772066732 Chr.17: 33016028-33016028 on Build GRCh38
rs572875747 Chr.17: 33015896-33015896 on Build GRCh38
rs761343086 Chr.17: 33016073-33016073 on Build GRCh38
rs757004100 Chr.17: 33015921-33015921 on Build GRCh38
rs750071243 Chr.17: 33015994-33015994 on Build GRCh38
rs923643738 Chr.17: 33016311-33016311 on Build GRCh38
rs28928 Chr.17: 33016351-33016351 on Build GRCh38
rs147559460 Chr.17: 33015995-33015995 on Build GRCh38
rs528893501 Chr.17: 33016273-33016273 on Build GRCh38
rs1033318302 Chr.17: 33015899-33015899 on Build GRCh38
rs1002433805 Chr.17: 33016109-33016109 on Build GRCh38

View Full Product Details