Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 33111835-33112290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC2
Entrez Gene ID
40
Gene Name
acid sensing ion channel subunit 2
Gene Aliases
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1
UniGene
Hs.368417
Gene Chromosome Location
Chr.17: 33013087-34156806 on Build GRCh38
Transcripts
NM_001094.4 NM_183377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752123 c.706G>T p.E236* Chr.17: 33111917-33111917 on Build GRCh38
6556287 c.627C>A p.V209V Chr.17: 33111996-33111996 on Build GRCh38
6605743 c.597C>A p.G199G Chr.17: 33112026-33112026 on Build GRCh38
3516035 c.849G>A p.W283* Chr.17: 33111927-33111927 on Build GRCh38
5966177 c.623C>A p.T208K Chr.17: 33112000-33112000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145139925 Chr.17: 33111939-33111939 on Build GRCh38
rs1022531793 Chr.17: 33112217-33112217 on Build GRCh38
rs898436479 Chr.17: 33112091-33112091 on Build GRCh38
rs539796708 Chr.17: 33112173-33112173 on Build GRCh38
rs1038304211 Chr.17: 33112053-33112053 on Build GRCh38
rs150196165 Chr.17: 33112117-33112117 on Build GRCh38
rs754675358 Chr.17: 33112018-33112018 on Build GRCh38
rs764324242 Chr.17: 33112035-33112035 on Build GRCh38
rs778651350 Chr.17: 33111959-33111959 on Build GRCh38
rs9916605 Chr.17: 33111887-33111887 on Build GRCh38
rs926373354 Chr.17: 33112000-33112000 on Build GRCh38
rs770538158 Chr.17: 33111982-33111982 on Build GRCh38
rs147153560 Chr.17: 33111990-33111990 on Build GRCh38
rs994502709 Chr.17: 33112126-33112126 on Build GRCh38
rs774139629 Chr.17: 33112014-33112014 on Build GRCh38
rs1010203553 Chr.17: 33112214-33112214 on Build GRCh38
rs758373199 Chr.17: 33112039-33112039 on Build GRCh38
rs554297751 Chr.17: 33112154-33112154 on Build GRCh38
rs759123215 Chr.17: 33112191-33112191 on Build GRCh38
rs534080116 Chr.17: 33112142-33112142 on Build GRCh38
rs201936906 Chr.17: 33112026-33112026 on Build GRCh38
rs748092997 Chr.17: 33111972-33111972 on Build GRCh38
rs112516665 Chr.17: 33111867-33111867 on Build GRCh38
rs556916303 Chr.17: 33112174-33112174 on Build GRCh38
rs34411967 Chr.17: 33112145-33112146 on Build GRCh38
rs770479044 Chr.17: 33112047-33112047 on Build GRCh38
rs926818437 Chr.17: 33112241-33112241 on Build GRCh38
rs139598307 Chr.17: 33111936-33111936 on Build GRCh38
rs760379981 Chr.17: 33111879-33111879 on Build GRCh38
rs200088098 Chr.17: 33111929-33111929 on Build GRCh38
rs772776813 Chr.17: 33111977-33111977 on Build GRCh38
rs771559356 Chr.17: 33111974-33111974 on Build GRCh38
rs1008942444 Chr.17: 33112198-33112198 on Build GRCh38
rs778055319 Chr.17: 33111926-33111926 on Build GRCh38
rs140895516 Chr.17: 33112080-33112081 on Build GRCh38
rs753699472 Chr.17: 33112079-33112080 on Build GRCh38
rs765622116 Chr.17: 33111897-33111897 on Build GRCh38
rs761276993 Chr.17: 33111866-33111866 on Build GRCh38
rs778829266 Chr.17: 33112025-33112025 on Build GRCh38
rs995303066 Chr.17: 33111856-33111856 on Build GRCh38
rs559883232 Chr.17: 33112135-33112135 on Build GRCh38
rs1018860658 Chr.17: 33112199-33112199 on Build GRCh38
rs777243834 Chr.17: 33112040-33112040 on Build GRCh38
rs980960404 Chr.17: 33112227-33112227 on Build GRCh38
rs770761664 Chr.17: 33111994-33111994 on Build GRCh38
rs796207003 Chr.17: 33112079-33112080 on Build GRCh38
rs968256135 Chr.17: 33111993-33111993 on Build GRCh38
rs549346772 Chr.17: 33112266-33112266 on Build GRCh38
rs199552243 Chr.17: 33111971-33111971 on Build GRCh38
rs776377083 Chr.17: 33111983-33111983 on Build GRCh38
rs148618079 Chr.17: 33111999-33111999 on Build GRCh38
rs207476368 Chr.17: 33111933-33111933 on Build GRCh38
rs368559997 Chr.17: 33111904-33111904 on Build GRCh38
rs956721891 Chr.17: 33112208-33112208 on Build GRCh38
rs568225586 Chr.17: 33112136-33112136 on Build GRCh38
rs758492772 Chr.17: 33111923-33111923 on Build GRCh38
rs751735036 Chr.17: 33111992-33111992 on Build GRCh38
rs776089839 Chr.17: 33112062-33112062 on Build GRCh38
rs764642591 Chr.17: 33112118-33112118 on Build GRCh38
rs770048021 Chr.17: 33112083-33112083 on Build GRCh38
rs375105736 Chr.17: 33111991-33111991 on Build GRCh38
rs750370890 Chr.17: 33112080-33112083 on Build GRCh38
rs745590349 Chr.17: 33112079-33112079 on Build GRCh38
rs112039708 Chr.17: 33112115-33112115 on Build GRCh38
rs1036007071 Chr.17: 33112087-33112087 on Build GRCh38
rs577318545 Chr.17: 33112170-33112170 on Build GRCh38
rs771437360 Chr.17: 33112220-33112220 on Build GRCh38
rs748958495 Chr.17: 33111978-33111978 on Build GRCh38
rs750666212 Chr.17: 33112017-33112017 on Build GRCh38
rs746474013 Chr.17: 33112046-33112046 on Build GRCh38
rs371727767 Chr.17: 33112146-33112146 on Build GRCh38
rs375913197 Chr.17: 33111896-33111896 on Build GRCh38
rs371777452 Chr.17: 33111919-33111919 on Build GRCh38

View Full Product Details