Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39213179-39213672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAC2
Entrez Gene ID
342667
Gene Name
SH3 and cysteine rich domain 2
Gene Aliases
24b2, 24b2/STAC2
UniGene
Hs.145068
Gene Chromosome Location
Chr.17: 39210536-39225787 on Build GRCh38
Transcripts
NM_198993.3 XM_017024580.1 XM_017024581.1 XM_017024582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3516761 c.944C>T p.P315L Chr.17: 39213556-39213556 on Build GRCh38
5386203 c.974C>T p.S325F Chr.17: 39213526-39213526 on Build GRCh38
5852380 c.947G>A p.G316E Chr.17: 39213553-39213553 on Build GRCh38
5386204 c.942G>A p.Q314Q Chr.17: 39213558-39213558 on Build GRCh38
7146793 c.949G>A p.D317N Chr.17: 39213551-39213551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543637223 Chr.17: 39213255-39213255 on Build GRCh38
rs578061600 Chr.17: 39213521-39213521 on Build GRCh38
rs554763439 Chr.17: 39213207-39213207 on Build GRCh38
rs372743371 Chr.17: 39213444-39213444 on Build GRCh38
rs749577616 Chr.17: 39213486-39213486 on Build GRCh38
rs907295453 Chr.17: 39213619-39213619 on Build GRCh38
rs541440597 Chr.17: 39213431-39213431 on Build GRCh38
rs577583712 Chr.17: 39213269-39213269 on Build GRCh38
rs904295460 Chr.17: 39213244-39213244 on Build GRCh38
rs545035226 Chr.17: 39213316-39213316 on Build GRCh38
rs887751073 Chr.17: 39213473-39213473 on Build GRCh38
rs963156930 Chr.17: 39213447-39213447 on Build GRCh38
rs761716743 Chr.17: 39213540-39213540 on Build GRCh38
rs16550 Chr.17: 39213326-39213326 on Build GRCh38
rs148047874 Chr.17: 39213514-39213514 on Build GRCh38
rs887843994 Chr.17: 39213256-39213256 on Build GRCh38
rs542750577 Chr.17: 39213343-39213343 on Build GRCh38
rs770870439 Chr.17: 39213522-39213522 on Build GRCh38
rs774481712 Chr.17: 39213501-39213501 on Build GRCh38
rs1036366610 Chr.17: 39213576-39213576 on Build GRCh38
rs944833599 Chr.17: 39213449-39213449 on Build GRCh38
rs576617095 Chr.17: 39213241-39213241 on Build GRCh38
rs375776325 Chr.17: 39213433-39213433 on Build GRCh38
rs16549 Chr.17: 39213321-39213321 on Build GRCh38
rs959534852 Chr.17: 39213633-39213633 on Build GRCh38
rs1029811683 Chr.17: 39213372-39213372 on Build GRCh38
rs775591339 Chr.17: 39213278-39213278 on Build GRCh38
rs767943958 Chr.17: 39213580-39213580 on Build GRCh38
rs559318774 Chr.17: 39213264-39213264 on Build GRCh38
rs972698290 Chr.17: 39213451-39213451 on Build GRCh38
rs376071211 Chr.17: 39213598-39213598 on Build GRCh38
rs111677226 Chr.17: 39213461-39213461 on Build GRCh38
rs1040988402 Chr.17: 39213206-39213206 on Build GRCh38
rs776732276 Chr.17: 39213347-39213347 on Build GRCh38
rs999861992 Chr.17: 39213304-39213304 on Build GRCh38
rs1034901982 Chr.17: 39213631-39213631 on Build GRCh38
rs377144142 Chr.17: 39213459-39213459 on Build GRCh38
rs990891554 Chr.17: 39213378-39213378 on Build GRCh38
rs756767397 Chr.17: 39213563-39213563 on Build GRCh38
rs748160225 Chr.17: 39213502-39213502 on Build GRCh38
rs376962733 Chr.17: 39213470-39213470 on Build GRCh38
rs996572599 Chr.17: 39213217-39213217 on Build GRCh38
rs1035317545 Chr.17: 39213333-39213333 on Build GRCh38
rs765188977 Chr.17: 39213555-39213555 on Build GRCh38
rs368895687 Chr.17: 39213452-39213452 on Build GRCh38
rs763831001 Chr.17: 39213552-39213552 on Build GRCh38
rs776584189 Chr.17: 39213531-39213531 on Build GRCh38
rs140834210 Chr.17: 39213548-39213548 on Build GRCh38
rs560807591 Chr.17: 39213402-39213402 on Build GRCh38
rs959354506 Chr.17: 39213362-39213362 on Build GRCh38
rs765120213 Chr.17: 39213547-39213547 on Build GRCh38
rs1052731724 Chr.17: 39213532-39213532 on Build GRCh38
rs772271970 Chr.17: 39213268-39213268 on Build GRCh38
rs780529691 Chr.17: 39213605-39213605 on Build GRCh38
rs762747972 Chr.17: 39213551-39213551 on Build GRCh38
rs914935584 Chr.17: 39213407-39213407 on Build GRCh38
rs1004859038 Chr.17: 39213543-39213543 on Build GRCh38
rs761722716 Chr.17: 39213525-39213526 on Build GRCh38
rs923685812 Chr.17: 39213448-39213448 on Build GRCh38
rs532234917 Chr.17: 39213627-39213627 on Build GRCh38
rs1040581448 Chr.17: 39213450-39213450 on Build GRCh38
rs759279470 Chr.17: 39213534-39213534 on Build GRCh38
rs1014976442 Chr.17: 39213286-39213286 on Build GRCh38
rs371416496 Chr.17: 39213526-39213526 on Build GRCh38
rs58545654 Chr.17: 39213451-39213451 on Build GRCh38
rs768856766 Chr.17: 39213492-39213492 on Build GRCh38
rs564518873 Chr.17: 39213453-39213453 on Build GRCh38
rs780253162 Chr.17: 39213475-39213475 on Build GRCh38

View Full Product Details