Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 70179357-70179867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ2
Entrez Gene ID
3759
Gene Name
potassium voltage-gated channel subfamily J member 2
Gene Aliases
ATFB9, HHBIRK1, HHIRK1, IRK1, KIR2.1, LQT7, SQT3
UniGene
Hs.1547
Gene Chromosome Location
Chr.17: 70169535-70180044 on Build GRCh38
Transcripts
NM_000891.2

SNP Detail

SNP ID Chromosome Location
rs1054888338 Chr.17: 70179842-70179842 on Build GRCh38
rs3841509 Chr.17: 70179786-70179786 on Build GRCh38
rs1036695738 Chr.17: 70179507-70179507 on Build GRCh38
rs976616212 Chr.17: 70179678-70179678 on Build GRCh38
rs546321373 Chr.17: 70179758-70179758 on Build GRCh38
rs3815306 Chr.17: 70179690-70179690 on Build GRCh38
rs886053345 Chr.17: 70179675-70179675 on Build GRCh38
rs1011120068 Chr.17: 70179642-70179642 on Build GRCh38
rs1029699049 Chr.17: 70179689-70179689 on Build GRCh38
rs886053347 Chr.17: 70179715-70179716 on Build GRCh38
rs3815307 Chr.17: 70179819-70179819 on Build GRCh38
rs1055258300 Chr.17: 70179615-70179615 on Build GRCh38
rs564383347 Chr.17: 70179772-70179772 on Build GRCh38
rs967069685 Chr.17: 70179675-70179675 on Build GRCh38
rs958362058 Chr.17: 70179643-70179643 on Build GRCh38
rs373948055 Chr.17: 70179409-70179409 on Build GRCh38
rs1013946089 Chr.17: 70179792-70179792 on Build GRCh38
rs531038680 Chr.17: 70179582-70179582 on Build GRCh38
rs1025243571 Chr.17: 70179571-70179571 on Build GRCh38
rs542519856 Chr.17: 70179595-70179595 on Build GRCh38
rs911259013 Chr.17: 70179760-70179760 on Build GRCh38
rs911408055 Chr.17: 70179463-70179463 on Build GRCh38
rs943920928 Chr.17: 70179398-70179398 on Build GRCh38
rs557623909 Chr.17: 70179661-70179661 on Build GRCh38
rs551384883 Chr.17: 70179732-70179732 on Build GRCh38
rs750672814 Chr.17: 70179619-70179622 on Build GRCh38
rs955330682 Chr.17: 70179700-70179700 on Build GRCh38
rs562088857 Chr.17: 70179826-70179826 on Build GRCh38
rs930225865 Chr.17: 70179542-70179542 on Build GRCh38
rs188533495 Chr.17: 70179531-70179531 on Build GRCh38
rs918743809 Chr.17: 70179777-70179777 on Build GRCh38
rs149224363 Chr.17: 70179486-70179486 on Build GRCh38
rs1022123230 Chr.17: 70179674-70179674 on Build GRCh38
rs1036790167 Chr.17: 70179777-70179777 on Build GRCh38
rs552636156 Chr.17: 70179711-70179712 on Build GRCh38
rs939725429 Chr.17: 70179768-70179768 on Build GRCh38
rs909755476 Chr.17: 70179735-70179735 on Build GRCh38
rs969519474 Chr.17: 70179580-70179580 on Build GRCh38
rs886053346 Chr.17: 70179676-70179676 on Build GRCh38
rs927649372 Chr.17: 70179817-70179817 on Build GRCh38
rs999427890 Chr.17: 70179676-70179676 on Build GRCh38
rs553647437 Chr.17: 70179503-70179503 on Build GRCh38
rs540233396 Chr.17: 70179822-70179822 on Build GRCh38
rs1040949412 Chr.17: 70179425-70179425 on Build GRCh38
rs895486955 Chr.17: 70179509-70179509 on Build GRCh38
rs1004000664 Chr.17: 70179505-70179505 on Build GRCh38
rs974685175 Chr.17: 70179770-70179770 on Build GRCh38
rs1045933133 Chr.17: 70179800-70179800 on Build GRCh38
rs1047408718 Chr.17: 70179558-70179558 on Build GRCh38
rs570896638 Chr.17: 70179783-70179783 on Build GRCh38
rs1002762249 Chr.17: 70179584-70179584 on Build GRCh38
rs962918722 Chr.17: 70179762-70179762 on Build GRCh38
rs894978518 Chr.17: 70179620-70179620 on Build GRCh38
rs921069066 Chr.17: 70179481-70179481 on Build GRCh38
rs988247605 Chr.17: 70179662-70179662 on Build GRCh38
rs184562002 Chr.17: 70179500-70179500 on Build GRCh38
rs906194331 Chr.17: 70179588-70179588 on Build GRCh38

View Full Product Details