Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75266366-75266907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIF4GD
Entrez Gene ID
57409
Gene Name
MIF4G domain containing
Gene Aliases
AD023, MIFD, SLIP1
UniGene
Hs.325631
Gene Chromosome Location
Chr.17: 75266228-75271292 on Build GRCh38
Transcripts
NM_001242498.1 NM_001242500.1 NM_001242501.1 NM_020679.3 XM_005257530.1 XM_005257532.4 XM_005257533.1 XM_011525051.2 XM_011525052.2 XM_011525053.2 XM_011525054.2 XM_011525055.2 XM_011525057.2 XM_017024888.1 XM_017024889.1 XM_017024890.1 XM_017024891.1 XM_017024893.1 XM_017024894.1 XM_017024895.1 NM_015971.3

Gene Symbol
MRPS7
Entrez Gene ID
51081
Gene Name
mitochondrial ribosomal protein S7
Gene Aliases
MRP-S, MRP-S7, RP-S7, RPMS7, S7mt, bMRP27a
UniGene
Hs.71787
Gene Chromosome Location
Chr.17: 75261668-75266376 on Build GRCh38
Transcripts
NM_001242498.1 NM_001242500.1 NM_001242501.1 NM_020679.3 XM_005257530.1 XM_005257532.4 XM_005257533.1 XM_011525051.2 XM_011525052.2 XM_011525053.2 XM_011525054.2 XM_011525055.2 XM_011525057.2 XM_017024888.1 XM_017024889.1 XM_017024890.1 XM_017024891.1 XM_017024893.1 XM_017024894.1 XM_017024895.1 NM_015971.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4800150 c.676C>T p.Q226* Chr.17: 75266835-75266835 on Build GRCh38
2978776 c.729G>A p.T243T Chr.17: 75266782-75266782 on Build GRCh38
4610665 c.607C>T p.R203C Chr.17: 75266904-75266904 on Build GRCh38
3820536 c.628C>T p.L210L Chr.17: 75266883-75266883 on Build GRCh38
4962415 c.619C>T p.L207F Chr.17: 75266892-75266892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768357177 Chr.17: 75266729-75266729 on Build GRCh38
rs765637526 Chr.17: 75266856-75266856 on Build GRCh38
rs557030914 Chr.17: 75266834-75266834 on Build GRCh38
rs754208867 Chr.17: 75266754-75266754 on Build GRCh38
rs981005833 Chr.17: 75266498-75266498 on Build GRCh38
rs148919655 Chr.17: 75266632-75266632 on Build GRCh38
rs751054843 Chr.17: 75266876-75266876 on Build GRCh38
rs112122132 Chr.17: 75266796-75266796 on Build GRCh38
rs749177272 Chr.17: 75266805-75266805 on Build GRCh38
rs762468381 Chr.17: 75266755-75266755 on Build GRCh38
rs773538560 Chr.17: 75266695-75266695 on Build GRCh38
rs779661148 Chr.17: 75266771-75266771 on Build GRCh38
rs775612995 Chr.17: 75266813-75266813 on Build GRCh38
rs765846932 Chr.17: 75266874-75266874 on Build GRCh38
rs764473423 Chr.17: 75266694-75266694 on Build GRCh38
rs757993469 Chr.17: 75266770-75266770 on Build GRCh38
rs772174471 Chr.17: 75266806-75266806 on Build GRCh38
rs780889452 Chr.17: 75266879-75266879 on Build GRCh38
rs777689116 Chr.17: 75266804-75266804 on Build GRCh38
rs577459943 Chr.17: 75266453-75266453 on Build GRCh38
rs754627506 Chr.17: 75266783-75266783 on Build GRCh38
rs750895932 Chr.17: 75266758-75266758 on Build GRCh38
rs971705013 Chr.17: 75266473-75266473 on Build GRCh38
rs760860411 Chr.17: 75266814-75266814 on Build GRCh38
rs543600375 Chr.17: 75266593-75266593 on Build GRCh38
rs768716409 Chr.17: 75266395-75266395 on Build GRCh38
rs138799236 Chr.17: 75266750-75266750 on Build GRCh38
rs775601225 Chr.17: 75266749-75266749 on Build GRCh38
rs772315905 Chr.17: 75266692-75266692 on Build GRCh38
rs567359960 Chr.17: 75266824-75266824 on Build GRCh38
rs1028234337 Chr.17: 75266481-75266481 on Build GRCh38
rs16967764 Chr.17: 75266534-75266534 on Build GRCh38
rs763512473 Chr.17: 75266870-75266870 on Build GRCh38
rs901196573 Chr.17: 75266655-75266655 on Build GRCh38
rs201029754 Chr.17: 75266878-75266878 on Build GRCh38
rs1024006254 Chr.17: 75266392-75266392 on Build GRCh38
rs985352165 Chr.17: 75266877-75266877 on Build GRCh38
rs200088097 Chr.17: 75266745-75266745 on Build GRCh38
rs774469537 Chr.17: 75266744-75266744 on Build GRCh38
rs945452382 Chr.17: 75266552-75266552 on Build GRCh38
rs369706063 Chr.17: 75266737-75266737 on Build GRCh38
rs757751425 Chr.17: 75266710-75266710 on Build GRCh38
rs993641380 Chr.17: 75266478-75266478 on Build GRCh38
rs750988954 Chr.17: 75266868-75266868 on Build GRCh38
rs761080665 Chr.17: 75266700-75266700 on Build GRCh38
rs200570459 Chr.17: 75266812-75266812 on Build GRCh38
rs747959236 Chr.17: 75266797-75266797 on Build GRCh38
rs915293677 Chr.17: 75266566-75266566 on Build GRCh38
rs769693796 Chr.17: 75266740-75266740 on Build GRCh38
rs143021436 Chr.17: 75266802-75266802 on Build GRCh38
rs745360304 Chr.17: 75266716-75266716 on Build GRCh38
rs12547 Chr.17: 75266542-75266542 on Build GRCh38
rs932449116 Chr.17: 75266836-75266836 on Build GRCh38
rs762275880 Chr.17: 75266838-75266838 on Build GRCh38
rs553707414 Chr.17: 75266521-75266521 on Build GRCh38
rs557315618 Chr.17: 75266416-75266416 on Build GRCh38
rs765835897 Chr.17: 75266756-75266756 on Build GRCh38
rs925097767 Chr.17: 75266575-75266575 on Build GRCh38
rs546142468 Chr.17: 75266504-75266504 on Build GRCh38
rs373373697 Chr.17: 75266782-75266782 on Build GRCh38
rs563297780 Chr.17: 75266606-75266606 on Build GRCh38
rs536784078 Chr.17: 75266830-75266830 on Build GRCh38
rs201777064 Chr.17: 75266746-75266746 on Build GRCh38
rs756451507 Chr.17: 75266693-75266693 on Build GRCh38
rs926632741 Chr.17: 75266533-75266533 on Build GRCh38
rs778615584 Chr.17: 75266715-75266715 on Build GRCh38
rs530937069 Chr.17: 75266562-75266562 on Build GRCh38
rs779855479 Chr.17: 75266724-75266724 on Build GRCh38
rs776223754 Chr.17: 75266800-75266800 on Build GRCh38
rs932610169 Chr.17: 75266650-75266650 on Build GRCh38
rs746614263 Chr.17: 75266726-75266726 on Build GRCh38
rs573248821 Chr.17: 75266526-75266526 on Build GRCh38
rs768910500 Chr.17: 75266815-75266815 on Build GRCh38
rs992255203 Chr.17: 75266535-75266535 on Build GRCh38
rs138489085 Chr.17: 75266752-75266752 on Build GRCh38
rs952363136 Chr.17: 75266485-75266485 on Build GRCh38
rs115133838 Chr.17: 75266717-75266717 on Build GRCh38
rs9906887 Chr.17: 75266671-75266671 on Build GRCh38
rs999567628 Chr.17: 75266534-75266543 on Build GRCh38
rs753170485 Chr.17: 75266690-75266690 on Build GRCh38
rs145006252 Chr.17: 75266732-75266732 on Build GRCh38

View Full Product Details