Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75828684-75829182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC13D
Entrez Gene ID
201294
Gene Name
unc-13 homolog D
Gene Aliases
FHL3, HLH3, HPLH3, Munc13-4
UniGene
Hs.41045
Gene Chromosome Location
Chr.17: 75827225-75844717 on Build GRCh38
Transcripts
NM_199242.2 XM_011524504.1 XM_011524506.1 XM_011524507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
330356 c.2983G>C p.A995P Chr.17: 75828955-75828955 on Build GRCh38
361904 c.2962C>G p.P988A Chr.17: 75828976-75828976 on Build GRCh38
5795085 c.3067C>T p.R1023C Chr.17: 75828871-75828871 on Build GRCh38
6816912 c.2978G>A p.R993H Chr.17: 75828960-75828960 on Build GRCh38
6383859 c.3092C>A p.S1031Y Chr.17: 75828846-75828846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372034111 Chr.17: 75828860-75828860 on Build GRCh38
rs144730861 Chr.17: 75828905-75828905 on Build GRCh38
rs375153464 Chr.17: 75828876-75828876 on Build GRCh38
rs765680080 Chr.17: 75828861-75828861 on Build GRCh38
rs757164072 Chr.17: 75828919-75828919 on Build GRCh38
rs773314418 Chr.17: 75828820-75828820 on Build GRCh38
rs778000267 Chr.17: 75829033-75829033 on Build GRCh38
rs1040965765 Chr.17: 75829031-75829031 on Build GRCh38
rs750743318 Chr.17: 75828793-75828793 on Build GRCh38
rs1042162693 Chr.17: 75828749-75828749 on Build GRCh38
rs145549734 Chr.17: 75828966-75828966 on Build GRCh38
rs760466426 Chr.17: 75828987-75828987 on Build GRCh38
rs994195331 Chr.17: 75829045-75829045 on Build GRCh38
rs201791093 Chr.17: 75828988-75828988 on Build GRCh38
rs111279039 Chr.17: 75828890-75828890 on Build GRCh38
rs768608291 Chr.17: 75828858-75828858 on Build GRCh38
rs1002065078 Chr.17: 75829133-75829133 on Build GRCh38
rs749372765 Chr.17: 75828933-75828933 on Build GRCh38
rs186479769 Chr.17: 75829143-75829143 on Build GRCh38
rs897139999 Chr.17: 75829038-75829038 on Build GRCh38
rs368211297 Chr.17: 75828887-75828887 on Build GRCh38
rs762847068 Chr.17: 75828763-75828763 on Build GRCh38
rs200462004 Chr.17: 75828931-75828931 on Build GRCh38
rs776400339 Chr.17: 75828755-75828755 on Build GRCh38
rs777496354 Chr.17: 75828915-75828915 on Build GRCh38
rs753334662 Chr.17: 75828989-75828989 on Build GRCh38
rs918825395 Chr.17: 75828771-75828771 on Build GRCh38
rs267605050 Chr.17: 75828982-75828982 on Build GRCh38
rs765735106 Chr.17: 75828792-75828792 on Build GRCh38
rs944169057 Chr.17: 75828728-75828728 on Build GRCh38
rs140837214 Chr.17: 75828901-75828901 on Build GRCh38
rs553992292 Chr.17: 75828886-75828886 on Build GRCh38
rs764279026 Chr.17: 75828788-75828788 on Build GRCh38
rs1018586945 Chr.17: 75829057-75829057 on Build GRCh38
rs752215201 Chr.17: 75828872-75828872 on Build GRCh38
rs140599939 Chr.17: 75828871-75828871 on Build GRCh38
rs775309464 Chr.17: 75829028-75829028 on Build GRCh38
rs562357679 Chr.17: 75828914-75828914 on Build GRCh38
rs368577676 Chr.17: 75828854-75828854 on Build GRCh38
rs745623386 Chr.17: 75828925-75828925 on Build GRCh38
rs748182186 Chr.17: 75828757-75828757 on Build GRCh38
rs770849476 Chr.17: 75828884-75828884 on Build GRCh38
rs758204874 Chr.17: 75829001-75829001 on Build GRCh38
rs138760432 Chr.17: 75828955-75828955 on Build GRCh38
rs756737472 Chr.17: 75828990-75828990 on Build GRCh38
rs1027451422 Chr.17: 75829066-75829066 on Build GRCh38
rs754299432 Chr.17: 75828789-75828789 on Build GRCh38
rs897193421 Chr.17: 75828837-75828837 on Build GRCh38
rs201146973 Chr.17: 75828804-75828804 on Build GRCh38
rs758781229 Chr.17: 75828796-75828796 on Build GRCh38
rs936530349 Chr.17: 75828761-75828761 on Build GRCh38
rs866899109 Chr.17: 75828797-75828797 on Build GRCh38
rs756260132 Chr.17: 75829017-75829017 on Build GRCh38
rs140452589 Chr.17: 75828960-75828960 on Build GRCh38
rs1048876742 Chr.17: 75828791-75828791 on Build GRCh38
rs775862197 Chr.17: 75828920-75828920 on Build GRCh38
rs775803518 Chr.17: 75828768-75828768 on Build GRCh38
rs766833119 Chr.17: 75828866-75828866 on Build GRCh38
rs1026243253 Chr.17: 75829146-75829146 on Build GRCh38
rs759922688 Chr.17: 75828961-75828961 on Build GRCh38
rs761028266 Chr.17: 75828857-75828857 on Build GRCh38
rs888543002 Chr.17: 75829108-75829108 on Build GRCh38
rs769789829 Chr.17: 75828817-75828817 on Build GRCh38
rs781440659 Chr.17: 75828928-75828928 on Build GRCh38
rs142571012 Chr.17: 75829147-75829147 on Build GRCh38
rs749044099 Chr.17: 75828829-75828829 on Build GRCh38
rs755709094 Chr.17: 75828875-75828875 on Build GRCh38
rs753816739 Chr.17: 75828916-75828916 on Build GRCh38
rs369108124 Chr.17: 75829063-75829063 on Build GRCh38
rs773217124 Chr.17: 75828932-75828932 on Build GRCh38
rs780811495 Chr.17: 75828811-75828811 on Build GRCh38
rs1056144854 Chr.17: 75828716-75828716 on Build GRCh38
rs769897808 Chr.17: 75828899-75828899 on Build GRCh38
rs751157590 Chr.17: 75828870-75828870 on Build GRCh38
rs770898851 Chr.17: 75828839-75828839 on Build GRCh38
rs539119297 Chr.17: 75828707-75828707 on Build GRCh38
rs773086620 Chr.17: 75828760-75828760 on Build GRCh38
rs993056426 Chr.17: 75829157-75829157 on Build GRCh38
rs573990991 Chr.17: 75829037-75829037 on Build GRCh38
rs749282003 Chr.17: 75828879-75828879 on Build GRCh38
rs779654621 Chr.17: 75828798-75828798 on Build GRCh38
rs776703437 Chr.17: 75828859-75828859 on Build GRCh38
rs769696902 Chr.17: 75828758-75828758 on Build GRCh38
rs776737156 Chr.17: 75828889-75828889 on Build GRCh38
rs767809533 Chr.17: 75828767-75828767 on Build GRCh38
rs138462345 Chr.17: 75828997-75828997 on Build GRCh38
rs945093660 Chr.17: 75828734-75828734 on Build GRCh38
rs564875531 Chr.17: 75828965-75828965 on Build GRCh38
rs374478310 Chr.17: 75828816-75828816 on Build GRCh38
rs746714637 Chr.17: 75828803-75828803 on Build GRCh38
rs779709756 Chr.17: 75828752-75828752 on Build GRCh38
rs750974136 Chr.17: 75828863-75828863 on Build GRCh38
rs767150175 Chr.17: 75828908-75828908 on Build GRCh38
rs374946979 Chr.17: 75829035-75829035 on Build GRCh38
rs147126779 Chr.17: 75828910-75828910 on Build GRCh38
rs75988962 Chr.17: 75828705-75828705 on Build GRCh38
rs751745917 Chr.17: 75828752-75828753 on Build GRCh38
rs765196017 Chr.17: 75828991-75828991 on Build GRCh38
rs1041960684 Chr.17: 75828984-75828984 on Build GRCh38
rs778829355 Chr.17: 75828885-75828885 on Build GRCh38
rs768262286 Chr.17: 75828753-75828753 on Build GRCh38
rs760653269 Chr.17: 75828772-75828772 on Build GRCh38
rs917629207 Chr.17: 75828719-75828719 on Build GRCh38
rs201238621 Chr.17: 75828902-75828902 on Build GRCh38
rs889647946 Chr.17: 75828995-75828995 on Build GRCh38
rs755058502 Chr.17: 75828966-75828966 on Build GRCh38
rs763271797 Chr.17: 75828865-75828865 on Build GRCh38
rs757821324 Chr.17: 75828790-75828790 on Build GRCh38
rs775281177 Chr.17: 75828972-75828972 on Build GRCh38
rs371940934 Chr.17: 75828927-75828927 on Build GRCh38
rs573855825 Chr.17: 75828906-75828906 on Build GRCh38
rs746819458 Chr.17: 75829010-75829010 on Build GRCh38

View Full Product Details