Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9466435-9466948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF560
Entrez Gene ID
147741
Gene Name
zinc finger protein 560
Gene Aliases
-
UniGene
Hs.631613
Gene Chromosome Location
Chr.19: 9445710-9506692 on Build GRCh38
Transcripts
NM_152476.2 XM_011527696.2 XM_011527697.2 XM_017026327.1 XM_017026328.1 XM_017026329.1 XM_017026330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1398292 c.2191G>A p.V731M Chr.19: 9466756-9466756 on Build GRCh38
4083634 c.2062G>A p.A688T Chr.19: 9466885-9466885 on Build GRCh38
1004293 c.2105G>A p.R702H Chr.19: 9466842-9466842 on Build GRCh38
1711765 c.2069G>A p.G690E Chr.19: 9466878-9466878 on Build GRCh38
5391524 c.2076C>T p.S692S Chr.19: 9466871-9466871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746950168 Chr.19: 9466753-9466753 on Build GRCh38
rs201622607 Chr.19: 9466596-9466596 on Build GRCh38
rs150705230 Chr.19: 9466534-9466534 on Build GRCh38
rs954904212 Chr.19: 9466514-9466514 on Build GRCh38
rs745493105 Chr.19: 9466745-9466745 on Build GRCh38
rs367555706 Chr.19: 9466559-9466559 on Build GRCh38
rs767682995 Chr.19: 9466704-9466704 on Build GRCh38
rs538535192 Chr.19: 9466827-9466827 on Build GRCh38
rs750724915 Chr.19: 9466899-9466899 on Build GRCh38
rs915200628 Chr.19: 9466688-9466688 on Build GRCh38
rs747315246 Chr.19: 9466752-9466752 on Build GRCh38
rs983860255 Chr.19: 9466842-9466842 on Build GRCh38
rs577305020 Chr.19: 9466913-9466913 on Build GRCh38
rs889472261 Chr.19: 9466509-9466509 on Build GRCh38
rs998856217 Chr.19: 9466476-9466476 on Build GRCh38
rs148650284 Chr.19: 9466866-9466866 on Build GRCh38
rs781229059 Chr.19: 9466848-9466848 on Build GRCh38
rs757976367 Chr.19: 9466843-9466843 on Build GRCh38
rs372386086 Chr.19: 9466759-9466759 on Build GRCh38
rs533277895 Chr.19: 9466594-9466594 on Build GRCh38
rs763505290 Chr.19: 9466849-9466850 on Build GRCh38
rs200998976 Chr.19: 9466829-9466829 on Build GRCh38
rs781780971 Chr.19: 9466611-9466611 on Build GRCh38
rs763287809 Chr.19: 9466922-9466922 on Build GRCh38
rs201321286 Chr.19: 9466853-9466853 on Build GRCh38
rs372137495 Chr.19: 9466590-9466590 on Build GRCh38
rs760553564 Chr.19: 9466677-9466677 on Build GRCh38
rs529704393 Chr.19: 9466683-9466683 on Build GRCh38
rs748925173 Chr.19: 9466649-9466649 on Build GRCh38
rs557801059 Chr.19: 9466893-9466893 on Build GRCh38
rs149418712 Chr.19: 9466615-9466615 on Build GRCh38
rs771569778 Chr.19: 9466661-9466661 on Build GRCh38
rs767736370 Chr.19: 9466804-9466804 on Build GRCh38
rs57811230 Chr.19: 9466906-9466906 on Build GRCh38
rs771465196 Chr.19: 9466605-9466608 on Build GRCh38
rs74178149 Chr.19: 9466692-9466692 on Build GRCh38
rs1051340121 Chr.19: 9466503-9466503 on Build GRCh38
rs138449365 Chr.19: 9466659-9466659 on Build GRCh38
rs147160704 Chr.19: 9466810-9466810 on Build GRCh38
rs748660672 Chr.19: 9466491-9466491 on Build GRCh38
rs766084939 Chr.19: 9466923-9466923 on Build GRCh38
rs539161316 Chr.19: 9466885-9466885 on Build GRCh38
rs777608482 Chr.19: 9466526-9466526 on Build GRCh38
rs558878776 Chr.19: 9466625-9466625 on Build GRCh38
rs775449370 Chr.19: 9466665-9466665 on Build GRCh38
rs144027276 Chr.19: 9466730-9466730 on Build GRCh38
rs764371835 Chr.19: 9466717-9466717 on Build GRCh38
rs866489063 Chr.19: 9466658-9466658 on Build GRCh38
rs879184568 Chr.19: 9466758-9466758 on Build GRCh38
rs560763946 Chr.19: 9466551-9466551 on Build GRCh38
rs775843407 Chr.19: 9466882-9466882 on Build GRCh38
rs374358325 Chr.19: 9466831-9466831 on Build GRCh38
rs754299500 Chr.19: 9466814-9466814 on Build GRCh38
rs775396039 Chr.19: 9466576-9466576 on Build GRCh38
rs534354156 Chr.19: 9466699-9466699 on Build GRCh38
rs746578033 Chr.19: 9466845-9466845 on Build GRCh38
rs549580362 Chr.19: 9466789-9466789 on Build GRCh38
rs762181644 Chr.19: 9466895-9466895 on Build GRCh38
rs751597194 Chr.19: 9466599-9466599 on Build GRCh38
rs962455739 Chr.19: 9466580-9466580 on Build GRCh38
rs759782520 Chr.19: 9466803-9466803 on Build GRCh38
rs760535052 Chr.19: 9466587-9466587 on Build GRCh38
rs924132965 Chr.19: 9466731-9466731 on Build GRCh38
rs768519538 Chr.19: 9466787-9466787 on Build GRCh38
rs775645027 Chr.19: 9466806-9466806 on Build GRCh38
rs754440378 Chr.19: 9466846-9466846 on Build GRCh38
rs778696686 Chr.19: 9466725-9466725 on Build GRCh38
rs907970191 Chr.19: 9466545-9466545 on Build GRCh38
rs767301877 Chr.19: 9466877-9466877 on Build GRCh38
rs144007259 Chr.19: 9466867-9466867 on Build GRCh38
rs777098947 Chr.19: 9466757-9466758 on Build GRCh38
rs1029568033 Chr.19: 9466485-9466485 on Build GRCh38
rs756649233 Chr.19: 9466715-9466715 on Build GRCh38
rs766679978 Chr.19: 9466689-9466689 on Build GRCh38
rs768466093 Chr.19: 9466680-9466680 on Build GRCh38
rs754176051 Chr.19: 9466718-9466718 on Build GRCh38
rs773873340 Chr.19: 9466548-9466548 on Build GRCh38
rs765634075 Chr.19: 9466898-9466898 on Build GRCh38
rs140170249 Chr.19: 9466790-9466790 on Build GRCh38
rs890050684 Chr.19: 9466826-9466826 on Build GRCh38
rs267605822 Chr.19: 9466859-9466859 on Build GRCh38
rs529130376 Chr.19: 9466520-9466520 on Build GRCh38
rs976707070 Chr.19: 9466478-9466478 on Build GRCh38
rs779550008 Chr.19: 9466844-9466844 on Build GRCh38
rs150203893 Chr.19: 9466600-9466600 on Build GRCh38
rs562895336 Chr.19: 9466556-9466556 on Build GRCh38
rs761197445 Chr.19: 9466596-9466600 on Build GRCh38
rs757552228 Chr.19: 9466720-9466720 on Build GRCh38
rs762507991 Chr.19: 9466795-9466796 on Build GRCh38
rs753224746 Chr.19: 9466612-9466612 on Build GRCh38
rs540153983 Chr.19: 9466481-9466481 on Build GRCh38
rs368872787 Chr.19: 9466886-9466886 on Build GRCh38
rs147120143 Chr.19: 9466653-9466653 on Build GRCh38
rs770277479 Chr.19: 9466525-9466525 on Build GRCh38
rs369977723 Chr.19: 9466698-9466698 on Build GRCh38
rs368763535 Chr.19: 9466533-9466533 on Build GRCh38
rs762574098 Chr.19: 9466840-9466840 on Build GRCh38
rs745443805 Chr.19: 9466660-9466660 on Build GRCh38
rs375379880 Chr.19: 9466591-9466591 on Build GRCh38
rs1018035076 Chr.19: 9466713-9466713 on Build GRCh38
rs561681989 Chr.19: 9466497-9466497 on Build GRCh38
rs549467533 Chr.19: 9466788-9466788 on Build GRCh38
rs377042499 Chr.19: 9466684-9466684 on Build GRCh38

View Full Product Details