Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9700057-9700551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF812P
Entrez Gene ID
729648
Gene Name
zinc finger protein 812, pseudogene
Gene Aliases
ZNF812
UniGene
Hs.626848
Gene Chromosome Location
Chr.19: 9688855-9700817 on Build GRCh38
Transcripts

SNP Detail

SNP ID Chromosome Location
rs561143121 Chr.19: 9700318-9700318 on Build GRCh38
rs935979348 Chr.19: 9700309-9700309 on Build GRCh38
rs946139535 Chr.19: 9700149-9700149 on Build GRCh38
rs1027306829 Chr.19: 9700111-9700111 on Build GRCh38
rs577969846 Chr.19: 9700231-9700231 on Build GRCh38
rs1005088358 Chr.19: 9700390-9700390 on Build GRCh38
rs774952273 Chr.19: 9700486-9700486 on Build GRCh38
rs951732702 Chr.19: 9700118-9700118 on Build GRCh38
rs1033598537 Chr.19: 9700245-9700245 on Build GRCh38
rs969060510 Chr.19: 9700324-9700324 on Build GRCh38
rs371898911 Chr.19: 9700507-9700507 on Build GRCh38
rs912392666 Chr.19: 9700350-9700350 on Build GRCh38
rs571898595 Chr.19: 9700354-9700354 on Build GRCh38
rs572185612 Chr.19: 9700261-9700261 on Build GRCh38
rs73921106 Chr.19: 9700498-9700498 on Build GRCh38
rs77401981 Chr.19: 9700232-9700232 on Build GRCh38
rs974959889 Chr.19: 9700126-9700126 on Build GRCh38
rs960530759 Chr.19: 9700137-9700137 on Build GRCh38
rs897567129 Chr.19: 9700207-9700207 on Build GRCh38
rs957460661 Chr.19: 9700516-9700516 on Build GRCh38
rs1004722 Chr.19: 9700193-9700193 on Build GRCh38
rs1981835 Chr.19: 9700302-9700302 on Build GRCh38
rs532898940 Chr.19: 9700120-9700120 on Build GRCh38
rs550384627 Chr.19: 9700401-9700401 on Build GRCh38
rs10413676 Chr.19: 9700341-9700341 on Build GRCh38
rs866084998 Chr.19: 9700434-9700434 on Build GRCh38
rs1009489139 Chr.19: 9700089-9700089 on Build GRCh38
rs553734897 Chr.19: 9700504-9700504 on Build GRCh38
rs1040813733 Chr.19: 9700223-9700223 on Build GRCh38
rs565538827 Chr.19: 9700362-9700362 on Build GRCh38
rs566542771 Chr.19: 9700155-9700155 on Build GRCh38
rs551371759 Chr.19: 9700400-9700400 on Build GRCh38
rs1032619141 Chr.19: 9700512-9700512 on Build GRCh38
rs910798460 Chr.19: 9700322-9700322 on Build GRCh38
rs979056442 Chr.19: 9700433-9700433 on Build GRCh38
rs752600263 Chr.19: 9700212-9700212 on Build GRCh38
rs956644766 Chr.19: 9700436-9700436 on Build GRCh38
rs960891273 Chr.19: 9700396-9700396 on Build GRCh38
rs549248383 Chr.19: 9700452-9700452 on Build GRCh38
rs141319305 Chr.19: 9700491-9700491 on Build GRCh38
rs890583821 Chr.19: 9700211-9700219 on Build GRCh38
rs572247319 Chr.19: 9700506-9700506 on Build GRCh38
rs1008982870 Chr.19: 9700216-9700216 on Build GRCh38
rs1043649799 Chr.19: 9700198-9700198 on Build GRCh38
rs992194573 Chr.19: 9700186-9700186 on Build GRCh38
rs947999315 Chr.19: 9700197-9700197 on Build GRCh38
rs570591920 Chr.19: 9700475-9700475 on Build GRCh38
rs907602179 Chr.19: 9700181-9700181 on Build GRCh38
rs1018658277 Chr.19: 9700092-9700092 on Build GRCh38
rs944818481 Chr.19: 9700353-9700353 on Build GRCh38
rs1027808492 Chr.19: 9700127-9700127 on Build GRCh38
rs891836445 Chr.19: 9700208-9700208 on Build GRCh38
rs758233324 Chr.19: 9700217-9700217 on Build GRCh38
rs960777551 Chr.19: 9700264-9700264 on Build GRCh38
rs551200579 Chr.19: 9700142-9700142 on Build GRCh38
rs976271028 Chr.19: 9700458-9700458 on Build GRCh38
rs750647170 Chr.19: 9700110-9700110 on Build GRCh38
rs1001867976 Chr.19: 9700364-9700364 on Build GRCh38
rs983182896 Chr.19: 9700143-9700143 on Build GRCh38
rs1009281530 Chr.19: 9700091-9700091 on Build GRCh38
rs534003042 Chr.19: 9700183-9700183 on Build GRCh38
rs375436038 Chr.19: 9700211-9700211 on Build GRCh38
rs567355950 Chr.19: 9700487-9700487 on Build GRCh38
rs768469940 Chr.19: 9700499-9700499 on Build GRCh38
rs1004721 Chr.19: 9700195-9700195 on Build GRCh38
rs556085267 Chr.19: 9700203-9700203 on Build GRCh38
rs998685 Chr.19: 9700188-9700188 on Build GRCh38
rs969054379 Chr.19: 9700432-9700432 on Build GRCh38
rs10415262 Chr.19: 9700349-9700349 on Build GRCh38
rs931985608 Chr.19: 9700469-9700469 on Build GRCh38
rs912410094 Chr.19: 9700450-9700450 on Build GRCh38
rs966550075 Chr.19: 9700267-9700267 on Build GRCh38
rs1049088391 Chr.19: 9700470-9700470 on Build GRCh38
rs768994858 Chr.19: 9700379-9700379 on Build GRCh38
rs1015541605 Chr.19: 9700394-9700394 on Build GRCh38
rs926035892 Chr.19: 9700303-9700303 on Build GRCh38
rs925160537 Chr.19: 9700333-9700333 on Build GRCh38
rs1049309615 Chr.19: 9700371-9700371 on Build GRCh38
rs998684 Chr.19: 9700252-9700252 on Build GRCh38
rs538007087 Chr.19: 9700490-9700490 on Build GRCh38
rs986576392 Chr.19: 9700311-9700311 on Build GRCh38
rs767684575 Chr.19: 9700107-9700107 on Build GRCh38
rs527369432 Chr.19: 9700447-9700447 on Build GRCh38
rs1008034798 Chr.19: 9700531-9700531 on Build GRCh38
rs1053205581 Chr.19: 9700206-9700206 on Build GRCh38
rs896382873 Chr.19: 9700377-9700377 on Build GRCh38
rs1004972005 Chr.19: 9700253-9700253 on Build GRCh38
rs532809869 Chr.19: 9700395-9700395 on Build GRCh38
rs775687296 Chr.19: 9700356-9700356 on Build GRCh38
rs904984166 Chr.19: 9700525-9700525 on Build GRCh38
rs979360666 Chr.19: 9700325-9700325 on Build GRCh38
rs900489896 Chr.19: 9700224-9700224 on Build GRCh38
rs916259682 Chr.19: 9700323-9700323 on Build GRCh38
rs939038288 Chr.19: 9700184-9700184 on Build GRCh38
rs1027840993 Chr.19: 9700234-9700234 on Build GRCh38
rs952196722 Chr.19: 9700141-9700141 on Build GRCh38
rs376476210 Chr.19: 9700448-9700448 on Build GRCh38
rs756835104 Chr.19: 9700336-9700336 on Build GRCh38
rs977399600 Chr.19: 9700171-9700171 on Build GRCh38
rs537749005 Chr.19: 9700204-9700204 on Build GRCh38
rs556378301 Chr.19: 9700205-9700205 on Build GRCh38
rs769040297 Chr.19: 9700451-9700451 on Build GRCh38
rs571492427 Chr.19: 9700492-9700492 on Build GRCh38
rs966502276 Chr.19: 9700415-9700415 on Build GRCh38
rs922006686 Chr.19: 9700467-9700467 on Build GRCh38
rs1040873577 Chr.19: 9700466-9700466 on Build GRCh38
rs1013774405 Chr.19: 9700397-9700397 on Build GRCh38

View Full Product Details