Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11106501-11106766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLR
Entrez Gene ID
3949
Gene Name
low density lipoprotein receptor
Gene Aliases
FH, FHC, LDLCQ2
UniGene
Hs.213289
Gene Chromosome Location
Chr.19: 11089362-11133830 on Build GRCh38
Transcripts
NM_000527.4 NM_001195798.1 NM_001195799.1 NM_001195800.1 NM_001195803.1 XM_011528010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5498316 c.769C>T p.R257W Chr.19: 11106639-11106639 on Build GRCh38
1390293 c.772G>A p.E258K Chr.19: 11106642-11106642 on Build GRCh38
4887983 c.810C>A p.C270* Chr.19: 11106680-11106680 on Build GRCh38
990777 c.718G>A p.E240K Chr.19: 11106588-11106588 on Build GRCh38
4642242 c.745A>G p.I249V Chr.19: 11106615-11106615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879254673 Chr.19: 11106646-11106647 on Build GRCh38
rs879254678 Chr.19: 11106667-11106667 on Build GRCh38
rs879254652 Chr.19: 11106564-11106564 on Build GRCh38
rs765565329 Chr.19: 11106721-11106721 on Build GRCh38
rs879254668 Chr.19: 11106633-11106633 on Build GRCh38
rs776223298 Chr.19: 11106590-11106590 on Build GRCh38
rs769808108 Chr.19: 11106606-11106606 on Build GRCh38
rs879254655 Chr.19: 11106576-11106576 on Build GRCh38
rs879254663 Chr.19: 11106613-11106613 on Build GRCh38
rs879254657 Chr.19: 11106579-11106579 on Build GRCh38
rs879254653 Chr.19: 11106573-11106574 on Build GRCh38
rs879254684 Chr.19: 11106684-11106687 on Build GRCh38
rs899246224 Chr.19: 11106730-11106730 on Build GRCh38
rs730882087 Chr.19: 11106640-11106641 on Build GRCh38
rs375163928 Chr.19: 11106696-11106696 on Build GRCh38
rs879254658 Chr.19: 11106594-11106594 on Build GRCh38
rs879254654 Chr.19: 11106575-11106576 on Build GRCh38
rs773672332 Chr.19: 11106693-11106693 on Build GRCh38
rs879254666 Chr.19: 11106622-11106623 on Build GRCh38
rs879254656 Chr.19: 11106578-11106579 on Build GRCh38
rs879254674 Chr.19: 11106651-11106651 on Build GRCh38
rs757635995 Chr.19: 11106545-11106545 on Build GRCh38
rs201875602 Chr.19: 11106701-11106701 on Build GRCh38
rs150673992 Chr.19: 11106627-11106627 on Build GRCh38
rs750900506 Chr.19: 11106657-11106657 on Build GRCh38
rs879254685 Chr.19: 11106688-11106688 on Build GRCh38
rs201041358 Chr.19: 11106567-11106567 on Build GRCh38
rs1037574172 Chr.19: 11106727-11106727 on Build GRCh38
rs772882099 Chr.19: 11106621-11106621 on Build GRCh38
rs148171426 Chr.19: 11106580-11106580 on Build GRCh38
rs747855640 Chr.19: 11106674-11106674 on Build GRCh38
rs879254675 Chr.19: 11106660-11106663 on Build GRCh38
rs146181232 Chr.19: 11106665-11106665 on Build GRCh38
rs780294768 Chr.19: 11106581-11106581 on Build GRCh38
rs780093891 Chr.19: 11106661-11106661 on Build GRCh38
rs879254660 Chr.19: 11106607-11106607 on Build GRCh38
rs879254683 Chr.19: 11106679-11106679 on Build GRCh38
rs879254670 Chr.19: 11106637-11106637 on Build GRCh38
rs764526789 Chr.19: 11106632-11106632 on Build GRCh38
rs879254650 Chr.19: 11106552-11106552 on Build GRCh38
rs773328511 Chr.19: 11106680-11106680 on Build GRCh38
rs879254659 Chr.19: 11106597-11106597 on Build GRCh38
rs750986321 Chr.19: 11106735-11106735 on Build GRCh38
rs567958343 Chr.19: 11106700-11106700 on Build GRCh38
rs141681167 Chr.19: 11106658-11106658 on Build GRCh38
rs572275000 Chr.19: 11106694-11106694 on Build GRCh38
rs875989907 Chr.19: 11106666-11106666 on Build GRCh38
rs778975482 Chr.19: 11106555-11106555 on Build GRCh38
rs879254679 Chr.19: 11106669-11106669 on Build GRCh38
rs879254676 Chr.19: 11106663-11106663 on Build GRCh38
rs768563000 Chr.19: 11106588-11106588 on Build GRCh38
rs376963526 Chr.19: 11106582-11106582 on Build GRCh38
rs200990725 Chr.19: 11106639-11106639 on Build GRCh38
rs121908040 Chr.19: 11106652-11106652 on Build GRCh38
rs143992984 Chr.19: 11106676-11106676 on Build GRCh38
rs762588484 Chr.19: 11106626-11106626 on Build GRCh38
rs141929940 Chr.19: 11106620-11106620 on Build GRCh38
rs749220643 Chr.19: 11106681-11106681 on Build GRCh38
rs879254681 Chr.19: 11106671-11106671 on Build GRCh38
rs730882088 Chr.19: 11106660-11106660 on Build GRCh38
rs748131256 Chr.19: 11106592-11106592 on Build GRCh38
rs759109699 Chr.19: 11106630-11106630 on Build GRCh38
rs879254661 Chr.19: 11106607-11106607 on Build GRCh38
rs879254671 Chr.19: 11106638-11106638 on Build GRCh38
rs770636985 Chr.19: 11106692-11106692 on Build GRCh38
rs139507589 Chr.19: 11106628-11106628 on Build GRCh38
rs879254680 Chr.19: 11106670-11106670 on Build GRCh38
rs754656393 Chr.19: 11106537-11106537 on Build GRCh38
rs879254667 Chr.19: 11106631-11106631 on Build GRCh38
rs139043155 Chr.19: 11106668-11106668 on Build GRCh38
rs879254662 Chr.19: 11106612-11106612 on Build GRCh38
rs879254672 Chr.19: 11106642-11106642 on Build GRCh38
rs879254664 Chr.19: 11106614-11106614 on Build GRCh38
rs879254682 Chr.19: 11106678-11106678 on Build GRCh38
rs879254651 Chr.19: 11106559-11106568 on Build GRCh38
rs879254677 Chr.19: 11106666-11106669 on Build GRCh38
rs202213054 Chr.19: 11106685-11106685 on Build GRCh38
rs531199430 Chr.19: 11106705-11106705 on Build GRCh38
rs879254665 Chr.19: 11106616-11106616 on Build GRCh38
rs879254669 Chr.19: 11106634-11106634 on Build GRCh38
rs781061095 Chr.19: 11106672-11106672 on Build GRCh38
rs745897889 Chr.19: 11106562-11106562 on Build GRCh38
rs879254686 Chr.19: 11106689-11106689 on Build GRCh38
rs757808215 Chr.19: 11106640-11106640 on Build GRCh38

View Full Product Details