Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11165055-11165564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK2
Entrez Gene ID
25959
Gene Name
KN motif and ankyrin repeat domains 2
Gene Aliases
ANKRD25, MXRA3, PPKWH, SIP
UniGene
Hs.284208
Gene Chromosome Location
Chr.19: 11164267-11197567 on Build GRCh38
Transcripts
NM_001136191.2 NM_015493.6

SNP Detail

SNP ID Chromosome Location
rs777212205 Chr.19: 11165199-11165199 on Build GRCh38
rs114268284 Chr.19: 11165279-11165279 on Build GRCh38
rs955600166 Chr.19: 11165305-11165305 on Build GRCh38
rs1003654378 Chr.19: 11165415-11165415 on Build GRCh38
rs765363913 Chr.19: 11165250-11165250 on Build GRCh38
rs975871613 Chr.19: 11165285-11165285 on Build GRCh38
rs750226123 Chr.19: 11165306-11165306 on Build GRCh38
rs925669843 Chr.19: 11165267-11165267 on Build GRCh38
rs116850924 Chr.19: 11165284-11165284 on Build GRCh38
rs75254697 Chr.19: 11165090-11165090 on Build GRCh38
rs796989406 Chr.19: 11165137-11165138 on Build GRCh38
rs543396003 Chr.19: 11165299-11165299 on Build GRCh38
rs78649129 Chr.19: 11165138-11165138 on Build GRCh38
rs941576229 Chr.19: 11165380-11165380 on Build GRCh38
rs779975715 Chr.19: 11165352-11165352 on Build GRCh38
rs975460561 Chr.19: 11165084-11165084 on Build GRCh38
rs143366219 Chr.19: 11165085-11165085 on Build GRCh38
rs796703897 Chr.19: 11165138-11165139 on Build GRCh38
rs370184830 Chr.19: 11165173-11165173 on Build GRCh38
rs759701944 Chr.19: 11165224-11165224 on Build GRCh38
rs555344581 Chr.19: 11165125-11165125 on Build GRCh38
rs566498783 Chr.19: 11165315-11165315 on Build GRCh38
rs955771153 Chr.19: 11165409-11165409 on Build GRCh38
rs898939440 Chr.19: 11165158-11165158 on Build GRCh38
rs866321522 Chr.19: 11165438-11165438 on Build GRCh38
rs545615672 Chr.19: 11165170-11165170 on Build GRCh38
rs186455621 Chr.19: 11165393-11165393 on Build GRCh38
rs1009247176 Chr.19: 11165146-11165146 on Build GRCh38
rs771561160 Chr.19: 11165189-11165189 on Build GRCh38
rs537420934 Chr.19: 11165442-11165442 on Build GRCh38
rs908793879 Chr.19: 11165137-11165140 on Build GRCh38
rs181480419 Chr.19: 11165281-11165281 on Build GRCh38
rs551168304 Chr.19: 11165300-11165300 on Build GRCh38
rs796803361 Chr.19: 11165336-11165336 on Build GRCh38
rs16423 Chr.19: 11165136-11165137 on Build GRCh38
rs534114673 Chr.19: 11165365-11165365 on Build GRCh38
rs1012495218 Chr.19: 11165370-11165370 on Build GRCh38
rs116256300 Chr.19: 11165388-11165388 on Build GRCh38
rs561358918 Chr.19: 11165266-11165266 on Build GRCh38
rs1005397397 Chr.19: 11165335-11165335 on Build GRCh38
rs934422405 Chr.19: 11165272-11165272 on Build GRCh38
rs150866270 Chr.19: 11165165-11165165 on Build GRCh38
rs573529252 Chr.19: 11165137-11165137 on Build GRCh38
rs1017418380 Chr.19: 11165155-11165155 on Build GRCh38
rs568756661 Chr.19: 11165419-11165419 on Build GRCh38
rs563382411 Chr.19: 11165539-11165539 on Build GRCh38
rs892113 Chr.19: 11165258-11165258 on Build GRCh38
rs1042819221 Chr.19: 11165078-11165078 on Build GRCh38
rs552151110 Chr.19: 11165191-11165191 on Build GRCh38
rs916744035 Chr.19: 11165246-11165246 on Build GRCh38
rs574507658 Chr.19: 11165205-11165205 on Build GRCh38
rs754513860 Chr.19: 11165485-11165485 on Build GRCh38
rs988995855 Chr.19: 11165484-11165484 on Build GRCh38
rs1010752704 Chr.19: 11165494-11165494 on Build GRCh38
rs3041983 Chr.19: 11165137-11165138 on Build GRCh38
rs3185010 Chr.19: 11165166-11165166 on Build GRCh38
rs746486819 Chr.19: 11165460-11165461 on Build GRCh38
rs886495353 Chr.19: 11165326-11165326 on Build GRCh38
rs556341115 Chr.19: 11165450-11165450 on Build GRCh38
rs979539803 Chr.19: 11165394-11165394 on Build GRCh38
rs534707421 Chr.19: 11165333-11165333 on Build GRCh38
rs950781604 Chr.19: 11165178-11165178 on Build GRCh38
rs528000214 Chr.19: 11165206-11165206 on Build GRCh38
rs1045173811 Chr.19: 11165401-11165401 on Build GRCh38
rs1039396002 Chr.19: 11165288-11165288 on Build GRCh38
rs1022093698 Chr.19: 11165500-11165500 on Build GRCh38
rs112053149 Chr.19: 11165395-11165395 on Build GRCh38
rs1035011571 Chr.19: 11165349-11165349 on Build GRCh38
rs991504821 Chr.19: 11165245-11165245 on Build GRCh38
rs1053204449 Chr.19: 11165119-11165119 on Build GRCh38

View Full Product Details