Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36605133-36605623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF382
Entrez Gene ID
84911
Gene Name
zinc finger protein 382
Gene Aliases
KS1
UniGene
Hs.631591
Gene Chromosome Location
Chr.19: 36605305-36628597 on Build GRCh38
Transcripts
NM_001256838.1 NM_032825.4 NM_001145649.1 NM_001145650.1 NR_027239.1 XM_011527164.2 XM_017027039.1 XM_017027042.1 XM_017027043.1

Gene Symbol
ZNF529
Entrez Gene ID
57711
Gene Name
zinc finger protein 529
Gene Aliases
-
UniGene
Hs.654960
Gene Chromosome Location
Chr.19: 36534491-36605411 on Build GRCh38
Transcripts
NM_001256838.1 NM_032825.4 NM_001145649.1 NM_001145650.1 NR_027239.1 XM_011527164.2 XM_017027039.1 XM_017027042.1 XM_017027043.1

SNP Detail

SNP ID Chromosome Location
rs986292879 Chr.19: 36605414-36605414 on Build GRCh38
rs977195747 Chr.19: 36605251-36605251 on Build GRCh38
rs111985242 Chr.19: 36605501-36605501 on Build GRCh38
rs1023715158 Chr.19: 36605177-36605177 on Build GRCh38
rs925751623 Chr.19: 36605309-36605309 on Build GRCh38
rs949729169 Chr.19: 36605359-36605359 on Build GRCh38
rs897504860 Chr.19: 36605408-36605408 on Build GRCh38
rs956917495 Chr.19: 36605205-36605205 on Build GRCh38
rs576089087 Chr.19: 36605410-36605410 on Build GRCh38
rs547580336 Chr.19: 36605538-36605538 on Build GRCh38
rs111647441 Chr.19: 36605444-36605444 on Build GRCh38
rs114196412 Chr.19: 36605255-36605255 on Build GRCh38
rs998183143 Chr.19: 36605514-36605514 on Build GRCh38
rs994337348 Chr.19: 36605199-36605199 on Build GRCh38
rs551966130 Chr.19: 36605227-36605227 on Build GRCh38
rs892462764 Chr.19: 36605380-36605380 on Build GRCh38
rs951021242 Chr.19: 36605516-36605516 on Build GRCh38
rs978635572 Chr.19: 36605586-36605586 on Build GRCh38
rs148516258 Chr.19: 36605592-36605592 on Build GRCh38
rs910956991 Chr.19: 36605237-36605237 on Build GRCh38
rs1007044750 Chr.19: 36605468-36605468 on Build GRCh38
rs888317070 Chr.19: 36605293-36605293 on Build GRCh38
rs559248830 Chr.19: 36605559-36605559 on Build GRCh38
rs3108603 Chr.19: 36605171-36605171 on Build GRCh38
rs1006892457 Chr.19: 36605226-36605226 on Build GRCh38
rs1018319857 Chr.19: 36605229-36605229 on Build GRCh38
rs1044869078 Chr.19: 36605572-36605572 on Build GRCh38
rs1018309714 Chr.19: 36605492-36605492 on Build GRCh38
rs1048631500 Chr.19: 36605439-36605439 on Build GRCh38
rs543582821 Chr.19: 36605435-36605435 on Build GRCh38
rs953696392 Chr.19: 36605412-36605412 on Build GRCh38
rs1003960312 Chr.19: 36605351-36605351 on Build GRCh38
rs1045572978 Chr.19: 36605331-36605331 on Build GRCh38
rs1020676525 Chr.19: 36605517-36605517 on Build GRCh38
rs142266051 Chr.19: 36605405-36605405 on Build GRCh38
rs1027083378 Chr.19: 36605201-36605201 on Build GRCh38
rs1018454937 Chr.19: 36605418-36605418 on Build GRCh38
rs989609007 Chr.19: 36605209-36605209 on Build GRCh38
rs1030920007 Chr.19: 36605360-36605360 on Build GRCh38
rs888537265 Chr.19: 36605167-36605167 on Build GRCh38
rs909785237 Chr.19: 36605545-36605545 on Build GRCh38
rs917043521 Chr.19: 36605341-36605341 on Build GRCh38
rs928039197 Chr.19: 36605575-36605578 on Build GRCh38
rs953564289 Chr.19: 36605228-36605228 on Build GRCh38
rs1055546276 Chr.19: 36605330-36605330 on Build GRCh38
rs559094141 Chr.19: 36605303-36605303 on Build GRCh38
rs1022457970 Chr.19: 36605222-36605222 on Build GRCh38
rs978688054 Chr.19: 36605291-36605291 on Build GRCh38
rs937159083 Chr.19: 36605325-36605325 on Build GRCh38
rs952902793 Chr.19: 36605220-36605220 on Build GRCh38
rs1048341764 Chr.19: 36605276-36605276 on Build GRCh38
rs751164479 Chr.19: 36605254-36605254 on Build GRCh38
rs189034696 Chr.19: 36605246-36605246 on Build GRCh38
rs929841087 Chr.19: 36605270-36605270 on Build GRCh38
rs984068541 Chr.19: 36605530-36605530 on Build GRCh38
rs947902213 Chr.19: 36605300-36605300 on Build GRCh38

View Full Product Details